Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129638491-129638993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB6
Entrez Gene ID
140459
Gene Name
ankyrin repeat and SOCS box containing 6
Gene Aliases
-
UniGene
Hs.125037
Gene Chromosome Location
Chr.9: 129634604-129642169 on Build GRCh38
Transcripts
NM_001202403.1 NM_017873.3 NM_177999.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3328452 c.580C>T p.R194C Chr.9: 129638591-129638591 on Build GRCh38
4657308 c.550G>A p.G184R Chr.9: 129638621-129638621 on Build GRCh38
3904717 c.563A>G p.H188R Chr.9: 129638608-129638608 on Build GRCh38
1755992 c.591G>A p.L197L Chr.9: 129638580-129638580 on Build GRCh38
1105865 c.546C>T p.S182S Chr.9: 129638625-129638625 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761008614 Chr.9: 129638600-129638600 on Build GRCh38
rs562818783 Chr.9: 129638804-129638804 on Build GRCh38
rs972054367 Chr.9: 129638863-129638863 on Build GRCh38
rs1021721583 Chr.9: 129638948-129638949 on Build GRCh38
rs755563337 Chr.9: 129638534-129638534 on Build GRCh38
rs370698648 Chr.9: 129638676-129638677 on Build GRCh38
rs572062485 Chr.9: 129638686-129638686 on Build GRCh38
rs753926985 Chr.9: 129638533-129638533 on Build GRCh38
rs376199179 Chr.9: 129638546-129638546 on Build GRCh38
rs1033594097 Chr.9: 129638834-129638834 on Build GRCh38
rs147281908 Chr.9: 129638748-129638749 on Build GRCh38
rs540721375 Chr.9: 129638594-129638594 on Build GRCh38
rs368515357 Chr.9: 129638653-129638653 on Build GRCh38
rs140484737 Chr.9: 129638559-129638559 on Build GRCh38
rs967611330 Chr.9: 129638724-129638724 on Build GRCh38
rs769461838 Chr.9: 129638639-129638639 on Build GRCh38
rs567346123 Chr.9: 129638514-129638514 on Build GRCh38
rs1000475577 Chr.9: 129638833-129638833 on Build GRCh38
rs774218305 Chr.9: 129638590-129638590 on Build GRCh38
rs1011206339 Chr.9: 129638933-129638933 on Build GRCh38
rs542845915 Chr.9: 129638777-129638777 on Build GRCh38
rs965996037 Chr.9: 129638859-129638859 on Build GRCh38
rs937082442 Chr.9: 129638761-129638761 on Build GRCh38
rs768105373 Chr.9: 129638663-129638663 on Build GRCh38
rs759044711 Chr.9: 129638680-129638682 on Build GRCh38
rs755041021 Chr.9: 129638621-129638621 on Build GRCh38
rs894218147 Chr.9: 129638884-129638884 on Build GRCh38
rs79562490 Chr.9: 129638661-129638661 on Build GRCh38
rs766657921 Chr.9: 129638604-129638604 on Build GRCh38
rs895447988 Chr.9: 129638797-129638797 on Build GRCh38
rs576432709 Chr.9: 129638547-129638547 on Build GRCh38
rs561081730 Chr.9: 129638618-129638618 on Build GRCh38
rs919286759 Chr.9: 129638866-129638866 on Build GRCh38
rs761792727 Chr.9: 129638591-129638591 on Build GRCh38
rs953606490 Chr.9: 129638840-129638840 on Build GRCh38
rs966027030 Chr.9: 129638707-129638707 on Build GRCh38
rs200288317 Chr.9: 129638535-129638535 on Build GRCh38
rs942004469 Chr.9: 129638792-129638792 on Build GRCh38
rs888296 Chr.9: 129638810-129638810 on Build GRCh38
rs895521462 Chr.9: 129638516-129638516 on Build GRCh38
rs112162300 Chr.9: 129638730-129638730 on Build GRCh38
rs773162470 Chr.9: 129638685-129638685 on Build GRCh38
rs199744552 Chr.9: 129638561-129638561 on Build GRCh38
rs778187823 Chr.9: 129638557-129638557 on Build GRCh38
rs909697372 Chr.9: 129638923-129638923 on Build GRCh38
rs747336110 Chr.9: 129638637-129638637 on Build GRCh38
rs1034410622 Chr.9: 129638513-129638513 on Build GRCh38
rs565089667 Chr.9: 129638851-129638851 on Build GRCh38
rs192560674 Chr.9: 129638729-129638729 on Build GRCh38
rs78799521 Chr.9: 129638662-129638662 on Build GRCh38
rs1001318077 Chr.9: 129638512-129638512 on Build GRCh38
rs1055581630 Chr.9: 129638770-129638770 on Build GRCh38
rs375226911 Chr.9: 129638644-129638644 on Build GRCh38
rs79636483 Chr.9: 129638652-129638652 on Build GRCh38
rs753202866 Chr.9: 129638624-129638624 on Build GRCh38
rs73670149 Chr.9: 129638850-129638850 on Build GRCh38
rs909739121 Chr.9: 129638752-129638752 on Build GRCh38
rs777630143 Chr.9: 129638922-129638922 on Build GRCh38
rs543684828 Chr.9: 129638907-129638907 on Build GRCh38
rs918523848 Chr.9: 129638717-129638717 on Build GRCh38
rs753418834 Chr.9: 129638726-129638726 on Build GRCh38
rs555168474 Chr.9: 129638527-129638527 on Build GRCh38
rs372266774 Chr.9: 129638625-129638625 on Build GRCh38
rs772004088 Chr.9: 129638678-129638678 on Build GRCh38
rs976601521 Chr.9: 129638709-129638709 on Build GRCh38
rs756379197 Chr.9: 129638526-129638526 on Build GRCh38
rs765192770 Chr.9: 129638640-129638640 on Build GRCh38
rs903423622 Chr.9: 129638824-129638824 on Build GRCh38
rs772014245 Chr.9: 129638861-129638861 on Build GRCh38
rs773885645 Chr.9: 129638667-129638667 on Build GRCh38
rs779046182 Chr.9: 129638622-129638622 on Build GRCh38
rs1002646306 Chr.9: 129638599-129638599 on Build GRCh38
rs778041755 Chr.9: 129638626-129638626 on Build GRCh38
rs1050505793 Chr.9: 129638753-129638753 on Build GRCh38
rs951353062 Chr.9: 129638898-129638898 on Build GRCh38
rs572015845 Chr.9: 129638568-129638568 on Build GRCh38
rs370190875 Chr.9: 129638524-129638524 on Build GRCh38
rs1034141503 Chr.9: 129638867-129638867 on Build GRCh38
rs994102616 Chr.9: 129638849-129638849 on Build GRCh38
rs1013931094 Chr.9: 129638799-129638799 on Build GRCh38
rs775015210 Chr.9: 129638560-129638560 on Build GRCh38
rs765351752 Chr.9: 129638538-129638539 on Build GRCh38
rs760487310 Chr.9: 129638691-129638691 on Build GRCh38
rs138905769 Chr.9: 129638673-129638674 on Build GRCh38
rs761693106 Chr.9: 129638675-129638675 on Build GRCh38

View Full Product Details