Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129888803-129889291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNBP1
Entrez Gene ID
23048
Gene Name
formin binding protein 1
Gene Aliases
FBP17
UniGene
Hs.189409
Gene Chromosome Location
Chr.9: 129887187-130053846 on Build GRCh38
Transcripts
NM_015033.2 XM_005251824.2 XM_005251826.2 XM_005251834.2 XM_011518401.1 XM_011518402.1 XM_017014490.1 XM_017014491.1 XM_017014494.1

SNP Detail

SNP ID Chromosome Location
rs1051779478 Chr.9: 129889073-129889073 on Build GRCh38
rs111766024 Chr.9: 129889084-129889084 on Build GRCh38
rs191412529 Chr.9: 129889258-129889258 on Build GRCh38
rs995311036 Chr.9: 129889087-129889087 on Build GRCh38
rs937465432 Chr.9: 129889096-129889096 on Build GRCh38
rs973819039 Chr.9: 129888895-129888895 on Build GRCh38
rs1039165142 Chr.9: 129889040-129889040 on Build GRCh38
rs1003004776 Chr.9: 129889090-129889090 on Build GRCh38
rs371735587 Chr.9: 129888858-129888858 on Build GRCh38
rs560605018 Chr.9: 129889209-129889209 on Build GRCh38
rs755535128 Chr.9: 129888898-129888898 on Build GRCh38
rs533409555 Chr.9: 129889076-129889076 on Build GRCh38
rs944397187 Chr.9: 129889097-129889097 on Build GRCh38
rs947517302 Chr.9: 129889037-129889037 on Build GRCh38
rs893341118 Chr.9: 129889168-129889168 on Build GRCh38
rs955897623 Chr.9: 129889161-129889161 on Build GRCh38
rs576228523 Chr.9: 129888985-129888985 on Build GRCh38
rs997125363 Chr.9: 129889273-129889273 on Build GRCh38
rs901488264 Chr.9: 129889268-129889268 on Build GRCh38
rs879074331 Chr.9: 129889086-129889086 on Build GRCh38
rs1004170897 Chr.9: 129889077-129889077 on Build GRCh38
rs891063083 Chr.9: 129888849-129888849 on Build GRCh38
rs544535574 Chr.9: 129888938-129888938 on Build GRCh38
rs989458573 Chr.9: 129888991-129888991 on Build GRCh38
rs1010484477 Chr.9: 129889079-129889081 on Build GRCh38
rs556094679 Chr.9: 129888965-129888965 on Build GRCh38
rs968931503 Chr.9: 129889220-129889220 on Build GRCh38
rs779449614 Chr.9: 129889090-129889094 on Build GRCh38
rs914781634 Chr.9: 129888996-129888996 on Build GRCh38
rs969753282 Chr.9: 129888919-129888919 on Build GRCh38
rs748605910 Chr.9: 129889216-129889216 on Build GRCh38
rs1033210230 Chr.9: 129888993-129888993 on Build GRCh38
rs756319198 Chr.9: 129888891-129888891 on Build GRCh38
rs150156540 Chr.9: 129889201-129889201 on Build GRCh38
rs1022658638 Chr.9: 129888900-129888900 on Build GRCh38
rs989101953 Chr.9: 129889190-129889190 on Build GRCh38
rs753987401 Chr.9: 129889082-129889086 on Build GRCh38
rs984695398 Chr.9: 129888830-129888830 on Build GRCh38
rs375504927 Chr.9: 129889078-129889078 on Build GRCh38
rs34975006 Chr.9: 129889083-129889084 on Build GRCh38
rs1015462602 Chr.9: 129888870-129888870 on Build GRCh38
rs1037030549 Chr.9: 129888867-129888867 on Build GRCh38
rs1052279153 Chr.9: 129889259-129889259 on Build GRCh38
rs1022727496 Chr.9: 129889088-129889088 on Build GRCh38
rs969779702 Chr.9: 129889089-129889089 on Build GRCh38
rs981152024 Chr.9: 129888952-129888952 on Build GRCh38
rs796676335 Chr.9: 129889082-129889083 on Build GRCh38
rs769053737 Chr.9: 129889086-129889087 on Build GRCh38
rs368630224 Chr.9: 129889081-129889081 on Build GRCh38
rs564846361 Chr.9: 129888845-129888845 on Build GRCh38
rs750590332 Chr.9: 129888878-129888878 on Build GRCh38
rs922055136 Chr.9: 129889248-129889248 on Build GRCh38
rs1055030001 Chr.9: 129889099-129889099 on Build GRCh38
rs200600167 Chr.9: 129889082-129889083 on Build GRCh38
rs748529231 Chr.9: 129888990-129888990 on Build GRCh38
rs369756638 Chr.9: 129888966-129888966 on Build GRCh38
rs907892909 Chr.9: 129889260-129889260 on Build GRCh38
rs933368679 Chr.9: 129889057-129889057 on Build GRCh38
rs869153440 Chr.9: 129889084-129889085 on Build GRCh38
rs900602378 Chr.9: 129889053-129889053 on Build GRCh38
rs1035398495 Chr.9: 129889091-129889091 on Build GRCh38
rs910295060 Chr.9: 129889094-129889094 on Build GRCh38
rs529336710 Chr.9: 129889229-129889229 on Build GRCh38
rs974926688 Chr.9: 129889226-129889226 on Build GRCh38
rs9650722 Chr.9: 129888969-129888969 on Build GRCh38
rs1030719135 Chr.9: 129888835-129888835 on Build GRCh38

View Full Product Details