Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 69532166-69532652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM155B
Entrez Gene ID
27112
Gene Name
family with sequence similarity 155 member B
Gene Aliases
CXorf63, TED, TMEM28, bB57D9.1
UniGene
Hs.87619
Gene Chromosome Location
Chr.X: 69505204-69532508 on Build GRCh38
Transcripts
NM_015686.2

SNP Detail

SNP ID Chromosome Location
rs1047710603 Chr.X: 69532486-69532486 on Build GRCh38
rs182565571 Chr.X: 69532423-69532423 on Build GRCh38
rs778282200 Chr.X: 69532322-69532322 on Build GRCh38
rs889642863 Chr.X: 69532192-69532192 on Build GRCh38
rs992595082 Chr.X: 69532427-69532427 on Build GRCh38
rs747292682 Chr.X: 69532411-69532411 on Build GRCh38
rs369378151 Chr.X: 69532470-69532470 on Build GRCh38
rs982274509 Chr.X: 69532565-69532565 on Build GRCh38
rs930578244 Chr.X: 69532477-69532477 on Build GRCh38
rs754241495 Chr.X: 69532188-69532188 on Build GRCh38
rs769739554 Chr.X: 69532577-69532577 on Build GRCh38
rs968747922 Chr.X: 69532281-69532281 on Build GRCh38
rs372522506 Chr.X: 69532475-69532475 on Build GRCh38
rs745989370 Chr.X: 69532270-69532270 on Build GRCh38
rs796201931 Chr.X: 69532465-69532465 on Build GRCh38
rs781499611 Chr.X: 69532426-69532426 on Build GRCh38
rs1032355017 Chr.X: 69532402-69532402 on Build GRCh38
rs756947662 Chr.X: 69532474-69532474 on Build GRCh38
rs745980418 Chr.X: 69532535-69532535 on Build GRCh38
rs753125213 Chr.X: 69532295-69532295 on Build GRCh38
rs1024293675 Chr.X: 69532499-69532499 on Build GRCh38
rs755537377 Chr.X: 69532256-69532256 on Build GRCh38
rs917045223 Chr.X: 69532467-69532467 on Build GRCh38
rs886458151 Chr.X: 69532548-69532548 on Build GRCh38
rs944769703 Chr.X: 69532611-69532611 on Build GRCh38
rs779331066 Chr.X: 69532289-69532289 on Build GRCh38
rs944620050 Chr.X: 69532575-69532575 on Build GRCh38
rs911778099 Chr.X: 69532380-69532380 on Build GRCh38
rs749936615 Chr.X: 69532189-69532189 on Build GRCh38
rs779499762 Chr.X: 69532279-69532279 on Build GRCh38
rs970919235 Chr.X: 69532563-69532564 on Build GRCh38
rs151113435 Chr.X: 69532296-69532296 on Build GRCh38
rs775537495 Chr.X: 69532593-69532593 on Build GRCh38
rs1038969098 Chr.X: 69532581-69532581 on Build GRCh38

View Full Product Details