Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153587844-153588346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM58A
Entrez Gene ID
92002
Gene Name
family with sequence similarity 58 member A
Gene Aliases
STAR
UniGene
Hs.496943
Gene Chromosome Location
Chr.X: 153587925-153599177 on Build GRCh38
Transcripts
NM_001130997.2 NM_152274.4 XM_005277920.4 XM_005277921.4 XM_011531214.2 XM_011531215.2

SNP Detail

SNP ID Chromosome Location
rs191478924 Chr.X: 153588191-153588191 on Build GRCh38
rs782345588 Chr.X: 153588032-153588032 on Build GRCh38
rs782593490 Chr.X: 153588237-153588237 on Build GRCh38
rs1042281403 Chr.X: 153587928-153587928 on Build GRCh38
rs1025791779 Chr.X: 153588114-153588114 on Build GRCh38
rs782109567 Chr.X: 153588099-153588099 on Build GRCh38
rs782818914 Chr.X: 153588131-153588131 on Build GRCh38
rs782460662 Chr.X: 153588208-153588208 on Build GRCh38
rs182984949 Chr.X: 153588287-153588287 on Build GRCh38
rs906608752 Chr.X: 153588297-153588297 on Build GRCh38
rs994283167 Chr.X: 153588109-153588109 on Build GRCh38
rs781798421 Chr.X: 153588187-153588187 on Build GRCh38
rs73637844 Chr.X: 153588121-153588121 on Build GRCh38
rs73637845 Chr.X: 153588135-153588135 on Build GRCh38
rs782490421 Chr.X: 153588281-153588281 on Build GRCh38
rs782330140 Chr.X: 153588323-153588323 on Build GRCh38
rs906665848 Chr.X: 153588100-153588100 on Build GRCh38
rs182816008 Chr.X: 153587966-153587966 on Build GRCh38
rs782042762 Chr.X: 153588184-153588184 on Build GRCh38
rs895108493 Chr.X: 153587990-153587990 on Build GRCh38
rs782005402 Chr.X: 153588037-153588037 on Build GRCh38
rs58399762 Chr.X: 153587916-153587916 on Build GRCh38
rs377294285 Chr.X: 153588162-153588162 on Build GRCh38
rs782036499 Chr.X: 153588295-153588295 on Build GRCh38
rs111226362 Chr.X: 153587904-153587904 on Build GRCh38
rs149068880 Chr.X: 153587969-153587969 on Build GRCh38
rs782820779 Chr.X: 153588185-153588185 on Build GRCh38
rs368748070 Chr.X: 153588209-153588209 on Build GRCh38
rs782177465 Chr.X: 153587929-153587929 on Build GRCh38
rs782535366 Chr.X: 153587898-153587898 on Build GRCh38
rs1046415443 Chr.X: 153588291-153588291 on Build GRCh38
rs782006693 Chr.X: 153588166-153588166 on Build GRCh38
rs933139843 Chr.X: 153587925-153587925 on Build GRCh38
rs781853813 Chr.X: 153588204-153588204 on Build GRCh38
rs782636575 Chr.X: 153588286-153588286 on Build GRCh38
rs188330990 Chr.X: 153588161-153588161 on Build GRCh38
rs781909311 Chr.X: 153588217-153588217 on Build GRCh38
rs782378900 Chr.X: 153588163-153588163 on Build GRCh38
rs782597779 Chr.X: 153588294-153588294 on Build GRCh38
rs782183736 Chr.X: 153588298-153588298 on Build GRCh38
rs781902732 Chr.X: 153587897-153587897 on Build GRCh38
rs782388501 Chr.X: 153587878-153587878 on Build GRCh38
rs782263331 Chr.X: 153588142-153588142 on Build GRCh38
rs781881712 Chr.X: 153588158-153588158 on Build GRCh38
rs782611529 Chr.X: 153588132-153588132 on Build GRCh38
rs782269013 Chr.X: 153588293-153588293 on Build GRCh38
rs782217172 Chr.X: 153588257-153588257 on Build GRCh38
rs952929380 Chr.X: 153588136-153588136 on Build GRCh38
rs371373348 Chr.X: 153588133-153588133 on Build GRCh38
rs1012256196 Chr.X: 153588013-153588013 on Build GRCh38
rs782740943 Chr.X: 153588199-153588199 on Build GRCh38
rs782430896 Chr.X: 153588228-153588228 on Build GRCh38
rs782103033 Chr.X: 153587975-153587975 on Build GRCh38

View Full Product Details