Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229431638-229432138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTA1
Entrez Gene ID
58
Gene Name
actin, alpha 1, skeletal muscle
Gene Aliases
ACTA, ASMA, CFTD, CFTD1, CFTDM, MPFD, NEM1, NEM2, NEM3, SHPM
UniGene
Hs.1288
Gene Chromosome Location
Chr.1: 229431245-229434096 on Build GRCh38
Transcripts
NM_001100.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4659087 c.690G>A p.A230A Chr.1: 229432112-229432112 on Build GRCh38
6592837 c.918G>A p.T306T Chr.1: 229431793-229431793 on Build GRCh38
905239 c.962C>T p.A321V Chr.1: 229431749-229431749 on Build GRCh38
6857056 c.692C>T p.T231M Chr.1: 229432110-229432110 on Build GRCh38
1997431 c.961G>A p.A321T Chr.1: 229431750-229431750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749262171 Chr.1: 229431805-229431805 on Build GRCh38
rs868200539 Chr.1: 229431909-229431909 on Build GRCh38
rs762406327 Chr.1: 229431946-229431947 on Build GRCh38
rs754757867 Chr.1: 229431675-229431675 on Build GRCh38
rs6673359 Chr.1: 229431916-229431916 on Build GRCh38
rs769671804 Chr.1: 229431683-229431683 on Build GRCh38
rs764754382 Chr.1: 229431666-229431666 on Build GRCh38
rs776941851 Chr.1: 229431889-229431889 on Build GRCh38
rs775891184 Chr.1: 229431875-229431875 on Build GRCh38
rs372686280 Chr.1: 229431720-229431720 on Build GRCh38
rs1036629663 Chr.1: 229431920-229431920 on Build GRCh38
rs761012261 Chr.1: 229431884-229431884 on Build GRCh38
rs777628810 Chr.1: 229431923-229431923 on Build GRCh38
rs398123565 Chr.1: 229431916-229431917 on Build GRCh38
rs772275013 Chr.1: 229431943-229431943 on Build GRCh38
rs368965736 Chr.1: 229431946-229431946 on Build GRCh38
rs762410878 Chr.1: 229431892-229431892 on Build GRCh38
rs749821094 Chr.1: 229432080-229432080 on Build GRCh38
rs794727714 Chr.1: 229432117-229432117 on Build GRCh38
rs752477289 Chr.1: 229431763-229431763 on Build GRCh38
rs770414205 Chr.1: 229432068-229432068 on Build GRCh38
rs140074813 Chr.1: 229431844-229431844 on Build GRCh38
rs201696359 Chr.1: 229431695-229431695 on Build GRCh38
rs535723510 Chr.1: 229431700-229431700 on Build GRCh38
rs746125735 Chr.1: 229431917-229431918 on Build GRCh38
rs201427429 Chr.1: 229431913-229431914 on Build GRCh38
rs756611370 Chr.1: 229431663-229431663 on Build GRCh38
rs201530525 Chr.1: 229431924-229431924 on Build GRCh38
rs373224284 Chr.1: 229431944-229431944 on Build GRCh38
rs771132330 Chr.1: 229431693-229431693 on Build GRCh38
rs748592740 Chr.1: 229432064-229432064 on Build GRCh38
rs768012676 Chr.1: 229431799-229431800 on Build GRCh38
rs149762697 Chr.1: 229432057-229432057 on Build GRCh38
rs745986816 Chr.1: 229431937-229431937 on Build GRCh38
rs951259177 Chr.1: 229431874-229431874 on Build GRCh38
rs1008265049 Chr.1: 229431949-229431949 on Build GRCh38
rs374599062 Chr.1: 229431748-229431748 on Build GRCh38
rs878854374 Chr.1: 229431788-229431788 on Build GRCh38
rs780183881 Chr.1: 229431997-229431997 on Build GRCh38
rs749949117 Chr.1: 229431670-229431670 on Build GRCh38
rs746783641 Chr.1: 229431674-229431674 on Build GRCh38
rs569500994 Chr.1: 229432112-229432112 on Build GRCh38
rs937931817 Chr.1: 229431901-229431901 on Build GRCh38
rs141030526 Chr.1: 229432016-229432016 on Build GRCh38
rs876657420 Chr.1: 229431916-229431916 on Build GRCh38
rs770509913 Chr.1: 229432091-229432093 on Build GRCh38
rs770931836 Chr.1: 229431823-229431823 on Build GRCh38
rs894074159 Chr.1: 229431688-229431688 on Build GRCh38
rs765996798 Chr.1: 229431745-229431745 on Build GRCh38
rs749398634 Chr.1: 229431687-229431687 on Build GRCh38
rs753716593 Chr.1: 229431972-229431972 on Build GRCh38
rs533868659 Chr.1: 229431814-229431814 on Build GRCh38
rs759235757 Chr.1: 229431915-229431915 on Build GRCh38
rs547455787 Chr.1: 229431820-229431820 on Build GRCh38
rs764547391 Chr.1: 229431709-229431709 on Build GRCh38
rs757210215 Chr.1: 229431928-229431928 on Build GRCh38
rs759758571 Chr.1: 229431701-229431701 on Build GRCh38
rs759179371 Chr.1: 229431951-229431951 on Build GRCh38
rs778943036 Chr.1: 229431933-229431933 on Build GRCh38
rs1007095495 Chr.1: 229431710-229431710 on Build GRCh38
rs772387021 Chr.1: 229431702-229431702 on Build GRCh38
rs367543051 Chr.1: 229432075-229432075 on Build GRCh38
rs755247915 Chr.1: 229432028-229432028 on Build GRCh38
rs781135829 Chr.1: 229431676-229431676 on Build GRCh38
rs549268481 Chr.1: 229431991-229431991 on Build GRCh38
rs878861182 Chr.1: 229431906-229431906 on Build GRCh38
rs777994788 Chr.1: 229431914-229431915 on Build GRCh38
rs773996373 Chr.1: 229431950-229431950 on Build GRCh38
rs979475402 Chr.1: 229432072-229432072 on Build GRCh38
rs187159043 Chr.1: 229431945-229431945 on Build GRCh38
rs755810986 Chr.1: 229431917-229431917 on Build GRCh38
rs200342114 Chr.1: 229431988-229431988 on Build GRCh38
rs59228224 Chr.1: 229431937-229431937 on Build GRCh38
rs763749374 Chr.1: 229431970-229431970 on Build GRCh38
rs776417838 Chr.1: 229431919-229431920 on Build GRCh38
rs375095091 Chr.1: 229431671-229431671 on Build GRCh38
rs748076340 Chr.1: 229431921-229431925 on Build GRCh38
rs121909523 Chr.1: 229432020-229432020 on Build GRCh38
rs748075887 Chr.1: 229431681-229431681 on Build GRCh38
rs979277183 Chr.1: 229431752-229431753 on Build GRCh38
rs542109877 Chr.1: 229432079-229432079 on Build GRCh38
rs778997995 Chr.1: 229431824-229431824 on Build GRCh38
rs751133731 Chr.1: 229431914-229431914 on Build GRCh38
rs886044062 Chr.1: 229432087-229432087 on Build GRCh38
rs398122936 Chr.1: 229431727-229431727 on Build GRCh38
rs536198269 Chr.1: 229432091-229432091 on Build GRCh38
rs771443604 Chr.1: 229432082-229432082 on Build GRCh38
rs756078196 Chr.1: 229431769-229431769 on Build GRCh38
rs539461449 Chr.1: 229431981-229431981 on Build GRCh38
rs774940494 Chr.1: 229431962-229431962 on Build GRCh38
rs769192485 Chr.1: 229431885-229431885 on Build GRCh38
rs121909529 Chr.1: 229431830-229431830 on Build GRCh38
rs770467323 Chr.1: 229431948-229431948 on Build GRCh38
rs760406921 Chr.1: 229431964-229431964 on Build GRCh38
rs368470632 Chr.1: 229431704-229431704 on Build GRCh38
rs772690824 Chr.1: 229431946-229431946 on Build GRCh38
rs121909525 Chr.1: 229431994-229431994 on Build GRCh38
rs549197605 Chr.1: 229431718-229431718 on Build GRCh38
rs755254834 Chr.1: 229431661-229431665 on Build GRCh38
rs555808828 Chr.1: 229432103-229432103 on Build GRCh38
rs749384329 Chr.1: 229431915-229431916 on Build GRCh38
rs569073359 Chr.1: 229431794-229431794 on Build GRCh38
rs767070539 Chr.1: 229431952-229431952 on Build GRCh38
rs773313243 Chr.1: 229431686-229431686 on Build GRCh38
rs746042348 Chr.1: 229431829-229431829 on Build GRCh38

View Full Product Details