Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229432191-229432685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTA1
Entrez Gene ID
58
Gene Name
actin, alpha 1, skeletal muscle
Gene Aliases
ACTA, ASMA, CFTD, CFTD1, CFTDM, MPFD, NEM1, NEM2, NEM3, SHPM
UniGene
Hs.1288
Gene Chromosome Location
Chr.1: 229431245-229434096 on Build GRCh38
Transcripts
NM_001100.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600146 c.353G>A p.R118H Chr.1: 229432657-229432657 on Build GRCh38
5009446 c.543C>T p.D181D Chr.1: 229432343-229432343 on Build GRCh38
6124665 c.463C>A p.L155M Chr.1: 229432423-229432423 on Build GRCh38
4957300 c.429C>T p.S143S Chr.1: 229432581-229432581 on Build GRCh38
6061493 c.488A>T p.H163L Chr.1: 229432398-229432398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770173089 Chr.1: 229432298-229432298 on Build GRCh38
rs200705851 Chr.1: 229432250-229432250 on Build GRCh38
rs751581251 Chr.1: 229432278-229432278 on Build GRCh38
rs758924495 Chr.1: 229432534-229432534 on Build GRCh38
rs200258538 Chr.1: 229432644-229432644 on Build GRCh38
rs121909526 Chr.1: 229432596-229432596 on Build GRCh38
rs765588353 Chr.1: 229432590-229432590 on Build GRCh38
rs774282451 Chr.1: 229432470-229432470 on Build GRCh38
rs121909522 Chr.1: 229432393-229432393 on Build GRCh38
rs759606148 Chr.1: 229432331-229432331 on Build GRCh38
rs367874241 Chr.1: 229432632-229432632 on Build GRCh38
rs770722574 Chr.1: 229432550-229432550 on Build GRCh38
rs778314735 Chr.1: 229432217-229432217 on Build GRCh38
rs374307939 Chr.1: 229432251-229432251 on Build GRCh38
rs779150156 Chr.1: 229432523-229432523 on Build GRCh38
rs757701835 Chr.1: 229432409-229432409 on Build GRCh38
rs527621 Chr.1: 229432484-229432484 on Build GRCh38
rs760949276 Chr.1: 229432511-229432511 on Build GRCh38
rs931098868 Chr.1: 229432248-229432248 on Build GRCh38
rs398123564 Chr.1: 229432564-229432564 on Build GRCh38
rs748900759 Chr.1: 229432549-229432549 on Build GRCh38
rs886039557 Chr.1: 229432580-229432580 on Build GRCh38
rs549240406 Chr.1: 229432490-229432490 on Build GRCh38
rs112919437 Chr.1: 229432433-229432433 on Build GRCh38
rs200976037 Chr.1: 229432553-229432553 on Build GRCh38
rs754270462 Chr.1: 229432389-229432389 on Build GRCh38
rs772957464 Chr.1: 229432440-229432440 on Build GRCh38
rs768897318 Chr.1: 229432617-229432617 on Build GRCh38
rs1042419556 Chr.1: 229432538-229432538 on Build GRCh38
rs755756897 Chr.1: 229432544-229432544 on Build GRCh38
rs121909520 Chr.1: 229432660-229432660 on Build GRCh38
rs551674486 Chr.1: 229432545-229432545 on Build GRCh38
rs201388631 Chr.1: 229432541-229432541 on Build GRCh38
rs371799971 Chr.1: 229432266-229432266 on Build GRCh38
rs756483510 Chr.1: 229432215-229432215 on Build GRCh38
rs775708474 Chr.1: 229432566-229432566 on Build GRCh38
rs757583419 Chr.1: 229432522-229432522 on Build GRCh38
rs749499154 Chr.1: 229432614-229432614 on Build GRCh38
rs764479229 Chr.1: 229432517-229432517 on Build GRCh38
rs1017212416 Chr.1: 229432387-229432387 on Build GRCh38
rs769533594 Chr.1: 229432263-229432263 on Build GRCh38
rs778261678 Chr.1: 229432364-229432364 on Build GRCh38
rs200094415 Chr.1: 229432337-229432337 on Build GRCh38
rs190340782 Chr.1: 229432391-229432391 on Build GRCh38
rs746374077 Chr.1: 229432424-229432424 on Build GRCh38
rs767928898 Chr.1: 229432436-229432437 on Build GRCh38
rs371410845 Chr.1: 229432575-229432575 on Build GRCh38
rs753181739 Chr.1: 229432537-229432537 on Build GRCh38
rs771519531 Chr.1: 229432247-229432247 on Build GRCh38
rs756428017 Chr.1: 229432342-229432342 on Build GRCh38
rs750797766 Chr.1: 229432609-229432609 on Build GRCh38
rs766934634 Chr.1: 229432621-229432621 on Build GRCh38
rs903232422 Chr.1: 229432527-229432527 on Build GRCh38
rs774293910 Chr.1: 229432662-229432662 on Build GRCh38
rs780711799 Chr.1: 229432427-229432427 on Build GRCh38
rs75926206 Chr.1: 229432582-229432582 on Build GRCh38
rs867087007 Chr.1: 229432432-229432432 on Build GRCh38
rs959694514 Chr.1: 229432605-229432605 on Build GRCh38
rs1057519311 Chr.1: 229432365-229432365 on Build GRCh38
rs111812550 Chr.1: 229432269-229432269 on Build GRCh38
rs569112604 Chr.1: 229432520-229432520 on Build GRCh38
rs369670582 Chr.1: 229432340-229432340 on Build GRCh38
rs553019935 Chr.1: 229432587-229432587 on Build GRCh38
rs587780272 Chr.1: 229432371-229432371 on Build GRCh38
rs377265320 Chr.1: 229432257-229432257 on Build GRCh38
rs750903065 Chr.1: 229432528-229432528 on Build GRCh38
rs893970878 Chr.1: 229432231-229432231 on Build GRCh38
rs767656452 Chr.1: 229432474-229432474 on Build GRCh38
rs768144106 Chr.1: 229432426-229432426 on Build GRCh38
rs201769848 Chr.1: 229432213-229432213 on Build GRCh38
rs869312739 Chr.1: 229432295-229432295 on Build GRCh38
rs76030344 Chr.1: 229432557-229432557 on Build GRCh38
rs752875062 Chr.1: 229432505-229432505 on Build GRCh38
rs370798526 Chr.1: 229432443-229432443 on Build GRCh38
rs769482139 Chr.1: 229432438-229432438 on Build GRCh38
rs773010488 Chr.1: 229432265-229432265 on Build GRCh38
rs759242559 Chr.1: 229432345-229432345 on Build GRCh38
rs749765603 Chr.1: 229432229-229432229 on Build GRCh38
rs398123563 Chr.1: 229432568-229432568 on Build GRCh38
rs755556717 Chr.1: 229432659-229432659 on Build GRCh38
rs727503797 Chr.1: 229432350-229432350 on Build GRCh38
rs587780271 Chr.1: 229432597-229432597 on Build GRCh38
rs560102755 Chr.1: 229432473-229432473 on Build GRCh38
rs544387567 Chr.1: 229432249-229432249 on Build GRCh38
rs770648114 Chr.1: 229432457-229432457 on Build GRCh38
rs779530455 Chr.1: 229432415-229432415 on Build GRCh38
rs747745049 Chr.1: 229432437-229432437 on Build GRCh38

View Full Product Details