Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229433852-229434341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTA1
Entrez Gene ID
58
Gene Name
actin, alpha 1, skeletal muscle
Gene Aliases
ACTA, ASMA, CFTD, CFTD1, CFTDM, MPFD, NEM1, NEM2, NEM3, SHPM
UniGene
Hs.1288
Gene Chromosome Location
Chr.1: 229431245-229434096 on Build GRCh38
Transcripts
NM_001100.3

SNP Detail

SNP ID Chromosome Location
rs970284813 Chr.1: 229433905-229433905 on Build GRCh38
rs898939535 Chr.1: 229434315-229434315 on Build GRCh38
rs1052079604 Chr.1: 229434295-229434295 on Build GRCh38
rs892878682 Chr.1: 229434136-229434136 on Build GRCh38
rs1054536531 Chr.1: 229434285-229434285 on Build GRCh38
rs386640096 Chr.1: 229434056-229434057 on Build GRCh38
rs982750683 Chr.1: 229434203-229434203 on Build GRCh38
rs1037836213 Chr.1: 229434156-229434156 on Build GRCh38
rs367844802 Chr.1: 229434057-229434057 on Build GRCh38
rs1012001775 Chr.1: 229434320-229434320 on Build GRCh38
rs605430 Chr.1: 229434056-229434056 on Build GRCh38
rs905385088 Chr.1: 229434313-229434313 on Build GRCh38
rs567089400 Chr.1: 229434321-229434321 on Build GRCh38
rs970338609 Chr.1: 229434235-229434235 on Build GRCh38
rs1004640381 Chr.1: 229434150-229434150 on Build GRCh38
rs928625808 Chr.1: 229433956-229433956 on Build GRCh38
rs115052102 Chr.1: 229433983-229433983 on Build GRCh38
rs933248823 Chr.1: 229434294-229434294 on Build GRCh38
rs951362380 Chr.1: 229434035-229434035 on Build GRCh38
rs947196871 Chr.1: 229434010-229434010 on Build GRCh38
rs377518258 Chr.1: 229434306-229434306 on Build GRCh38
rs982321168 Chr.1: 229433991-229433991 on Build GRCh38
rs968758659 Chr.1: 229434277-229434277 on Build GRCh38
rs933219405 Chr.1: 229434064-229434064 on Build GRCh38
rs892795782 Chr.1: 229434293-229434293 on Build GRCh38
rs767031604 Chr.1: 229434283-229434283 on Build GRCh38
rs974569759 Chr.1: 229434290-229434290 on Build GRCh38
rs147267882 Chr.1: 229433967-229433967 on Build GRCh38
rs908197657 Chr.1: 229434151-229434151 on Build GRCh38
rs1051475948 Chr.1: 229434127-229434127 on Build GRCh38
rs879681626 Chr.1: 229434242-229434243 on Build GRCh38
rs902879482 Chr.1: 229434083-229434083 on Build GRCh38
rs71574315 Chr.1: 229434056-229434056 on Build GRCh38
rs963156837 Chr.1: 229434213-229434213 on Build GRCh38
rs914413396 Chr.1: 229434002-229434002 on Build GRCh38
rs565479298 Chr.1: 229434037-229434037 on Build GRCh38
rs988930009 Chr.1: 229434253-229434253 on Build GRCh38
rs556542043 Chr.1: 229433949-229433949 on Build GRCh38
rs998536621 Chr.1: 229434091-229434091 on Build GRCh38
rs956182252 Chr.1: 229433962-229433962 on Build GRCh38
rs1003527120 Chr.1: 229434238-229434238 on Build GRCh38
rs974342704 Chr.1: 229433884-229433884 on Build GRCh38
rs1037358844 Chr.1: 229434312-229434312 on Build GRCh38
rs1054100162 Chr.1: 229434092-229434092 on Build GRCh38
rs1050985027 Chr.1: 229434080-229434080 on Build GRCh38
rs962834721 Chr.1: 229433878-229433878 on Build GRCh38
rs886072130 Chr.1: 229434147-229434147 on Build GRCh38
rs981489760 Chr.1: 229433922-229433922 on Build GRCh38
rs34465109 Chr.1: 229434053-229434054 on Build GRCh38
rs957116147 Chr.1: 229433889-229433889 on Build GRCh38
rs116217268 Chr.1: 229433871-229433871 on Build GRCh38
rs749345801 Chr.1: 229434026-229434026 on Build GRCh38
rs928005893 Chr.1: 229434027-229434027 on Build GRCh38
rs751941168 Chr.1: 229434251-229434251 on Build GRCh38
rs551349915 Chr.1: 229434225-229434225 on Build GRCh38
rs921763884 Chr.1: 229434292-229434292 on Build GRCh38
rs941416928 Chr.1: 229434054-229434054 on Build GRCh38
rs761180150 Chr.1: 229434258-229434258 on Build GRCh38
rs533815304 Chr.1: 229434252-229434252 on Build GRCh38
rs1035125190 Chr.1: 229434316-229434316 on Build GRCh38
rs572772527 Chr.1: 229434038-229434038 on Build GRCh38
rs73112253 Chr.1: 229434248-229434248 on Build GRCh38
rs921120572 Chr.1: 229434247-229434247 on Build GRCh38
rs879788591 Chr.1: 229434056-229434057 on Build GRCh38
rs548779559 Chr.1: 229434157-229434157 on Build GRCh38
rs1016903625 Chr.1: 229434208-229434208 on Build GRCh38
rs371683324 Chr.1: 229434148-229434148 on Build GRCh38
rs909967841 Chr.1: 229434041-229434041 on Build GRCh38
rs547224722 Chr.1: 229434305-229434305 on Build GRCh38
rs1023194278 Chr.1: 229433896-229433896 on Build GRCh38
rs605428 Chr.1: 229434057-229434057 on Build GRCh38
rs368699658 Chr.1: 229434063-229434063 on Build GRCh38
rs562509475 Chr.1: 229434164-229434164 on Build GRCh38
rs1011509357 Chr.1: 229433945-229433945 on Build GRCh38
rs1023122069 Chr.1: 229434205-229434205 on Build GRCh38
rs1024257386 Chr.1: 229434152-229434152 on Build GRCh38
rs117656252 Chr.1: 229434178-229434178 on Build GRCh38

View Full Product Details