Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229521223-229521728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB10
Entrez Gene ID
23456
Gene Name
ATP binding cassette subfamily B member 10
Gene Aliases
EST20237, M-ABC2, MTABC2
UniGene
Hs.17614
Gene Chromosome Location
Chr.1: 229516582-229558987 on Build GRCh38
Transcripts
NM_012089.2 XM_011544135.2 XM_011544136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5987011 c.1942C>A p.L648M Chr.1: 229521600-229521600 on Build GRCh38
4443756 c.1937G>A p.R646H Chr.1: 229521605-229521605 on Build GRCh38
5458053 c.1932T>G p.I644M Chr.1: 229521610-229521610 on Build GRCh38
3671661 c.1922G>T p.R641L Chr.1: 229521620-229521620 on Build GRCh38
4422268 c.1923G>T p.R641R Chr.1: 229521619-229521619 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs978662634 Chr.1: 229521459-229521459 on Build GRCh38
rs994416554 Chr.1: 229521271-229521271 on Build GRCh38
rs186656454 Chr.1: 229521436-229521436 on Build GRCh38
rs779696259 Chr.1: 229521563-229521563 on Build GRCh38
rs539537539 Chr.1: 229521688-229521688 on Build GRCh38
rs367927448 Chr.1: 229521558-229521558 on Build GRCh38
rs540327102 Chr.1: 229521312-229521312 on Build GRCh38
rs932154201 Chr.1: 229521326-229521326 on Build GRCh38
rs754553026 Chr.1: 229521654-229521654 on Build GRCh38
rs888192713 Chr.1: 229521315-229521315 on Build GRCh38
rs1017882170 Chr.1: 229521325-229521325 on Build GRCh38
rs771868423 Chr.1: 229521573-229521573 on Build GRCh38
rs752148489 Chr.1: 229521674-229521674 on Build GRCh38
rs575826331 Chr.1: 229521457-229521457 on Build GRCh38
rs759092914 Chr.1: 229521316-229521316 on Build GRCh38
rs377051670 Chr.1: 229521614-229521614 on Build GRCh38
rs929951773 Chr.1: 229521552-229521552 on Build GRCh38
rs746564965 Chr.1: 229521641-229521641 on Build GRCh38
rs753468956 Chr.1: 229521681-229521681 on Build GRCh38
rs3738186 Chr.1: 229521483-229521483 on Build GRCh38
rs138401798 Chr.1: 229521698-229521698 on Build GRCh38
rs373815737 Chr.1: 229521555-229521555 on Build GRCh38
rs759383118 Chr.1: 229521568-229521568 on Build GRCh38
rs559268041 Chr.1: 229521248-229521248 on Build GRCh38
rs1031567400 Chr.1: 229521699-229521699 on Build GRCh38
rs986294687 Chr.1: 229521335-229521335 on Build GRCh38
rs983318608 Chr.1: 229521458-229521458 on Build GRCh38
rs867847624 Chr.1: 229521333-229521333 on Build GRCh38
rs947931460 Chr.1: 229521250-229521250 on Build GRCh38
rs761962076 Chr.1: 229521631-229521631 on Build GRCh38
rs958209178 Chr.1: 229521701-229521701 on Build GRCh38
rs578146227 Chr.1: 229521269-229521269 on Build GRCh38
rs77633913 Chr.1: 229521475-229521475 on Build GRCh38
rs72751736 Chr.1: 229521487-229521487 on Build GRCh38
rs897829018 Chr.1: 229521500-229521500 on Build GRCh38
rs749015900 Chr.1: 229521556-229521556 on Build GRCh38
rs780854611 Chr.1: 229521653-229521654 on Build GRCh38
rs368158613 Chr.1: 229521473-229521474 on Build GRCh38
rs1025354225 Chr.1: 229521291-229521291 on Build GRCh38
rs758811279 Chr.1: 229521666-229521666 on Build GRCh38
rs755682478 Chr.1: 229521682-229521682 on Build GRCh38
rs955120300 Chr.1: 229521692-229521692 on Build GRCh38
rs4148762 Chr.1: 229521484-229521484 on Build GRCh38
rs974943405 Chr.1: 229521410-229521410 on Build GRCh38
rs780641738 Chr.1: 229521541-229521541 on Build GRCh38
rs760653297 Chr.1: 229521613-229521613 on Build GRCh38
rs368398099 Chr.1: 229521668-229521668 on Build GRCh38
rs1031006596 Chr.1: 229521419-229521420 on Build GRCh38
rs564271976 Chr.1: 229521268-229521268 on Build GRCh38
rs750319612 Chr.1: 229521541-229521542 on Build GRCh38
rs867589451 Chr.1: 229521480-229521480 on Build GRCh38
rs774296144 Chr.1: 229521361-229521361 on Build GRCh38
rs750711754 Chr.1: 229521664-229521664 on Build GRCh38
rs754042899 Chr.1: 229521615-229521615 on Build GRCh38
rs747774187 Chr.1: 229521659-229521661 on Build GRCh38
rs912134573 Chr.1: 229521344-229521344 on Build GRCh38
rs371797996 Chr.1: 229521563-229521563 on Build GRCh38
rs113076209 Chr.1: 229521637-229521637 on Build GRCh38
rs1036806707 Chr.1: 229521373-229521373 on Build GRCh38
rs765448204 Chr.1: 229521643-229521643 on Build GRCh38
rs777484197 Chr.1: 229521685-229521685 on Build GRCh38
rs769477656 Chr.1: 229521554-229521554 on Build GRCh38
rs916559397 Chr.1: 229521483-229521484 on Build GRCh38
rs989585055 Chr.1: 229521474-229521474 on Build GRCh38
rs747561901 Chr.1: 229521543-229521543 on Build GRCh38
rs201648623 Chr.1: 229521579-229521579 on Build GRCh38
rs911237 Chr.1: 229521418-229521418 on Build GRCh38
rs866273357 Chr.1: 229521609-229521609 on Build GRCh38
rs936724128 Chr.1: 229521469-229521469 on Build GRCh38
rs758476301 Chr.1: 229521545-229521545 on Build GRCh38
rs10799534 Chr.1: 229521466-229521466 on Build GRCh38

View Full Product Details