Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49366027-49366531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRGX
Entrez Gene ID
644168
Gene Name
dorsal root ganglia homeobox
Gene Aliases
DRG11, PRRXL1
UniGene
Hs.534530
Gene Chromosome Location
Chr.10: 49364085-49396016 on Build GRCh38
Transcripts
NM_001276451.1 XM_011540089.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4887875 c.743C>T p.P248L Chr.10: 49366180-49366180 on Build GRCh38
918647 c.738G>A p.A246A Chr.10: 49366185-49366185 on Build GRCh38
918649 c.565G>A p.V189I Chr.10: 49366358-49366358 on Build GRCh38
4905118 c.654C>T p.A218A Chr.10: 49366269-49366269 on Build GRCh38
5002376 c.727G>T p.A243S Chr.10: 49366196-49366196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371865203 Chr.10: 49366346-49366346 on Build GRCh38
rs753856701 Chr.10: 49366158-49366158 on Build GRCh38
rs201807477 Chr.10: 49366267-49366267 on Build GRCh38
rs374365523 Chr.10: 49366257-49366257 on Build GRCh38
rs926432631 Chr.10: 49366415-49366415 on Build GRCh38
rs770284135 Chr.10: 49366363-49366363 on Build GRCh38
rs368998576 Chr.10: 49366403-49366403 on Build GRCh38
rs375388153 Chr.10: 49366280-49366280 on Build GRCh38
rs375343755 Chr.10: 49366171-49366171 on Build GRCh38
rs745374751 Chr.10: 49366277-49366277 on Build GRCh38
rs768125202 Chr.10: 49366073-49366073 on Build GRCh38
rs200974922 Chr.10: 49366344-49366344 on Build GRCh38
rs776886652 Chr.10: 49366292-49366292 on Build GRCh38
rs201659578 Chr.10: 49366266-49366266 on Build GRCh38
rs781071913 Chr.10: 49366099-49366099 on Build GRCh38
rs759957813 Chr.10: 49366137-49366137 on Build GRCh38
rs761028683 Chr.10: 49366146-49366146 on Build GRCh38
rs773212963 Chr.10: 49366301-49366301 on Build GRCh38
rs370735058 Chr.10: 49366268-49366268 on Build GRCh38
rs770614690 Chr.10: 49366187-49366187 on Build GRCh38
rs765331314 Chr.10: 49366331-49366331 on Build GRCh38
rs748609775 Chr.10: 49366299-49366299 on Build GRCh38
rs761756732 Chr.10: 49366408-49366408 on Build GRCh38
rs540249602 Chr.10: 49366481-49366481 on Build GRCh38
rs760356952 Chr.10: 49366312-49366312 on Build GRCh38
rs985453164 Chr.10: 49366303-49366303 on Build GRCh38
rs769796474 Chr.10: 49366100-49366100 on Build GRCh38
rs192890807 Chr.10: 49366452-49366452 on Build GRCh38
rs543215196 Chr.10: 49366453-49366453 on Build GRCh38
rs764665485 Chr.10: 49366154-49366154 on Build GRCh38
rs187937853 Chr.10: 49366432-49366432 on Build GRCh38
rs774206760 Chr.10: 49366123-49366123 on Build GRCh38
rs371537979 Chr.10: 49366080-49366080 on Build GRCh38
rs777092913 Chr.10: 49366108-49366108 on Build GRCh38
rs772526015 Chr.10: 49366140-49366140 on Build GRCh38
rs776562089 Chr.10: 49366404-49366404 on Build GRCh38
rs775944524 Chr.10: 49366141-49366141 on Build GRCh38
rs35005538 Chr.10: 49366107-49366108 on Build GRCh38
rs538583055 Chr.10: 49366189-49366189 on Build GRCh38
rs965969926 Chr.10: 49366121-49366121 on Build GRCh38
rs765162373 Chr.10: 49366358-49366359 on Build GRCh38
rs528433437 Chr.10: 49366077-49366077 on Build GRCh38
rs748838114 Chr.10: 49366109-49366109 on Build GRCh38
rs770302415 Chr.10: 49366300-49366300 on Build GRCh38
rs774326355 Chr.10: 49366400-49366400 on Build GRCh38
rs564966737 Chr.10: 49366495-49366495 on Build GRCh38
rs370954422 Chr.10: 49366159-49366159 on Build GRCh38
rs751338807 Chr.10: 49366095-49366095 on Build GRCh38
rs769027545 Chr.10: 49366205-49366205 on Build GRCh38
rs201682854 Chr.10: 49366305-49366305 on Build GRCh38
rs916573836 Chr.10: 49366060-49366060 on Build GRCh38
rs777835103 Chr.10: 49366185-49366185 on Build GRCh38
rs770704583 Chr.10: 49366115-49366115 on Build GRCh38
rs868407889 Chr.10: 49366330-49366330 on Build GRCh38
rs758654754 Chr.10: 49366163-49366163 on Build GRCh38
rs779170357 Chr.10: 49366084-49366084 on Build GRCh38
rs755940392 Chr.10: 49366172-49366172 on Build GRCh38
rs896150026 Chr.10: 49366207-49366207 on Build GRCh38
rs533549211 Chr.10: 49366328-49366328 on Build GRCh38
rs774791991 Chr.10: 49366067-49366067 on Build GRCh38
rs572199315 Chr.10: 49366503-49366503 on Build GRCh38
rs769100690 Chr.10: 49366290-49366290 on Build GRCh38
rs561177476 Chr.10: 49366460-49366460 on Build GRCh38
rs754648308 Chr.10: 49366379-49366379 on Build GRCh38
rs748520186 Chr.10: 49366360-49366360 on Build GRCh38
rs138046893 Chr.10: 49366081-49366081 on Build GRCh38
rs753489697 Chr.10: 49366279-49366279 on Build GRCh38
rs775648598 Chr.10: 49366196-49366196 on Build GRCh38
rs926389796 Chr.10: 49366178-49366178 on Build GRCh38
rs778676897 Chr.10: 49366188-49366188 on Build GRCh38
rs765241499 Chr.10: 49366430-49366430 on Build GRCh38
rs950281554 Chr.10: 49366248-49366248 on Build GRCh38
rs749350307 Chr.10: 49366388-49366388 on Build GRCh38
rs545740512 Chr.10: 49366378-49366378 on Build GRCh38
rs765256410 Chr.10: 49366246-49366246 on Build GRCh38
rs750680023 Chr.10: 49366161-49366161 on Build GRCh38
rs369280603 Chr.10: 49366359-49366359 on Build GRCh38
rs867296023 Chr.10: 49366340-49366340 on Build GRCh38
rs761165458 Chr.10: 49366197-49366197 on Build GRCh38
rs940870733 Chr.10: 49366341-49366341 on Build GRCh38
rs372695489 Chr.10: 49366468-49366468 on Build GRCh38
rs150871858 Chr.10: 49366052-49366052 on Build GRCh38
rs554330479 Chr.10: 49366304-49366304 on Build GRCh38
rs746628925 Chr.10: 49366353-49366353 on Build GRCh38
rs763910282 Chr.10: 49366315-49366315 on Build GRCh38
rs771084075 Chr.10: 49366391-49366391 on Build GRCh38
rs1015130635 Chr.10: 49366051-49366051 on Build GRCh38
rs763107153 Chr.10: 49366265-49366265 on Build GRCh38
rs532392924 Chr.10: 49366502-49366502 on Build GRCh38
rs766145052 Chr.10: 49366066-49366066 on Build GRCh38
rs577210939 Chr.10: 49366402-49366402 on Build GRCh38
rs761847176 Chr.10: 49366224-49366224 on Build GRCh38
rs368054205 Chr.10: 49366186-49366186 on Build GRCh38
rs370393487 Chr.10: 49366206-49366206 on Build GRCh38
rs377685998 Chr.10: 49366093-49366093 on Build GRCh38
rs375866724 Chr.10: 49366384-49366384 on Build GRCh38
rs771863541 Chr.10: 49366191-49366191 on Build GRCh38
rs753699459 Chr.10: 49366324-49366324 on Build GRCh38
rs867908614 Chr.10: 49366136-49366136 on Build GRCh38
rs558290426 Chr.10: 49366205-49366206 on Build GRCh38
rs749937635 Chr.10: 49366336-49366336 on Build GRCh38
rs948320801 Chr.10: 49366063-49366063 on Build GRCh38
rs183224892 Chr.10: 49366269-49366269 on Build GRCh38
rs143541395 Chr.10: 49366358-49366358 on Build GRCh38
rs754812158 Chr.10: 49366096-49366096 on Build GRCh38
rs778507201 Chr.10: 49366276-49366276 on Build GRCh38
rs1011452390 Chr.10: 49366150-49366150 on Build GRCh38

View Full Product Details