Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104393200-104393747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP58
Entrez Gene ID
159686
Gene Name
cilia and flagella associated protein 58
Gene Aliases
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1
UniGene
Hs.253576
Gene Chromosome Location
Chr.10: 104339124-104455090 on Build GRCh38
Transcripts
NM_001008723.1 XM_011539377.2 XM_017015787.1 XM_017015788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199870 c.1546A>C p.K516Q Chr.10: 104393347-104393347 on Build GRCh38
5360080 c.1531G>T p.E511* Chr.10: 104393332-104393332 on Build GRCh38
3433982 c.1656G>A p.K552K Chr.10: 104393457-104393457 on Build GRCh38
5898383 c.1613C>T p.S538F Chr.10: 104393414-104393414 on Build GRCh38
200475 c.1646G>A p.R549Q Chr.10: 104393447-104393447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751389686 Chr.10: 104393349-104393349 on Build GRCh38
rs371289437 Chr.10: 104393358-104393358 on Build GRCh38
rs914162600 Chr.10: 104393514-104393514 on Build GRCh38
rs61732088 Chr.10: 104393447-104393447 on Build GRCh38
rs138726534 Chr.10: 104393446-104393446 on Build GRCh38
rs947967103 Chr.10: 104393609-104393609 on Build GRCh38
rs773414802 Chr.10: 104393477-104393477 on Build GRCh38
rs781410571 Chr.10: 104393356-104393356 on Build GRCh38
rs971232560 Chr.10: 104393411-104393411 on Build GRCh38
rs4582900 Chr.10: 104393281-104393281 on Build GRCh38
rs140999253 Chr.10: 104393416-104393416 on Build GRCh38
rs754720963 Chr.10: 104393330-104393330 on Build GRCh38
rs745580272 Chr.10: 104393480-104393480 on Build GRCh38
rs775144506 Chr.10: 104393497-104393497 on Build GRCh38
rs1047674872 Chr.10: 104393357-104393357 on Build GRCh38
rs926864032 Chr.10: 104393228-104393228 on Build GRCh38
rs548675831 Chr.10: 104393241-104393241 on Build GRCh38
rs759103172 Chr.10: 104393385-104393385 on Build GRCh38
rs899233276 Chr.10: 104393722-104393722 on Build GRCh38
rs780990612 Chr.10: 104393400-104393400 on Build GRCh38
rs764637221 Chr.10: 104393324-104393324 on Build GRCh38
rs148917745 Chr.10: 104393237-104393237 on Build GRCh38
rs185106263 Chr.10: 104393544-104393544 on Build GRCh38
rs759098745 Chr.10: 104393303-104393303 on Build GRCh38
rs779968827 Chr.10: 104393362-104393362 on Build GRCh38
rs895072485 Chr.10: 104393593-104393593 on Build GRCh38
rs568976257 Chr.10: 104393654-104393654 on Build GRCh38
rs930503082 Chr.10: 104393646-104393646 on Build GRCh38
rs779575094 Chr.10: 104393452-104393452 on Build GRCh38
rs761708432 Chr.10: 104393511-104393511 on Build GRCh38
rs936543885 Chr.10: 104393574-104393574 on Build GRCh38
rs781047705 Chr.10: 104393236-104393236 on Build GRCh38
rs757105858 Chr.10: 104393438-104393438 on Build GRCh38
rs974558065 Chr.10: 104393579-104393579 on Build GRCh38
rs985549875 Chr.10: 104393483-104393483 on Build GRCh38
rs751444811 Chr.10: 104393429-104393429 on Build GRCh38
rs752743350 Chr.10: 104393328-104393328 on Build GRCh38
rs148099758 Chr.10: 104393692-104393693 on Build GRCh38
rs374090530 Chr.10: 104393488-104393488 on Build GRCh38
rs4614364 Chr.10: 104393280-104393280 on Build GRCh38
rs148052621 Chr.10: 104393685-104393685 on Build GRCh38
rs764026343 Chr.10: 104393418-104393418 on Build GRCh38
rs1004217759 Chr.10: 104393398-104393398 on Build GRCh38
rs772507702 Chr.10: 104393375-104393375 on Build GRCh38
rs143609667 Chr.10: 104393521-104393521 on Build GRCh38
rs958516205 Chr.10: 104393230-104393230 on Build GRCh38
rs201643990 Chr.10: 104393443-104393443 on Build GRCh38
rs920416281 Chr.10: 104393588-104393588 on Build GRCh38
rs531700296 Chr.10: 104393282-104393282 on Build GRCh38
rs779739165 Chr.10: 104393233-104393233 on Build GRCh38
rs748651852 Chr.10: 104393470-104393470 on Build GRCh38
rs148122772 Chr.10: 104393383-104393383 on Build GRCh38
rs923689912 Chr.10: 104393550-104393550 on Build GRCh38
rs774838433 Chr.10: 104393415-104393415 on Build GRCh38
rs760476539 Chr.10: 104393523-104393523 on Build GRCh38
rs1024950552 Chr.10: 104393392-104393392 on Build GRCh38
rs753364521 Chr.10: 104393302-104393302 on Build GRCh38
rs555570642 Chr.10: 104393354-104393354 on Build GRCh38
rs764839849 Chr.10: 104393404-104393404 on Build GRCh38
rs769185326 Chr.10: 104393292-104393292 on Build GRCh38
rs774957687 Chr.10: 104393369-104393369 on Build GRCh38
rs776399141 Chr.10: 104393378-104393378 on Build GRCh38
rs554777395 Chr.10: 104393711-104393711 on Build GRCh38
rs758234926 Chr.10: 104393344-104393344 on Build GRCh38
rs763584794 Chr.10: 104393504-104393504 on Build GRCh38
rs113432791 Chr.10: 104393715-104393715 on Build GRCh38
rs1049390917 Chr.10: 104393234-104393234 on Build GRCh38
rs762218842 Chr.10: 104393296-104393296 on Build GRCh38
rs749858144 Chr.10: 104393365-104393365 on Build GRCh38
rs769352236 Chr.10: 104393496-104393496 on Build GRCh38
rs772628264 Chr.10: 104393474-104393474 on Build GRCh38
rs762467083 Chr.10: 104393498-104393498 on Build GRCh38
rs774339371 Chr.10: 104393506-104393506 on Build GRCh38
rs748527186 Chr.10: 104393372-104393372 on Build GRCh38
rs939975196 Chr.10: 104393683-104393683 on Build GRCh38
rs772816145 Chr.10: 104393656-104393656 on Build GRCh38
rs943104288 Chr.10: 104393556-104393556 on Build GRCh38
rs911588413 Chr.10: 104393529-104393529 on Build GRCh38
rs769307766 Chr.10: 104393366-104393366 on Build GRCh38
rs551844936 Chr.10: 104393558-104393558 on Build GRCh38
rs774835210 Chr.10: 104393293-104393293 on Build GRCh38
rs756285571 Chr.10: 104393360-104393360 on Build GRCh38
rs1046839311 Chr.10: 104393647-104393647 on Build GRCh38
rs1012691763 Chr.10: 104393386-104393386 on Build GRCh38
rs756269674 Chr.10: 104393445-104393445 on Build GRCh38
rs150206828 Chr.10: 104393441-104393441 on Build GRCh38
rs571131681 Chr.10: 104393530-104393530 on Build GRCh38
rs745969872 Chr.10: 104393279-104393279 on Build GRCh38
rs145923348 Chr.10: 104393346-104393346 on Build GRCh38
rs191845175 Chr.10: 104393522-104393522 on Build GRCh38

View Full Product Details