Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110510708-110511217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP5
Entrez Gene ID
1847
Gene Name
dual specificity phosphatase 5
Gene Aliases
DUSP, HVH3
UniGene
Hs.2128
Gene Chromosome Location
Chr.10: 110497867-110511544 on Build GRCh38
Transcripts
NM_004419.3

SNP Detail

SNP ID Chromosome Location
rs1042606 Chr.10: 110510917-110510917 on Build GRCh38
rs538430754 Chr.10: 110511006-110511006 on Build GRCh38
rs959734208 Chr.10: 110510827-110510827 on Build GRCh38
rs773047464 Chr.10: 110510952-110510952 on Build GRCh38
rs896054849 Chr.10: 110511052-110511052 on Build GRCh38
rs759127203 Chr.10: 110511178-110511178 on Build GRCh38
rs933251322 Chr.10: 110510994-110510994 on Build GRCh38
rs41291910 Chr.10: 110511023-110511023 on Build GRCh38
rs923184191 Chr.10: 110510961-110510961 on Build GRCh38
rs147624233 Chr.10: 110510935-110510935 on Build GRCh38
rs182331064 Chr.10: 110510923-110510923 on Build GRCh38
rs1037879194 Chr.10: 110511014-110511014 on Build GRCh38
rs142624976 Chr.10: 110510862-110510862 on Build GRCh38
rs1050264957 Chr.10: 110510756-110510756 on Build GRCh38
rs529289174 Chr.10: 110511015-110511015 on Build GRCh38
rs1033829991 Chr.10: 110510908-110510908 on Build GRCh38
rs75306833 Chr.10: 110510997-110510997 on Build GRCh38
rs543863068 Chr.10: 110510921-110510921 on Build GRCh38
rs1016819143 Chr.10: 110510755-110510755 on Build GRCh38
rs945729312 Chr.10: 110510933-110510933 on Build GRCh38
rs41291908 Chr.10: 110510882-110510882 on Build GRCh38
rs1025411687 Chr.10: 110511101-110511101 on Build GRCh38
rs968448488 Chr.10: 110511081-110511081 on Build GRCh38
rs564582088 Chr.10: 110510806-110510806 on Build GRCh38
rs922505098 Chr.10: 110510913-110510913 on Build GRCh38
rs1020018484 Chr.10: 110510856-110510856 on Build GRCh38
rs565366601 Chr.10: 110511188-110511188 on Build GRCh38
rs539790780 Chr.10: 110511164-110511164 on Build GRCh38
rs978879995 Chr.10: 110511113-110511113 on Build GRCh38
rs767544115 Chr.10: 110511187-110511187 on Build GRCh38
rs980879361 Chr.10: 110510901-110510901 on Build GRCh38
rs1018022571 Chr.10: 110511175-110511175 on Build GRCh38
rs1001706133 Chr.10: 110511022-110511022 on Build GRCh38
rs773766417 Chr.10: 110511038-110511038 on Build GRCh38
rs906125999 Chr.10: 110510767-110510767 on Build GRCh38
rs1019880830 Chr.10: 110511077-110511077 on Build GRCh38
rs760927416 Chr.10: 110510912-110510912 on Build GRCh38
rs988424411 Chr.10: 110510851-110510851 on Build GRCh38
rs1009879029 Chr.10: 110511068-110511068 on Build GRCh38
rs942637063 Chr.10: 110510894-110510894 on Build GRCh38
rs142409394 Chr.10: 110511132-110511132 on Build GRCh38
rs554624859 Chr.10: 110511103-110511103 on Build GRCh38
rs542612784 Chr.10: 110511065-110511065 on Build GRCh38
rs1025183769 Chr.10: 110510898-110510898 on Build GRCh38
rs569339451 Chr.10: 110511055-110511055 on Build GRCh38
rs888937764 Chr.10: 110511001-110511001 on Build GRCh38
rs527330809 Chr.10: 110510900-110510900 on Build GRCh38
rs565310822 Chr.10: 110510802-110510802 on Build GRCh38
rs548145148 Chr.10: 110510881-110510881 on Build GRCh38
rs1035363252 Chr.10: 110510825-110510825 on Build GRCh38

View Full Product Details