Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112153388-112153907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAM
Entrez Gene ID
57678
Gene Name
glycerol-3-phosphate acyltransferase, mitochondrial
Gene Aliases
GPAT, GPAT1
UniGene
Hs.42586
Gene Chromosome Location
Chr.10: 112149864-112215395 on Build GRCh38
Transcripts
NM_001244949.1 NM_020918.5 XM_005269998.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6518275 c.2380G>T p.E794* Chr.10: 112153657-112153657 on Build GRCh38
244741 c.2467C>G p.L823V Chr.10: 112153570-112153570 on Build GRCh38
3434117 c.2443C>G p.R815G Chr.10: 112153594-112153594 on Build GRCh38
4480580 c.2431C>T p.P811S Chr.10: 112153606-112153606 on Build GRCh38
5789024 c.2463T>C p.Y821Y Chr.10: 112153574-112153574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774897716 Chr.10: 112153711-112153712 on Build GRCh38
rs773872084 Chr.10: 112153650-112153650 on Build GRCh38
rs543418172 Chr.10: 112153444-112153444 on Build GRCh38
rs943183478 Chr.10: 112153814-112153814 on Build GRCh38
rs770815376 Chr.10: 112153574-112153574 on Build GRCh38
rs987772123 Chr.10: 112153738-112153738 on Build GRCh38
rs141269875 Chr.10: 112153632-112153632 on Build GRCh38
rs866340670 Chr.10: 112153872-112153872 on Build GRCh38
rs757445515 Chr.10: 112153823-112153823 on Build GRCh38
rs140848655 Chr.10: 112153811-112153811 on Build GRCh38
rs554838657 Chr.10: 112153437-112153437 on Build GRCh38
rs953410339 Chr.10: 112153677-112153677 on Build GRCh38
rs559993696 Chr.10: 112153545-112153545 on Build GRCh38
rs750493652 Chr.10: 112153505-112153505 on Build GRCh38
rs1043575621 Chr.10: 112153457-112153457 on Build GRCh38
rs946521397 Chr.10: 112153778-112153778 on Build GRCh38
rs980895788 Chr.10: 112153788-112153788 on Build GRCh38
rs398054732 Chr.10: 112153881-112153882 on Build GRCh38
rs770917151 Chr.10: 112153684-112153688 on Build GRCh38
rs1986438 Chr.10: 112153860-112153860 on Build GRCh38
rs776502566 Chr.10: 112153698-112153698 on Build GRCh38
rs762302871 Chr.10: 112153606-112153606 on Build GRCh38
rs1053880225 Chr.10: 112153807-112153807 on Build GRCh38
rs201807534 Chr.10: 112153577-112153577 on Build GRCh38
rs573551480 Chr.10: 112153733-112153733 on Build GRCh38
rs369722933 Chr.10: 112153817-112153817 on Build GRCh38
rs745322829 Chr.10: 112153586-112153586 on Build GRCh38
rs375893400 Chr.10: 112153540-112153540 on Build GRCh38
rs780267872 Chr.10: 112153539-112153539 on Build GRCh38
rs71492050 Chr.10: 112153871-112153871 on Build GRCh38
rs371976321 Chr.10: 112153870-112153870 on Build GRCh38
rs35417372 Chr.10: 112153870-112153871 on Build GRCh38
rs577026882 Chr.10: 112153462-112153462 on Build GRCh38
rs755264433 Chr.10: 112153544-112153544 on Build GRCh38
rs781022466 Chr.10: 112153718-112153718 on Build GRCh38
rs146130725 Chr.10: 112153580-112153580 on Build GRCh38
rs17129557 Chr.10: 112153435-112153435 on Build GRCh38
rs140106403 Chr.10: 112153659-112153659 on Build GRCh38
rs149582863 Chr.10: 112153867-112153867 on Build GRCh38
rs776665288 Chr.10: 112153775-112153775 on Build GRCh38
rs970907162 Chr.10: 112153744-112153744 on Build GRCh38
rs757056863 Chr.10: 112153696-112153696 on Build GRCh38
rs150134813 Chr.10: 112153828-112153828 on Build GRCh38
rs576001210 Chr.10: 112153438-112153438 on Build GRCh38
rs1807800 Chr.10: 112153794-112153794 on Build GRCh38
rs889224643 Chr.10: 112153857-112153857 on Build GRCh38
rs371886268 Chr.10: 112153681-112153681 on Build GRCh38
rs928905443 Chr.10: 112153609-112153609 on Build GRCh38
rs530276012 Chr.10: 112153593-112153593 on Build GRCh38
rs901871825 Chr.10: 112153470-112153470 on Build GRCh38
rs936383662 Chr.10: 112153800-112153800 on Build GRCh38
rs945019398 Chr.10: 112153812-112153812 on Build GRCh38
rs768405384 Chr.10: 112153602-112153602 on Build GRCh38
rs748862078 Chr.10: 112153601-112153601 on Build GRCh38
rs558718035 Chr.10: 112153461-112153461 on Build GRCh38
rs745815679 Chr.10: 112153705-112153712 on Build GRCh38
rs993509139 Chr.10: 112153484-112153484 on Build GRCh38
rs936373590 Chr.10: 112153451-112153451 on Build GRCh38
rs972373937 Chr.10: 112153713-112153713 on Build GRCh38
rs774530128 Chr.10: 112153605-112153605 on Build GRCh38
rs769172409 Chr.10: 112153591-112153591 on Build GRCh38
rs901083139 Chr.10: 112153824-112153824 on Build GRCh38
rs1016750705 Chr.10: 112153874-112153874 on Build GRCh38
rs753996445 Chr.10: 112153674-112153674 on Build GRCh38
rs2803607 Chr.10: 112153870-112153870 on Build GRCh38
rs199766613 Chr.10: 112153883-112153883 on Build GRCh38
rs751287587 Chr.10: 112153691-112153691 on Build GRCh38
rs934892467 Chr.10: 112153729-112153729 on Build GRCh38
rs371437205 Chr.10: 112153654-112153654 on Build GRCh38
rs3052610 Chr.10: 112153880-112153882 on Build GRCh38
rs377723956 Chr.10: 112153553-112153553 on Build GRCh38
rs3216019 Chr.10: 112153712-112153712 on Build GRCh38
rs985243828 Chr.10: 112153549-112153549 on Build GRCh38
rs1049552400 Chr.10: 112153850-112153850 on Build GRCh38
rs79304665 Chr.10: 112153871-112153871 on Build GRCh38
rs756270705 Chr.10: 112153534-112153534 on Build GRCh38
rs113388222 Chr.10: 112153869-112153869 on Build GRCh38
rs755101863 Chr.10: 112153679-112153679 on Build GRCh38
rs762157658 Chr.10: 112153869-112153869 on Build GRCh38
rs2297991 Chr.10: 112153464-112153464 on Build GRCh38
rs542202549 Chr.10: 112153625-112153625 on Build GRCh38

View Full Product Details