Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112157894-112158395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAM
Entrez Gene ID
57678
Gene Name
glycerol-3-phosphate acyltransferase, mitochondrial
Gene Aliases
GPAT, GPAT1
UniGene
Hs.42586
Gene Chromosome Location
Chr.10: 112149864-112215395 on Build GRCh38
Transcripts
NM_001244949.1 NM_020918.5 XM_005269998.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011624 c.1956G>A p.Q652Q Chr.10: 112158340-112158340 on Build GRCh38
3665425 c.1940T>G p.V647G Chr.10: 112158356-112158356 on Build GRCh38
5802255 c.1918delT p.Y640fs*8 Chr.10: 112158378-112158378 on Build GRCh38
5039103 c.1947G>T p.K649N Chr.10: 112158349-112158349 on Build GRCh38
6285884 c.1980+7G>A p.? Chr.10: 112158309-112158309 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1027951483 Chr.10: 112158265-112158265 on Build GRCh38
rs777245248 Chr.10: 112158177-112158177 on Build GRCh38
rs1000375951 Chr.10: 112158144-112158144 on Build GRCh38
rs766703401 Chr.10: 112158286-112158286 on Build GRCh38
rs749127175 Chr.10: 112158368-112158368 on Build GRCh38
rs867130844 Chr.10: 112158205-112158205 on Build GRCh38
rs752761962 Chr.10: 112158307-112158307 on Build GRCh38
rs1023531475 Chr.10: 112158110-112158110 on Build GRCh38
rs758392793 Chr.10: 112158328-112158328 on Build GRCh38
rs544956899 Chr.10: 112158132-112158132 on Build GRCh38
rs779127742 Chr.10: 112158178-112158178 on Build GRCh38
rs779945457 Chr.10: 112158353-112158353 on Build GRCh38
rs1011262800 Chr.10: 112158261-112158261 on Build GRCh38
rs1048997866 Chr.10: 112158367-112158367 on Build GRCh38
rs953749750 Chr.10: 112158034-112158034 on Build GRCh38
rs986633811 Chr.10: 112158041-112158041 on Build GRCh38
rs745621628 Chr.10: 112158324-112158324 on Build GRCh38
rs919951500 Chr.10: 112158097-112158097 on Build GRCh38
rs987098706 Chr.10: 112158213-112158213 on Build GRCh38
rs777997024 Chr.10: 112158046-112158046 on Build GRCh38
rs1019904164 Chr.10: 112158304-112158304 on Build GRCh38
rs931344852 Chr.10: 112158121-112158121 on Build GRCh38
rs747200815 Chr.10: 112158342-112158342 on Build GRCh38
rs6585139 Chr.10: 112158075-112158075 on Build GRCh38
rs573020012 Chr.10: 112157980-112157980 on Build GRCh38
rs2792738 Chr.10: 112158194-112158194 on Build GRCh38
rs756726975 Chr.10: 112157944-112157944 on Build GRCh38
rs775301888 Chr.10: 112158339-112158339 on Build GRCh38
rs972672292 Chr.10: 112158076-112158076 on Build GRCh38
rs754253583 Chr.10: 112158287-112158287 on Build GRCh38
rs955724370 Chr.10: 112158207-112158207 on Build GRCh38
rs771327956 Chr.10: 112158352-112158352 on Build GRCh38
rs920455181 Chr.10: 112158309-112158309 on Build GRCh38
rs757301679 Chr.10: 112158159-112158159 on Build GRCh38
rs144873485 Chr.10: 112158337-112158337 on Build GRCh38
rs1054026610 Chr.10: 112158216-112158216 on Build GRCh38
rs964597775 Chr.10: 112158264-112158264 on Build GRCh38
rs1000212317 Chr.10: 112158209-112158209 on Build GRCh38
rs367762052 Chr.10: 112158289-112158289 on Build GRCh38
rs142171427 Chr.10: 112157945-112157945 on Build GRCh38
rs562152284 Chr.10: 112158239-112158239 on Build GRCh38
rs957044790 Chr.10: 112158007-112158007 on Build GRCh38
rs1055025453 Chr.10: 112158157-112158157 on Build GRCh38
rs6585140 Chr.10: 112158224-112158224 on Build GRCh38
rs902679378 Chr.10: 112158196-112158196 on Build GRCh38
rs1031795045 Chr.10: 112158005-112158005 on Build GRCh38
rs989926447 Chr.10: 112158015-112158015 on Build GRCh38
rs1028087393 Chr.10: 112158029-112158029 on Build GRCh38
rs540035072 Chr.10: 112158036-112158036 on Build GRCh38
rs561410806 Chr.10: 112158292-112158292 on Build GRCh38
rs768871941 Chr.10: 112158146-112158146 on Build GRCh38

View Full Product Details