Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113721677-113722184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP7
Entrez Gene ID
840
Gene Name
caspase 7
Gene Aliases
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3
UniGene
Hs.9216
Gene Chromosome Location
Chr.10: 113679162-113730909 on Build GRCh38
Transcripts
NM_001227.4 NM_001267056.1 NM_001267057.1 NM_001267058.1 NM_001320911.1 NM_033338.5 NM_033339.4 NM_033340.3 XM_006718017.3 XM_011540260.1 XM_017016763.1 XM_017016764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6065115 c.310C>T p.L104L Chr.10: 113721713-113721713 on Build GRCh38
1345893 c.469delA p.A159fs*17 Chr.10: 113721773-113721773 on Build GRCh38
3434229 c.380C>T p.A127V Chr.10: 113721684-113721684 on Build GRCh38
7324103 c.283G>A p.E95K Chr.10: 113721686-113721686 on Build GRCh38
7295288 c.474A>G p.K158K Chr.10: 113721778-113721778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763656398 Chr.10: 113721755-113721755 on Build GRCh38
rs974667620 Chr.10: 113721907-113721907 on Build GRCh38
rs377579991 Chr.10: 113721956-113721956 on Build GRCh38
rs775398249 Chr.10: 113721811-113721811 on Build GRCh38
rs374750668 Chr.10: 113722100-113722100 on Build GRCh38
rs115952269 Chr.10: 113722028-113722028 on Build GRCh38
rs907967323 Chr.10: 113721969-113721969 on Build GRCh38
rs368309877 Chr.10: 113721766-113721766 on Build GRCh38
rs992236169 Chr.10: 113722007-113722007 on Build GRCh38
rs1028832385 Chr.10: 113721908-113721909 on Build GRCh38
rs143659216 Chr.10: 113721697-113721697 on Build GRCh38
rs202060178 Chr.10: 113721707-113721707 on Build GRCh38
rs748363388 Chr.10: 113721802-113721802 on Build GRCh38
rs933964814 Chr.10: 113722034-113722034 on Build GRCh38
rs1009211567 Chr.10: 113722143-113722143 on Build GRCh38
rs181037368 Chr.10: 113722160-113722160 on Build GRCh38
rs1021133243 Chr.10: 113722148-113722148 on Build GRCh38
rs540474794 Chr.10: 113721824-113721824 on Build GRCh38
rs985007139 Chr.10: 113721944-113721944 on Build GRCh38
rs940798728 Chr.10: 113721972-113721972 on Build GRCh38
rs138342520 Chr.10: 113722142-113722142 on Build GRCh38
rs527334380 Chr.10: 113722112-113722112 on Build GRCh38
rs768132549 Chr.10: 113721821-113721821 on Build GRCh38
rs1019052518 Chr.10: 113721820-113721820 on Build GRCh38
rs539802026 Chr.10: 113721762-113721762 on Build GRCh38
rs1038529000 Chr.10: 113722099-113722099 on Build GRCh38
rs189137028 Chr.10: 113722104-113722104 on Build GRCh38
rs1006141561 Chr.10: 113722098-113722098 on Build GRCh38
rs544547687 Chr.10: 113721895-113721895 on Build GRCh38
rs1047153209 Chr.10: 113722038-113722038 on Build GRCh38
rs868780443 Chr.10: 113721847-113721847 on Build GRCh38
rs2229998 Chr.10: 113721712-113721712 on Build GRCh38
rs754590030 Chr.10: 113721908-113721908 on Build GRCh38
rs772140271 Chr.10: 113721806-113721806 on Build GRCh38
rs758096353 Chr.10: 113722064-113722064 on Build GRCh38
rs569130136 Chr.10: 113721760-113721760 on Build GRCh38
rs574290212 Chr.10: 113721864-113721864 on Build GRCh38
rs982007571 Chr.10: 113721979-113721979 on Build GRCh38
rs750105794 Chr.10: 113721830-113721830 on Build GRCh38
rs372563999 Chr.10: 113721809-113721809 on Build GRCh38
rs979716396 Chr.10: 113722153-113722153 on Build GRCh38
rs563173342 Chr.10: 113721902-113721902 on Build GRCh38
rs748453320 Chr.10: 113721725-113721725 on Build GRCh38
rs770732568 Chr.10: 113721730-113721730 on Build GRCh38
rs144555540 Chr.10: 113721739-113721739 on Build GRCh38
rs762432682 Chr.10: 113721744-113721744 on Build GRCh38
rs779179291 Chr.10: 113721778-113721778 on Build GRCh38
rs887160576 Chr.10: 113722050-113722050 on Build GRCh38
rs751261334 Chr.10: 113722159-113722159 on Build GRCh38
rs115421189 Chr.10: 113721769-113721769 on Build GRCh38
rs759028666 Chr.10: 113721827-113721827 on Build GRCh38
rs892618753 Chr.10: 113722123-113722123 on Build GRCh38
rs1002437068 Chr.10: 113722128-113722128 on Build GRCh38
rs759399881 Chr.10: 113721737-113721737 on Build GRCh38
rs913738779 Chr.10: 113721894-113721894 on Build GRCh38
rs776342160 Chr.10: 113721731-113721731 on Build GRCh38
rs146796754 Chr.10: 113721708-113721708 on Build GRCh38
rs755104278 Chr.10: 113721773-113721773 on Build GRCh38
rs557341293 Chr.10: 113721724-113721724 on Build GRCh38
rs376577730 Chr.10: 113721819-113721819 on Build GRCh38
rs555497840 Chr.10: 113721828-113721828 on Build GRCh38
rs544492071 Chr.10: 113722150-113722150 on Build GRCh38
rs765015515 Chr.10: 113721738-113721738 on Build GRCh38
rs61866465 Chr.10: 113721905-113721905 on Build GRCh38
rs532823560 Chr.10: 113722016-113722016 on Build GRCh38
rs111894827 Chr.10: 113722138-113722138 on Build GRCh38
rs769685192 Chr.10: 113721810-113721810 on Build GRCh38
rs951884306 Chr.10: 113721918-113721918 on Build GRCh38

View Full Product Details