Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113725283-113725781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP7
Entrez Gene ID
840
Gene Name
caspase 7
Gene Aliases
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3
UniGene
Hs.9216
Gene Chromosome Location
Chr.10: 113679162-113730909 on Build GRCh38
Transcripts
NM_001227.4 NM_001267056.1 NM_001267057.1 NM_001267058.1 NM_001320911.1 NM_033338.5 NM_033339.4 NM_033340.3 XM_006718017.3 XM_011540260.1 XM_017016763.1 XM_017016764.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2053626 c.625G>A p.E209K Chr.10: 113725511-113725511 on Build GRCh38
2053624 c.516C>T p.C172C Chr.10: 113725402-113725402 on Build GRCh38
3434230 c.577A>G p.K193E Chr.10: 113725463-113725463 on Build GRCh38
7201078 c.546A>G p.V182V Chr.10: 113725432-113725432 on Build GRCh38
4816594 c.645C>T p.F215F Chr.10: 113725531-113725531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951468740 Chr.10: 113725743-113725743 on Build GRCh38
rs181033605 Chr.10: 113725687-113725687 on Build GRCh38
rs981503980 Chr.10: 113725751-113725751 on Build GRCh38
rs748184526 Chr.10: 113725523-113725523 on Build GRCh38
rs779873226 Chr.10: 113725388-113725388 on Build GRCh38
rs199687948 Chr.10: 113725543-113725543 on Build GRCh38
rs151040178 Chr.10: 113725728-113725728 on Build GRCh38
rs779589082 Chr.10: 113725340-113725340 on Build GRCh38
rs760936227 Chr.10: 113725311-113725311 on Build GRCh38
rs770669926 Chr.10: 113725531-113725531 on Build GRCh38
rs75516871 Chr.10: 113725519-113725519 on Build GRCh38
rs143673557 Chr.10: 113725695-113725695 on Build GRCh38
rs1018566256 Chr.10: 113725392-113725392 on Build GRCh38
rs201040237 Chr.10: 113725497-113725497 on Build GRCh38
rs1028094544 Chr.10: 113725572-113725572 on Build GRCh38
rs768539831 Chr.10: 113725398-113725398 on Build GRCh38
rs1018160002 Chr.10: 113725681-113725681 on Build GRCh38
rs759963752 Chr.10: 113725564-113725564 on Build GRCh38
rs749103359 Chr.10: 113725397-113725397 on Build GRCh38
rs570242654 Chr.10: 113725355-113725355 on Build GRCh38
rs558082924 Chr.10: 113725404-113725404 on Build GRCh38
rs764271040 Chr.10: 113725485-113725485 on Build GRCh38
rs999219006 Chr.10: 113725649-113725649 on Build GRCh38
rs1006797838 Chr.10: 113725677-113725677 on Build GRCh38
rs773710326 Chr.10: 113725517-113725517 on Build GRCh38
rs374492870 Chr.10: 113725612-113725612 on Build GRCh38
rs754365567 Chr.10: 113725318-113725318 on Build GRCh38
rs765774624 Chr.10: 113725474-113725474 on Build GRCh38
rs763050011 Chr.10: 113725396-113725396 on Build GRCh38
rs11592536 Chr.10: 113725641-113725641 on Build GRCh38
rs141266925 Chr.10: 113725526-113725526 on Build GRCh38
rs375167870 Chr.10: 113725339-113725339 on Build GRCh38
rs779523386 Chr.10: 113725424-113725425 on Build GRCh38
rs754779330 Chr.10: 113725514-113725514 on Build GRCh38
rs4414176 Chr.10: 113725715-113725715 on Build GRCh38
rs369880412 Chr.10: 113725577-113725577 on Build GRCh38
rs541644893 Chr.10: 113725566-113725566 on Build GRCh38
rs536786215 Chr.10: 113725445-113725445 on Build GRCh38
rs928848795 Chr.10: 113725709-113725709 on Build GRCh38
rs372396520 Chr.10: 113725386-113725386 on Build GRCh38
rs760415288 Chr.10: 113725415-113725415 on Build GRCh38
rs878911865 Chr.10: 113725668-113725668 on Build GRCh38
rs746527405 Chr.10: 113725529-113725529 on Build GRCh38
rs776431264 Chr.10: 113725548-113725548 on Build GRCh38
rs760013015 Chr.10: 113725473-113725473 on Build GRCh38
rs747318342 Chr.10: 113725384-113725384 on Build GRCh38
rs193124738 Chr.10: 113725471-113725471 on Build GRCh38
rs1019926611 Chr.10: 113725582-113725582 on Build GRCh38
rs763561638 Chr.10: 113725587-113725587 on Build GRCh38
rs766918526 Chr.10: 113725315-113725315 on Build GRCh38
rs116709623 Chr.10: 113725480-113725480 on Build GRCh38
rs772829055 Chr.10: 113725528-113725528 on Build GRCh38
rs916066138 Chr.10: 113725675-113725675 on Build GRCh38
rs758516148 Chr.10: 113725349-113725349 on Build GRCh38
rs532672396 Chr.10: 113725644-113725644 on Build GRCh38
rs953992859 Chr.10: 113725575-113725575 on Build GRCh38
rs761367047 Chr.10: 113725430-113725430 on Build GRCh38
rs143369246 Chr.10: 113725620-113725620 on Build GRCh38
rs537094698 Chr.10: 113725399-113725399 on Build GRCh38
rs143354631 Chr.10: 113725486-113725486 on Build GRCh38
rs984978820 Chr.10: 113725662-113725662 on Build GRCh38
rs748986164 Chr.10: 113725484-113725485 on Build GRCh38
rs752681631 Chr.10: 113725344-113725344 on Build GRCh38
rs372251562 Chr.10: 113725755-113725755 on Build GRCh38
rs112225268 Chr.10: 113725651-113725651 on Build GRCh38
rs188201053 Chr.10: 113725591-113725591 on Build GRCh38
rs778332173 Chr.10: 113725729-113725729 on Build GRCh38
rs374718188 Chr.10: 113725408-113725408 on Build GRCh38
rs955320166 Chr.10: 113725338-113725338 on Build GRCh38
rs1056047198 Chr.10: 113725573-113725573 on Build GRCh38
rs369067741 Chr.10: 113725476-113725476 on Build GRCh38
rs934379984 Chr.10: 113725727-113725727 on Build GRCh38

View Full Product Details