Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113730007-113730553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP7
Entrez Gene ID
840
Gene Name
caspase 7
Gene Aliases
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3
UniGene
Hs.9216
Gene Chromosome Location
Chr.10: 113679162-113730909 on Build GRCh38
Transcripts
NM_001227.4 NM_001267056.1 NM_001267057.1 NM_001267058.1 NM_001320911.1 NM_033338.5 NM_033339.4 NM_033340.3 XM_006718017.3 XM_011540260.1 XM_017016763.1 XM_017016764.1

SNP Detail

SNP ID Chromosome Location
rs12247588 Chr.10: 113730431-113730431 on Build GRCh38
rs576757788 Chr.10: 113730210-113730210 on Build GRCh38
rs907721175 Chr.10: 113730398-113730398 on Build GRCh38
rs971752841 Chr.10: 113730524-113730524 on Build GRCh38
rs572010376 Chr.10: 113730359-113730359 on Build GRCh38
rs998431583 Chr.10: 113730412-113730412 on Build GRCh38
rs560518113 Chr.10: 113730274-113730274 on Build GRCh38
rs1028951014 Chr.10: 113730416-113730416 on Build GRCh38
rs1055115409 Chr.10: 113730530-113730530 on Build GRCh38
rs565689460 Chr.10: 113730490-113730490 on Build GRCh38
rs181350515 Chr.10: 113730306-113730306 on Build GRCh38
rs79838074 Chr.10: 113730228-113730228 on Build GRCh38
rs1032515979 Chr.10: 113730254-113730254 on Build GRCh38
rs541386567 Chr.10: 113730461-113730461 on Build GRCh38
rs1055652417 Chr.10: 113730046-113730046 on Build GRCh38
rs1007324231 Chr.10: 113730463-113730463 on Build GRCh38
rs553514657 Chr.10: 113730487-113730487 on Build GRCh38
rs953275087 Chr.10: 113730062-113730062 on Build GRCh38
rs1015982354 Chr.10: 113730481-113730481 on Build GRCh38
rs141018894 Chr.10: 113730102-113730102 on Build GRCh38
rs770653556 Chr.10: 113730196-113730196 on Build GRCh38
rs762846988 Chr.10: 113730140-113730140 on Build GRCh38
rs535836568 Chr.10: 113730495-113730495 on Build GRCh38
rs556922705 Chr.10: 113730388-113730388 on Build GRCh38
rs954466761 Chr.10: 113730380-113730380 on Build GRCh38
rs531946102 Chr.10: 113730318-113730318 on Build GRCh38
rs554656613 Chr.10: 113730529-113730529 on Build GRCh38
rs901453575 Chr.10: 113730405-113730405 on Build GRCh38
rs748415913 Chr.10: 113730478-113730478 on Build GRCh38
rs1025130085 Chr.10: 113730110-113730110 on Build GRCh38
rs895229264 Chr.10: 113730077-113730077 on Build GRCh38
rs1127687 Chr.10: 113730350-113730350 on Build GRCh38
rs545036860 Chr.10: 113730247-113730247 on Build GRCh38
rs987739220 Chr.10: 113730395-113730395 on Build GRCh38
rs116185385 Chr.10: 113730314-113730314 on Build GRCh38
rs762437935 Chr.10: 113730501-113730501 on Build GRCh38
rs985876225 Chr.10: 113730064-113730064 on Build GRCh38
rs999752427 Chr.10: 113730240-113730240 on Build GRCh38
rs893808991 Chr.10: 113730362-113730362 on Build GRCh38
rs982376818 Chr.10: 113730273-113730273 on Build GRCh38
rs12247479 Chr.10: 113730301-113730301 on Build GRCh38
rs940481181 Chr.10: 113730409-113730409 on Build GRCh38
rs543924275 Chr.10: 113730272-113730272 on Build GRCh38
rs963059553 Chr.10: 113730514-113730514 on Build GRCh38

View Full Product Details