Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114288846-114289349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1L2
Entrez Gene ID
84632
Gene Name
actin filament associated protein 1 like 2
Gene Aliases
CTB-1144G6.4, KIAA1914, XB130
UniGene
Hs.501106
Gene Chromosome Location
Chr.10: 114280730-114405143 on Build GRCh38
Transcripts
XM_005270233.4 XM_005270237.4 XM_005270241.4 XM_005270242.4 XM_011540286.2 XM_017016792.1 XM_017016793.1 XM_017016794.1 XM_017016795.1 XM_017016796.1 XM_017016797.1 XM_017016798.1 XM_017016804.1 XM_017016814.1 XM_017016815.1 XM_017016818.1 XM_017016829.1 XM_017016832.1 NM_001272046.1 NM_001320804.1 XM_011539754.2 XM_011539757.2 XM_017016177.1 XM_017016178.1 XM_017016179.1 XM_017016180.1

Gene Symbol
VWA2
Entrez Gene ID
340706
Gene Name
von Willebrand factor A domain containing 2
Gene Aliases
AMACO, CCSP-2, NET42
UniGene
Hs.197741
Gene Chromosome Location
Chr.10: 114239254-114294500 on Build GRCh38
Transcripts
XM_005270233.4 XM_005270237.4 XM_005270241.4 XM_005270242.4 XM_011540286.2 XM_017016792.1 XM_017016793.1 XM_017016794.1 XM_017016795.1 XM_017016796.1 XM_017016797.1 XM_017016798.1 XM_017016804.1 XM_017016814.1 XM_017016815.1 XM_017016818.1 XM_017016829.1 XM_017016832.1 NM_001272046.1 NM_001320804.1 XM_011539754.2 XM_011539757.2 XM_017016177.1 XM_017016178.1 XM_017016179.1 XM_017016180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011770 c.1779G>A p.A593A Chr.10: 114289146-114289146 on Build GRCh38
458902 c.1749C>T p.F583F Chr.10: 114289116-114289116 on Build GRCh38
5541632 c.1689G>A p.V563V Chr.10: 114289056-114289056 on Build GRCh38
7397477 c.1684G>T p.E562* Chr.10: 114289051-114289051 on Build GRCh38
2054032 c.1771C>T p.R591W Chr.10: 114289138-114289138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748964738 Chr.10: 114289146-114289146 on Build GRCh38
rs376601832 Chr.10: 114288881-114288881 on Build GRCh38
rs754575985 Chr.10: 114289305-114289305 on Build GRCh38
rs778517109 Chr.10: 114289306-114289306 on Build GRCh38
rs755950349 Chr.10: 114289224-114289224 on Build GRCh38
rs757112781 Chr.10: 114289208-114289208 on Build GRCh38
rs533160127 Chr.10: 114289059-114289059 on Build GRCh38
rs867469596 Chr.10: 114289249-114289249 on Build GRCh38
rs761247064 Chr.10: 114289272-114289272 on Build GRCh38
rs755328335 Chr.10: 114288929-114288929 on Build GRCh38
rs969728576 Chr.10: 114288943-114288943 on Build GRCh38
rs747714601 Chr.10: 114289252-114289252 on Build GRCh38
rs769380790 Chr.10: 114289139-114289139 on Build GRCh38
rs768137633 Chr.10: 114289066-114289066 on Build GRCh38
rs771831377 Chr.10: 114288889-114288889 on Build GRCh38
rs866382749 Chr.10: 114289051-114289051 on Build GRCh38
rs759962614 Chr.10: 114288904-114288905 on Build GRCh38
rs1044548082 Chr.10: 114288880-114288880 on Build GRCh38
rs190448182 Chr.10: 114289000-114289000 on Build GRCh38
rs777891947 Chr.10: 114289200-114289200 on Build GRCh38
rs140567963 Chr.10: 114289158-114289158 on Build GRCh38
rs568092737 Chr.10: 114289078-114289078 on Build GRCh38
rs763040664 Chr.10: 114289294-114289294 on Build GRCh38
rs771555917 Chr.10: 114289314-114289314 on Build GRCh38
rs770449464 Chr.10: 114288911-114288911 on Build GRCh38
rs906022290 Chr.10: 114289179-114289179 on Build GRCh38
rs759658035 Chr.10: 114289290-114289290 on Build GRCh38
rs765498186 Chr.10: 114289204-114289204 on Build GRCh38
rs763423732 Chr.10: 114289273-114289274 on Build GRCh38
rs781235341 Chr.10: 114289127-114289127 on Build GRCh38
rs147215815 Chr.10: 114289154-114289154 on Build GRCh38
rs767751676 Chr.10: 114289174-114289174 on Build GRCh38
rs764526217 Chr.10: 114288964-114288964 on Build GRCh38
rs776426558 Chr.10: 114289008-114289008 on Build GRCh38
rs1019355745 Chr.10: 114289268-114289268 on Build GRCh38
rs929413917 Chr.10: 114289123-114289123 on Build GRCh38
rs200921074 Chr.10: 114288883-114288883 on Build GRCh38
rs774196736 Chr.10: 114289247-114289247 on Build GRCh38
rs779718596 Chr.10: 114289229-114289229 on Build GRCh38
rs144705004 Chr.10: 114289300-114289300 on Build GRCh38
rs111616652 Chr.10: 114289064-114289064 on Build GRCh38
rs149792922 Chr.10: 114289295-114289295 on Build GRCh38
rs535139009 Chr.10: 114289180-114289180 on Build GRCh38
rs761038837 Chr.10: 114289186-114289186 on Build GRCh38
rs535731405 Chr.10: 114289080-114289080 on Build GRCh38
rs149262685 Chr.10: 114289001-114289001 on Build GRCh38
rs200257950 Chr.10: 114289213-114289213 on Build GRCh38
rs745844089 Chr.10: 114289220-114289220 on Build GRCh38
rs887635332 Chr.10: 114289141-114289141 on Build GRCh38
rs780382305 Chr.10: 114288985-114288985 on Build GRCh38
rs201102050 Chr.10: 114289075-114289075 on Build GRCh38
rs750452924 Chr.10: 114288976-114288976 on Build GRCh38
rs34757551 Chr.10: 114288999-114288999 on Build GRCh38
rs773174149 Chr.10: 114288892-114288892 on Build GRCh38
rs766858511 Chr.10: 114289279-114289279 on Build GRCh38
rs199710371 Chr.10: 114288963-114288963 on Build GRCh38
rs777106857 Chr.10: 114289287-114289287 on Build GRCh38
rs769160519 Chr.10: 114289035-114289035 on Build GRCh38
rs531061260 Chr.10: 114288995-114288995 on Build GRCh38
rs759139603 Chr.10: 114289009-114289009 on Build GRCh38
rs201659824 Chr.10: 114289040-114289040 on Build GRCh38
rs781146365 Chr.10: 114288971-114288971 on Build GRCh38
rs143465186 Chr.10: 114289116-114289116 on Build GRCh38
rs760563857 Chr.10: 114288898-114288898 on Build GRCh38
rs372729237 Chr.10: 114289153-114289153 on Build GRCh38
rs368163458 Chr.10: 114289315-114289315 on Build GRCh38
rs544925685 Chr.10: 114289303-114289303 on Build GRCh38
rs200171942 Chr.10: 114289138-114289138 on Build GRCh38
rs776027827 Chr.10: 114289250-114289250 on Build GRCh38
rs150763897 Chr.10: 114289281-114289281 on Build GRCh38
rs1002967042 Chr.10: 114288960-114288960 on Build GRCh38
rs199884277 Chr.10: 114289065-114289065 on Build GRCh38
rs752274903 Chr.10: 114288944-114288944 on Build GRCh38
rs762658059 Chr.10: 114289045-114289045 on Build GRCh38
rs750122369 Chr.10: 114289297-114289297 on Build GRCh38
rs775094333 Chr.10: 114289145-114289145 on Build GRCh38
rs891878881 Chr.10: 114289246-114289246 on Build GRCh38
rs202219404 Chr.10: 114289236-114289236 on Build GRCh38
rs376331052 Chr.10: 114289237-114289237 on Build GRCh38
rs764577364 Chr.10: 114289047-114289047 on Build GRCh38
rs776146572 Chr.10: 114288925-114288925 on Build GRCh38
rs35749056 Chr.10: 114289247-114289247 on Build GRCh38
rs751918496 Chr.10: 114289053-114289053 on Build GRCh38
rs747850496 Chr.10: 114289308-114289308 on Build GRCh38
rs756311283 Chr.10: 114289140-114289140 on Build GRCh38
rs143776898 Chr.10: 114289119-114289119 on Build GRCh38
rs750881586 Chr.10: 114289068-114289068 on Build GRCh38
rs1023037279 Chr.10: 114288899-114288899 on Build GRCh38
rs765003591 Chr.10: 114288936-114288936 on Build GRCh38
rs747387911 Chr.10: 114289318-114289318 on Build GRCh38
rs756143333 Chr.10: 114288980-114288980 on Build GRCh38
rs765575743 Chr.10: 114289069-114289070 on Build GRCh38
rs142806839 Chr.10: 114289203-114289203 on Build GRCh38
rs10082357 Chr.10: 114289291-114289291 on Build GRCh38
rs376160370 Chr.10: 114289299-114289299 on Build GRCh38
rs371230333 Chr.10: 114289253-114289253 on Build GRCh38
rs377210274 Chr.10: 114289198-114289198 on Build GRCh38
rs143337523 Chr.10: 114289117-114289117 on Build GRCh38
rs755133926 Chr.10: 114289089-114289089 on Build GRCh38
rs752890861 Chr.10: 114289207-114289207 on Build GRCh38
rs950205258 Chr.10: 114288867-114288867 on Build GRCh38
rs1040404585 Chr.10: 114289102-114289102 on Build GRCh38

View Full Product Details