Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10304970-10305461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM
Entrez Gene ID
133
Gene Name
adrenomedullin
Gene Aliases
AM, PAMP
UniGene
Hs.441047
Gene Chromosome Location
Chr.11: 10304980-10307402 on Build GRCh38
Transcripts
NM_001124.2

SNP Detail

SNP ID Chromosome Location
rs1050973266 Chr.11: 10305141-10305141 on Build GRCh38
rs945535489 Chr.11: 10305035-10305035 on Build GRCh38
rs967667211 Chr.11: 10305425-10305425 on Build GRCh38
rs995170152 Chr.11: 10305210-10305210 on Build GRCh38
rs34570663 Chr.11: 10305343-10305343 on Build GRCh38
rs933383844 Chr.11: 10305121-10305121 on Build GRCh38
rs1033305560 Chr.11: 10305211-10305211 on Build GRCh38
rs796450021 Chr.11: 10305245-10305245 on Build GRCh38
rs917305638 Chr.11: 10305208-10305208 on Build GRCh38
rs1018669605 Chr.11: 10305079-10305079 on Build GRCh38
rs900626965 Chr.11: 10305118-10305118 on Build GRCh38
rs930655669 Chr.11: 10305289-10305289 on Build GRCh38
rs925559617 Chr.11: 10305350-10305350 on Build GRCh38
rs13306112 Chr.11: 10305294-10305294 on Build GRCh38
rs1015470545 Chr.11: 10305185-10305185 on Build GRCh38
rs1028611391 Chr.11: 10305122-10305122 on Build GRCh38
rs1000505179 Chr.11: 10305431-10305431 on Build GRCh38
rs553327430 Chr.11: 10305288-10305288 on Build GRCh38
rs950175920 Chr.11: 10305262-10305262 on Build GRCh38
rs902047117 Chr.11: 10305376-10305376 on Build GRCh38
rs966197275 Chr.11: 10305096-10305096 on Build GRCh38
rs750953470 Chr.11: 10305178-10305178 on Build GRCh38
rs973670448 Chr.11: 10305015-10305015 on Build GRCh38
rs992415077 Chr.11: 10305206-10305206 on Build GRCh38
rs540228856 Chr.11: 10305189-10305189 on Build GRCh38
rs1044424415 Chr.11: 10305012-10305012 on Build GRCh38
rs565195866 Chr.11: 10305410-10305410 on Build GRCh38
rs979303129 Chr.11: 10305174-10305174 on Build GRCh38
rs5003 Chr.11: 10305190-10305190 on Build GRCh38
rs890967718 Chr.11: 10305167-10305167 on Build GRCh38
rs1044533670 Chr.11: 10305273-10305273 on Build GRCh38
rs974763208 Chr.11: 10305415-10305415 on Build GRCh38
rs1049370955 Chr.11: 10305017-10305017 on Build GRCh38
rs886589982 Chr.11: 10305018-10305018 on Build GRCh38
rs549878905 Chr.11: 10305014-10305014 on Build GRCh38
rs967280263 Chr.11: 10305325-10305325 on Build GRCh38
rs532427130 Chr.11: 10305377-10305377 on Build GRCh38
rs569455947 Chr.11: 10305034-10305034 on Build GRCh38
rs967719363 Chr.11: 10305130-10305130 on Build GRCh38
rs542301603 Chr.11: 10305318-10305318 on Build GRCh38
rs1015439333 Chr.11: 10305003-10305003 on Build GRCh38
rs558421832 Chr.11: 10305114-10305114 on Build GRCh38
rs562372223 Chr.11: 10305330-10305330 on Build GRCh38
rs577604305 Chr.11: 10305142-10305142 on Build GRCh38
rs13306111 Chr.11: 10305010-10305010 on Build GRCh38
rs925486168 Chr.11: 10305111-10305111 on Build GRCh38
rs1004026634 Chr.11: 10305169-10305169 on Build GRCh38
rs905902471 Chr.11: 10305013-10305013 on Build GRCh38
rs538182588 Chr.11: 10305113-10305113 on Build GRCh38
rs545015719 Chr.11: 10305355-10305355 on Build GRCh38
rs1028727341 Chr.11: 10305403-10305403 on Build GRCh38
rs886705129 Chr.11: 10305302-10305302 on Build GRCh38
rs978355785 Chr.11: 10305039-10305039 on Build GRCh38

View Full Product Details