Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10307009-10307504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM
Entrez Gene ID
133
Gene Name
adrenomedullin
Gene Aliases
AM, PAMP
UniGene
Hs.441047
Gene Chromosome Location
Chr.11: 10304980-10307402 on Build GRCh38
Transcripts
NM_001124.2

SNP Detail

SNP ID Chromosome Location
rs983795406 Chr.11: 10307057-10307057 on Build GRCh38
rs551565001 Chr.11: 10307250-10307250 on Build GRCh38
rs184443338 Chr.11: 10307440-10307440 on Build GRCh38
rs4698 Chr.11: 10307220-10307220 on Build GRCh38
rs756289325 Chr.11: 10307312-10307312 on Build GRCh38
rs1049304694 Chr.11: 10307329-10307329 on Build GRCh38
rs1033397074 Chr.11: 10307441-10307441 on Build GRCh38
rs953080044 Chr.11: 10307216-10307216 on Build GRCh38
rs549764311 Chr.11: 10307409-10307409 on Build GRCh38
rs35440828 Chr.11: 10307196-10307196 on Build GRCh38
rs747812735 Chr.11: 10307484-10307484 on Build GRCh38
rs1012953715 Chr.11: 10307378-10307378 on Build GRCh38
rs200599471 Chr.11: 10307206-10307206 on Build GRCh38
rs377262939 Chr.11: 10307460-10307460 on Build GRCh38
rs1007474908 Chr.11: 10307215-10307215 on Build GRCh38
rs965655002 Chr.11: 10307485-10307485 on Build GRCh38
rs893832147 Chr.11: 10307252-10307252 on Build GRCh38
rs778532135 Chr.11: 10307479-10307479 on Build GRCh38
rs565520785 Chr.11: 10307267-10307267 on Build GRCh38
rs930788702 Chr.11: 10307277-10307277 on Build GRCh38
rs561156410 Chr.11: 10307134-10307134 on Build GRCh38
rs531869867 Chr.11: 10307246-10307246 on Build GRCh38
rs543043922 Chr.11: 10307209-10307209 on Build GRCh38
rs572125585 Chr.11: 10307115-10307115 on Build GRCh38
rs939951805 Chr.11: 10307088-10307088 on Build GRCh38
rs919438478 Chr.11: 10307259-10307259 on Build GRCh38
rs1010708353 Chr.11: 10307418-10307418 on Build GRCh38
rs780374999 Chr.11: 10307368-10307368 on Build GRCh38
rs900993429 Chr.11: 10307142-10307142 on Build GRCh38
rs1022244463 Chr.11: 10307423-10307423 on Build GRCh38
rs944433043 Chr.11: 10307249-10307249 on Build GRCh38
rs907310284 Chr.11: 10307387-10307387 on Build GRCh38
rs199897268 Chr.11: 10307205-10307205 on Build GRCh38
rs74834059 Chr.11: 10307200-10307200 on Build GRCh38
rs933836969 Chr.11: 10307155-10307155 on Build GRCh38
rs757767382 Chr.11: 10307064-10307064 on Build GRCh38
rs201588733 Chr.11: 10307207-10307207 on Build GRCh38
rs967279959 Chr.11: 10307439-10307439 on Build GRCh38
rs397737206 Chr.11: 10307204-10307205 on Build GRCh38
rs753320727 Chr.11: 10307175-10307175 on Build GRCh38
rs34354539 Chr.11: 10307200-10307201 on Build GRCh38
rs754519436 Chr.11: 10307263-10307263 on Build GRCh38
rs959115538 Chr.11: 10307452-10307452 on Build GRCh38
rs541381616 Chr.11: 10307120-10307120 on Build GRCh38
rs5008 Chr.11: 10307335-10307335 on Build GRCh38
rs34056369 Chr.11: 10307384-10307384 on Build GRCh38
rs937630115 Chr.11: 10307055-10307055 on Build GRCh38
rs192534446 Chr.11: 10307393-10307393 on Build GRCh38
rs537248444 Chr.11: 10307067-10307067 on Build GRCh38
rs114485041 Chr.11: 10307478-10307478 on Build GRCh38
rs894494120 Chr.11: 10307116-10307117 on Build GRCh38
rs1051768156 Chr.11: 10307191-10307191 on Build GRCh38
rs911682659 Chr.11: 10307235-10307235 on Build GRCh38
rs1024402641 Chr.11: 10307135-10307136 on Build GRCh38
rs1126716 Chr.11: 10307321-10307321 on Build GRCh38
rs995037571 Chr.11: 10307081-10307081 on Build GRCh38
rs1021646099 Chr.11: 10307332-10307332 on Build GRCh38
rs1031733302 Chr.11: 10307401-10307401 on Build GRCh38
rs145100579 Chr.11: 10307395-10307395 on Build GRCh38

View Full Product Details