Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 11352996-11353493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK2A3
Entrez Gene ID
283106
Gene Name
casein kinase 2 alpha 3
Gene Aliases
CSNK2A1P
UniGene
Hs.654675
Gene Chromosome Location
Chr.11: 11351942-11353357 on Build GRCh38
Transcripts
NM_001256686.1 NM_198516.2 XM_006718224.2 XM_011520068.2 XM_011520071.2

Gene Symbol
GALNT18
Entrez Gene ID
374378
Gene Name
polypeptide N-acetylgalactosaminyltransferase 18
Gene Aliases
GALNACT18, GALNT15, GALNTL4, GalNAc-T15, GalNAc-T18
UniGene
Hs.655152
Gene Chromosome Location
Chr.11: 11270874-11622045 on Build GRCh38
Transcripts
NM_001256686.1 NM_198516.2 XM_006718224.2 XM_011520068.2 XM_011520071.2

SNP Detail

SNP ID Chromosome Location
rs1047562496 Chr.11: 11353248-11353248 on Build GRCh38
rs998980759 Chr.11: 11353323-11353323 on Build GRCh38
rs550139220 Chr.11: 11353364-11353364 on Build GRCh38
rs534275803 Chr.11: 11353293-11353293 on Build GRCh38
rs755996286 Chr.11: 11353059-11353059 on Build GRCh38
rs113276278 Chr.11: 11353135-11353135 on Build GRCh38
rs2129523 Chr.11: 11353382-11353382 on Build GRCh38
rs1014739053 Chr.11: 11353358-11353358 on Build GRCh38
rs755594317 Chr.11: 11353138-11353138 on Build GRCh38
rs201527804 Chr.11: 11353291-11353291 on Build GRCh38
rs758556224 Chr.11: 11353080-11353080 on Build GRCh38
rs746004196 Chr.11: 11353079-11353079 on Build GRCh38
rs532739784 Chr.11: 11353404-11353404 on Build GRCh38
rs944502759 Chr.11: 11353455-11353455 on Build GRCh38
rs970479746 Chr.11: 11353378-11353378 on Build GRCh38
rs906065541 Chr.11: 11353405-11353405 on Build GRCh38
rs757468139 Chr.11: 11353072-11353072 on Build GRCh38
rs921679884 Chr.11: 11353465-11353465 on Build GRCh38
rs747544221 Chr.11: 11353091-11353091 on Build GRCh38
rs753636414 Chr.11: 11353108-11353108 on Build GRCh38
rs748527421 Chr.11: 11353095-11353095 on Build GRCh38
rs987041459 Chr.11: 11353187-11353187 on Build GRCh38
rs748534656 Chr.11: 11353144-11353144 on Build GRCh38
rs1043721187 Chr.11: 11353294-11353294 on Build GRCh38
rs779184038 Chr.11: 11353078-11353078 on Build GRCh38
rs773788910 Chr.11: 11353041-11353041 on Build GRCh38
rs577098272 Chr.11: 11353260-11353260 on Build GRCh38
rs370595804 Chr.11: 11353340-11353340 on Build GRCh38
rs186628667 Chr.11: 11353344-11353344 on Build GRCh38
rs750687783 Chr.11: 11353118-11353118 on Build GRCh38
rs911046789 Chr.11: 11353019-11353019 on Build GRCh38
rs535086321 Chr.11: 11353464-11353464 on Build GRCh38
rs368248470 Chr.11: 11353058-11353058 on Build GRCh38
rs878934200 Chr.11: 11353242-11353242 on Build GRCh38
rs759127002 Chr.11: 11353098-11353098 on Build GRCh38
rs115934936 Chr.11: 11353341-11353341 on Build GRCh38
rs148146417 Chr.11: 11353252-11353253 on Build GRCh38
rs904749694 Chr.11: 11353305-11353305 on Build GRCh38
rs539005921 Chr.11: 11353089-11353089 on Build GRCh38
rs781718630 Chr.11: 11353143-11353143 on Build GRCh38
rs751405463 Chr.11: 11353045-11353045 on Build GRCh38
rs566174971 Chr.11: 11353096-11353096 on Build GRCh38
rs780225559 Chr.11: 11353082-11353082 on Build GRCh38
rs2129524 Chr.11: 11353267-11353267 on Build GRCh38
rs764874134 Chr.11: 11353115-11353115 on Build GRCh38
rs193042900 Chr.11: 11353060-11353060 on Build GRCh38
rs777947472 Chr.11: 11353161-11353161 on Build GRCh38
rs566089114 Chr.11: 11353449-11353449 on Build GRCh38
rs779427208 Chr.11: 11353021-11353021 on Build GRCh38
rs780316019 Chr.11: 11353134-11353134 on Build GRCh38
rs770203452 Chr.11: 11353156-11353156 on Build GRCh38
rs558971315 Chr.11: 11353282-11353282 on Build GRCh38
rs915887401 Chr.11: 11353206-11353206 on Build GRCh38
rs1022987342 Chr.11: 11353372-11353372 on Build GRCh38
rs559030633 Chr.11: 11353093-11353093 on Build GRCh38
rs758648808 Chr.11: 11353129-11353129 on Build GRCh38
rs911719708 Chr.11: 11353448-11353448 on Build GRCh38
rs992881230 Chr.11: 11353194-11353194 on Build GRCh38
rs143373580 Chr.11: 11353290-11353290 on Build GRCh38
rs920520104 Chr.11: 11353384-11353384 on Build GRCh38
rs535125193 Chr.11: 11353097-11353097 on Build GRCh38
rs757063204 Chr.11: 11353073-11353074 on Build GRCh38
rs139538538 Chr.11: 11353197-11353197 on Build GRCh38
rs929138873 Chr.11: 11353245-11353245 on Build GRCh38
rs558423099 Chr.11: 11353107-11353107 on Build GRCh38
rs1003416293 Chr.11: 11353413-11353413 on Build GRCh38
rs950576716 Chr.11: 11353385-11353385 on Build GRCh38
rs767036375 Chr.11: 11353100-11353100 on Build GRCh38
rs985358985 Chr.11: 11353279-11353279 on Build GRCh38
rs191384102 Chr.11: 11353445-11353445 on Build GRCh38
rs960074758 Chr.11: 11353192-11353192 on Build GRCh38
rs753635572 Chr.11: 11353067-11353067 on Build GRCh38
rs914509733 Chr.11: 11353262-11353262 on Build GRCh38
rs759040823 Chr.11: 11353049-11353049 on Build GRCh38
rs766027302 Chr.11: 11353022-11353022 on Build GRCh38
rs774941515 Chr.11: 11353106-11353106 on Build GRCh38
rs1023118281 Chr.11: 11353104-11353104 on Build GRCh38
rs920485579 Chr.11: 11353243-11353243 on Build GRCh38
rs571719102 Chr.11: 11353175-11353175 on Build GRCh38
rs182860137 Chr.11: 11353114-11353114 on Build GRCh38
rs970025958 Chr.11: 11353232-11353232 on Build GRCh38
rs983325195 Chr.11: 11353434-11353434 on Build GRCh38

View Full Product Details