Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14457791-14458307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPB1
Entrez Gene ID
1315
Gene Name
coatomer protein complex subunit beta 1
Gene Aliases
COPB
UniGene
Hs.339278
Gene Chromosome Location
Chr.11: 14457503-14499895 on Build GRCh38
Transcripts
NM_001144061.1 NM_001144062.1 NM_016451.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1352822 c.2852C>T p.T951I Chr.11: 14457834-14457834 on Build GRCh38
3700023 c.2857A>G p.I953V Chr.11: 14457829-14457829 on Build GRCh38
6132029 c.2840C>T p.S947L Chr.11: 14457846-14457846 on Build GRCh38
4791701 c.2809G>A p.A937T Chr.11: 14457877-14457877 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1045051216 Chr.11: 14457984-14457984 on Build GRCh38
rs752682987 Chr.11: 14457934-14457934 on Build GRCh38
rs775998909 Chr.11: 14457887-14457887 on Build GRCh38
rs999152020 Chr.11: 14458162-14458162 on Build GRCh38
rs988285278 Chr.11: 14458230-14458230 on Build GRCh38
rs1016679094 Chr.11: 14458118-14458118 on Build GRCh38
rs746341161 Chr.11: 14457880-14457880 on Build GRCh38
rs768685954 Chr.11: 14457892-14457892 on Build GRCh38
rs550648879 Chr.11: 14457965-14457965 on Build GRCh38
rs771309231 Chr.11: 14457839-14457839 on Build GRCh38
rs905123003 Chr.11: 14457985-14457985 on Build GRCh38
rs376286436 Chr.11: 14458022-14458022 on Build GRCh38
rs775591113 Chr.11: 14457903-14457904 on Build GRCh38
rs781719255 Chr.11: 14457879-14457880 on Build GRCh38
rs761991539 Chr.11: 14457898-14457898 on Build GRCh38
rs5789837 Chr.11: 14458075-14458076 on Build GRCh38
rs915323244 Chr.11: 14457958-14457958 on Build GRCh38
rs376124362 Chr.11: 14458207-14458207 on Build GRCh38
rs555569069 Chr.11: 14458065-14458065 on Build GRCh38
rs1051674203 Chr.11: 14458053-14458053 on Build GRCh38
rs1004044645 Chr.11: 14458103-14458103 on Build GRCh38
rs760682268 Chr.11: 14457891-14457891 on Build GRCh38
rs996477273 Chr.11: 14458158-14458158 on Build GRCh38
rs1043486844 Chr.11: 14458105-14458105 on Build GRCh38
rs768793573 Chr.11: 14457915-14457916 on Build GRCh38
rs372741936 Chr.11: 14457960-14457964 on Build GRCh38
rs377718558 Chr.11: 14458191-14458191 on Build GRCh38
rs776900167 Chr.11: 14457895-14457895 on Build GRCh38
rs542580626 Chr.11: 14457955-14457955 on Build GRCh38
rs969182748 Chr.11: 14458208-14458208 on Build GRCh38
rs973001917 Chr.11: 14457990-14457990 on Build GRCh38
rs946700510 Chr.11: 14457961-14457961 on Build GRCh38
rs879481850 Chr.11: 14458221-14458221 on Build GRCh38
rs774626201 Chr.11: 14457877-14457877 on Build GRCh38
rs746251442 Chr.11: 14457886-14457886 on Build GRCh38
rs533463683 Chr.11: 14458236-14458236 on Build GRCh38
rs191748188 Chr.11: 14458035-14458035 on Build GRCh38
rs898211508 Chr.11: 14458139-14458139 on Build GRCh38
rs886971918 Chr.11: 14458102-14458102 on Build GRCh38
rs183178329 Chr.11: 14458171-14458171 on Build GRCh38
rs934187404 Chr.11: 14458011-14458011 on Build GRCh38
rs72868007 Chr.11: 14458007-14458007 on Build GRCh38
rs532426996 Chr.11: 14457928-14457928 on Build GRCh38
rs1016974686 Chr.11: 14458222-14458222 on Build GRCh38
rs901565058 Chr.11: 14458069-14458069 on Build GRCh38
rs962680242 Chr.11: 14457990-14457991 on Build GRCh38
rs770011031 Chr.11: 14457888-14457893 on Build GRCh38
rs749325380 Chr.11: 14457904-14457905 on Build GRCh38
rs36063774 Chr.11: 14458170-14458170 on Build GRCh38
rs777415338 Chr.11: 14457828-14457828 on Build GRCh38
rs867788373 Chr.11: 14458253-14458253 on Build GRCh38
rs988665796 Chr.11: 14457899-14457899 on Build GRCh38
rs1019184782 Chr.11: 14458205-14458205 on Build GRCh38
rs909916954 Chr.11: 14457972-14457972 on Build GRCh38
rs748562590 Chr.11: 14458097-14458098 on Build GRCh38
rs759849901 Chr.11: 14458204-14458204 on Build GRCh38
rs567413247 Chr.11: 14458156-14458156 on Build GRCh38
rs1056367282 Chr.11: 14458073-14458073 on Build GRCh38
rs962691129 Chr.11: 14458229-14458229 on Build GRCh38
rs942454923 Chr.11: 14458066-14458066 on Build GRCh38
rs761792234 Chr.11: 14457918-14457923 on Build GRCh38
rs769204761 Chr.11: 14458074-14458075 on Build GRCh38
rs371492671 Chr.11: 14457909-14457909 on Build GRCh38
rs146071739 Chr.11: 14458178-14458178 on Build GRCh38
rs748955941 Chr.11: 14457829-14457829 on Build GRCh38
rs896365928 Chr.11: 14458180-14458180 on Build GRCh38

View Full Product Details