Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14460093-14460495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPB1
Entrez Gene ID
1315
Gene Name
coatomer protein complex subunit beta 1
Gene Aliases
COPB
UniGene
Hs.339278
Gene Chromosome Location
Chr.11: 14457503-14499895 on Build GRCh38
Transcripts
NM_001144061.1 NM_001144062.1 NM_016451.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466597 c.2639C>A p.P880Q Chr.11: 14460215-14460215 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931473233 Chr.11: 14460256-14460256 on Build GRCh38
rs746167085 Chr.11: 14460328-14460328 on Build GRCh38
rs892435115 Chr.11: 14460154-14460154 on Build GRCh38
rs958669061 Chr.11: 14460458-14460458 on Build GRCh38
rs138121968 Chr.11: 14460210-14460210 on Build GRCh38
rs771236146 Chr.11: 14460192-14460192 on Build GRCh38
rs753918856 Chr.11: 14460314-14460314 on Build GRCh38
rs765960575 Chr.11: 14460240-14460240 on Build GRCh38
rs1048501825 Chr.11: 14460317-14460317 on Build GRCh38
rs756724699 Chr.11: 14460308-14460308 on Build GRCh38
rs751233037 Chr.11: 14460241-14460241 on Build GRCh38
rs757799194 Chr.11: 14460321-14460323 on Build GRCh38
rs746723778 Chr.11: 14460201-14460201 on Build GRCh38
rs550840044 Chr.11: 14460452-14460452 on Build GRCh38
rs762475541 Chr.11: 14460233-14460233 on Build GRCh38
rs776420485 Chr.11: 14460226-14460226 on Build GRCh38
rs994522382 Chr.11: 14460342-14460342 on Build GRCh38
rs781770428 Chr.11: 14460336-14460337 on Build GRCh38
rs778935378 Chr.11: 14460325-14460325 on Build GRCh38
rs764696961 Chr.11: 14460229-14460229 on Build GRCh38
rs906742323 Chr.11: 14460194-14460194 on Build GRCh38
rs756244522 Chr.11: 14460165-14460165 on Build GRCh38
rs1010595917 Chr.11: 14460356-14460356 on Build GRCh38
rs778045297 Chr.11: 14460179-14460179 on Build GRCh38
rs949644665 Chr.11: 14460218-14460218 on Build GRCh38
rs1019275065 Chr.11: 14460406-14460406 on Build GRCh38
rs773307855 Chr.11: 14460279-14460279 on Build GRCh38
rs964572295 Chr.11: 14460427-14460427 on Build GRCh38
rs572981167 Chr.11: 14460341-14460341 on Build GRCh38
rs753118849 Chr.11: 14460307-14460307 on Build GRCh38
rs945658644 Chr.11: 14460160-14460160 on Build GRCh38
rs748752586 Chr.11: 14460164-14460164 on Build GRCh38
rs764887895 Chr.11: 14460309-14460309 on Build GRCh38
rs1041415100 Chr.11: 14460350-14460350 on Build GRCh38
rs771192103 Chr.11: 14460372-14460372 on Build GRCh38
rs755387008 Chr.11: 14460162-14460162 on Build GRCh38
rs1020202280 Chr.11: 14460371-14460371 on Build GRCh38
rs778452025 Chr.11: 14460309-14460309 on Build GRCh38
rs376516852 Chr.11: 14460193-14460193 on Build GRCh38
rs752081939 Chr.11: 14460308-14460309 on Build GRCh38
rs999989889 Chr.11: 14460428-14460428 on Build GRCh38
rs761859954 Chr.11: 14460227-14460227 on Build GRCh38
rs755297746 Chr.11: 14460246-14460246 on Build GRCh38
rs901109843 Chr.11: 14460353-14460353 on Build GRCh38
rs141058262 Chr.11: 14460457-14460457 on Build GRCh38
rs565101512 Chr.11: 14460133-14460133 on Build GRCh38
rs767980170 Chr.11: 14460250-14460250 on Build GRCh38
rs768314332 Chr.11: 14460336-14460336 on Build GRCh38
rs747928611 Chr.11: 14460345-14460345 on Build GRCh38
rs889829699 Chr.11: 14460334-14460334 on Build GRCh38
rs772670397 Chr.11: 14460230-14460230 on Build GRCh38
rs368830467 Chr.11: 14460322-14460322 on Build GRCh38

View Full Product Details