Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17775417-17775913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC1
Entrez Gene ID
3746
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
EPM7, KV3.1, KV4, NGK2
UniGene
Hs.552896
Gene Chromosome Location
Chr.11: 17734810-17783055 on Build GRCh38
Transcripts
NM_001112741.1 NM_004976.4 XM_011520080.2

SNP Detail

SNP ID Chromosome Location
rs905129439 Chr.11: 17775736-17775736 on Build GRCh38
rs535927107 Chr.11: 17775798-17775798 on Build GRCh38
rs943383444 Chr.11: 17775830-17775830 on Build GRCh38
rs527302293 Chr.11: 17775756-17775756 on Build GRCh38
rs1033324725 Chr.11: 17775737-17775737 on Build GRCh38
rs113156764 Chr.11: 17775517-17775517 on Build GRCh38
rs563606727 Chr.11: 17775795-17775795 on Build GRCh38
rs574477819 Chr.11: 17775724-17775724 on Build GRCh38
rs887152322 Chr.11: 17775773-17775773 on Build GRCh38
rs568573551 Chr.11: 17775605-17775605 on Build GRCh38
rs925428968 Chr.11: 17775668-17775668 on Build GRCh38
rs972649771 Chr.11: 17775769-17775769 on Build GRCh38
rs958961751 Chr.11: 17775611-17775611 on Build GRCh38
rs1021159253 Chr.11: 17775514-17775514 on Build GRCh38
rs975659970 Chr.11: 17775824-17775824 on Build GRCh38
rs926271242 Chr.11: 17775884-17775884 on Build GRCh38
rs919404892 Chr.11: 17775778-17775778 on Build GRCh38
rs901755370 Chr.11: 17775716-17775716 on Build GRCh38
rs967822896 Chr.11: 17775515-17775515 on Build GRCh38
rs917482710 Chr.11: 17775622-17775622 on Build GRCh38
rs1019598861 Chr.11: 17775793-17775793 on Build GRCh38
rs972254557 Chr.11: 17775647-17775647 on Build GRCh38
rs758613509 Chr.11: 17775699-17775699 on Build GRCh38
rs930738492 Chr.11: 17775800-17775800 on Build GRCh38
rs566425804 Chr.11: 17775877-17775877 on Build GRCh38
rs764719236 Chr.11: 17775690-17775690 on Build GRCh38
rs910624686 Chr.11: 17775814-17775814 on Build GRCh38
rs1026107385 Chr.11: 17775587-17775587 on Build GRCh38
rs930711067 Chr.11: 17775669-17775669 on Build GRCh38
rs1040342298 Chr.11: 17775705-17775705 on Build GRCh38
rs563083977 Chr.11: 17775689-17775689 on Build GRCh38
rs541906600 Chr.11: 17775725-17775725 on Build GRCh38
rs956098401 Chr.11: 17775504-17775525 on Build GRCh38
rs979513971 Chr.11: 17775590-17775590 on Build GRCh38
rs750661235 Chr.11: 17775698-17775698 on Build GRCh38
rs905078480 Chr.11: 17775503-17775503 on Build GRCh38
rs544494234 Chr.11: 17775651-17775651 on Build GRCh38
rs188741850 Chr.11: 17775602-17775602 on Build GRCh38
rs984895261 Chr.11: 17775803-17775803 on Build GRCh38
rs375756457 Chr.11: 17775864-17775864 on Build GRCh38
rs950294261 Chr.11: 17775644-17775644 on Build GRCh38
rs867291341 Chr.11: 17775472-17775472 on Build GRCh38
rs559758472 Chr.11: 17775754-17775754 on Build GRCh38
rs1009388054 Chr.11: 17775464-17775464 on Build GRCh38
rs941532826 Chr.11: 17775458-17775458 on Build GRCh38
rs991773962 Chr.11: 17775748-17775748 on Build GRCh38
rs991678838 Chr.11: 17775618-17775618 on Build GRCh38
rs12421233 Chr.11: 17775445-17775445 on Build GRCh38
rs748654836 Chr.11: 17775847-17775847 on Build GRCh38
rs781632895 Chr.11: 17775839-17775839 on Build GRCh38
rs763211449 Chr.11: 17775747-17775747 on Build GRCh38
rs986767162 Chr.11: 17775606-17775606 on Build GRCh38
rs1009855524 Chr.11: 17775723-17775723 on Build GRCh38
rs964267280 Chr.11: 17775650-17775650 on Build GRCh38
rs995587599 Chr.11: 17775524-17775524 on Build GRCh38
rs1000592361 Chr.11: 17775729-17775729 on Build GRCh38
rs913668343 Chr.11: 17775855-17775855 on Build GRCh38

View Full Product Details