Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 17776211-17776715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC1
Entrez Gene ID
3746
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
EPM7, KV3.1, KV4, NGK2
UniGene
Hs.552896
Gene Chromosome Location
Chr.11: 17734810-17783055 on Build GRCh38
Transcripts
NM_001112741.1 NM_004976.4 XM_011520080.2

SNP Detail

SNP ID Chromosome Location
rs879162160 Chr.11: 17776674-17776674 on Build GRCh38
rs765118776 Chr.11: 17776429-17776429 on Build GRCh38
rs556528888 Chr.11: 17776268-17776268 on Build GRCh38
rs888038606 Chr.11: 17776478-17776478 on Build GRCh38
rs1003686908 Chr.11: 17776448-17776448 on Build GRCh38
rs1006452052 Chr.11: 17776529-17776529 on Build GRCh38
rs189046428 Chr.11: 17776643-17776643 on Build GRCh38
rs543807518 Chr.11: 17776442-17776442 on Build GRCh38
rs529086710 Chr.11: 17776563-17776563 on Build GRCh38
rs368744164 Chr.11: 17776262-17776262 on Build GRCh38
rs145156513 Chr.11: 17776419-17776419 on Build GRCh38
rs559821051 Chr.11: 17776308-17776308 on Build GRCh38
rs527769858 Chr.11: 17776658-17776658 on Build GRCh38
rs577645864 Chr.11: 17776255-17776255 on Build GRCh38
rs1030451221 Chr.11: 17776595-17776595 on Build GRCh38
rs915297746 Chr.11: 17776657-17776657 on Build GRCh38
rs925264149 Chr.11: 17776415-17776415 on Build GRCh38
rs959093262 Chr.11: 17776252-17776252 on Build GRCh38
rs866664067 Chr.11: 17776395-17776395 on Build GRCh38
rs989961270 Chr.11: 17776648-17776649 on Build GRCh38
rs944525413 Chr.11: 17776309-17776309 on Build GRCh38
rs187170829 Chr.11: 17776399-17776399 on Build GRCh38
rs916391771 Chr.11: 17776443-17776443 on Build GRCh38
rs896887581 Chr.11: 17776463-17776463 on Build GRCh38
rs978124105 Chr.11: 17776640-17776640 on Build GRCh38
rs1048496911 Chr.11: 17776477-17776477 on Build GRCh38
rs11024395 Chr.11: 17776422-17776422 on Build GRCh38
rs976144717 Chr.11: 17776398-17776398 on Build GRCh38
rs966764567 Chr.11: 17776404-17776404 on Build GRCh38
rs138238610 Chr.11: 17776284-17776284 on Build GRCh38
rs182820085 Chr.11: 17776397-17776397 on Build GRCh38
rs547585716 Chr.11: 17776638-17776638 on Build GRCh38
rs906757825 Chr.11: 17776445-17776445 on Build GRCh38
rs921744015 Chr.11: 17776646-17776646 on Build GRCh38
rs900695661 Chr.11: 17776543-17776543 on Build GRCh38
rs148938810 Chr.11: 17776647-17776648 on Build GRCh38
rs574968276 Chr.11: 17776279-17776279 on Build GRCh38
rs11024396 Chr.11: 17776549-17776549 on Build GRCh38
rs957966091 Chr.11: 17776650-17776650 on Build GRCh38
rs371698703 Chr.11: 17776256-17776256 on Build GRCh38
rs975651589 Chr.11: 17776661-17776661 on Build GRCh38
rs965922336 Chr.11: 17776594-17776594 on Build GRCh38
rs183727461 Chr.11: 17776535-17776535 on Build GRCh38
rs1038861984 Chr.11: 17776374-17776374 on Build GRCh38
rs1053813486 Chr.11: 17776400-17776400 on Build GRCh38
rs561791248 Chr.11: 17776545-17776545 on Build GRCh38
rs147007362 Chr.11: 17776263-17776263 on Build GRCh38
rs1046172349 Chr.11: 17776458-17776458 on Build GRCh38
rs1027362714 Chr.11: 17776514-17776514 on Build GRCh38
rs916482974 Chr.11: 17776660-17776660 on Build GRCh38
rs911829248 Chr.11: 17776546-17776546 on Build GRCh38
rs929696172 Chr.11: 17776475-17776475 on Build GRCh38
rs750282993 Chr.11: 17776449-17776449 on Build GRCh38
rs1017753775 Chr.11: 17776325-17776325 on Build GRCh38
rs563725846 Chr.11: 17776412-17776412 on Build GRCh38
rs896839055 Chr.11: 17776251-17776251 on Build GRCh38

View Full Product Details