Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27040881-27041393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBOX1
Entrez Gene ID
8424
Gene Name
gamma-butyrobetaine hydroxylase 1
Gene Aliases
BBH, BBOX, G-BBH, gamma-BBH
UniGene
Hs.591996
Gene Chromosome Location
Chr.11: 27040705-27127807 on Build GRCh38
Transcripts
NM_003986.2 XM_005253159.2 XM_005253160.2 XM_005253161.2 XM_011520402.1

SNP Detail

SNP ID Chromosome Location
rs1010025512 Chr.11: 27041240-27041240 on Build GRCh38
rs60996491 Chr.11: 27041202-27041202 on Build GRCh38
rs7358459 Chr.11: 27041303-27041303 on Build GRCh38
rs750646771 Chr.11: 27041321-27041321 on Build GRCh38
rs910029201 Chr.11: 27041328-27041328 on Build GRCh38
rs965920053 Chr.11: 27041049-27041049 on Build GRCh38
rs138682156 Chr.11: 27041325-27041325 on Build GRCh38
rs918620227 Chr.11: 27041315-27041315 on Build GRCh38
rs1010323789 Chr.11: 27040987-27040987 on Build GRCh38
rs531984839 Chr.11: 27041201-27041202 on Build GRCh38
rs869267578 Chr.11: 27041185-27041186 on Build GRCh38
rs905497848 Chr.11: 27041160-27041160 on Build GRCh38
rs11300902 Chr.11: 27041185-27041185 on Build GRCh38
rs112838088 Chr.11: 27041353-27041353 on Build GRCh38
rs1045669070 Chr.11: 27041346-27041346 on Build GRCh38
rs16916476 Chr.11: 27040941-27040941 on Build GRCh38
rs201541456 Chr.11: 27041203-27041203 on Build GRCh38
rs564970366 Chr.11: 27041000-27041000 on Build GRCh38
rs563800651 Chr.11: 27040905-27040905 on Build GRCh38
rs950273682 Chr.11: 27041196-27041203 on Build GRCh38
rs537344988 Chr.11: 27041219-27041219 on Build GRCh38
rs1010625524 Chr.11: 27041329-27041330 on Build GRCh38
rs200882350 Chr.11: 27041185-27041192 on Build GRCh38
rs372447625 Chr.11: 27041355-27041355 on Build GRCh38
rs948265294 Chr.11: 27041130-27041130 on Build GRCh38
rs201650142 Chr.11: 27041203-27041203 on Build GRCh38
rs965697227 Chr.11: 27041290-27041290 on Build GRCh38
rs906154086 Chr.11: 27041277-27041277 on Build GRCh38
rs12417081 Chr.11: 27041159-27041159 on Build GRCh38
rs568556114 Chr.11: 27041005-27041005 on Build GRCh38
rs75938218 Chr.11: 27041199-27041202 on Build GRCh38
rs10528120 Chr.11: 27041202-27041202 on Build GRCh38
rs564463227 Chr.11: 27040955-27040955 on Build GRCh38
rs199685296 Chr.11: 27041192-27041192 on Build GRCh38
rs891549663 Chr.11: 27041196-27041196 on Build GRCh38
rs1035380892 Chr.11: 27041175-27041175 on Build GRCh38
rs1054674787 Chr.11: 27041318-27041318 on Build GRCh38
rs141688554 Chr.11: 27041276-27041276 on Build GRCh38
rs779247502 Chr.11: 27041271-27041271 on Build GRCh38
rs1028891234 Chr.11: 27041102-27041102 on Build GRCh38
rs1020765075 Chr.11: 27041029-27041029 on Build GRCh38
rs948261102 Chr.11: 27041332-27041332 on Build GRCh38
rs548371244 Chr.11: 27041120-27041120 on Build GRCh38
rs531238252 Chr.11: 27040922-27040922 on Build GRCh38
rs751561059 Chr.11: 27041191-27041192 on Build GRCh38
rs182762363 Chr.11: 27040984-27040984 on Build GRCh38
rs147128499 Chr.11: 27041310-27041310 on Build GRCh38
rs1002530448 Chr.11: 27041173-27041173 on Build GRCh38
rs73448723 Chr.11: 27041079-27041079 on Build GRCh38
rs953297052 Chr.11: 27041113-27041113 on Build GRCh38
rs558472122 Chr.11: 27041239-27041239 on Build GRCh38
rs556712353 Chr.11: 27041136-27041136 on Build GRCh38
rs777990734 Chr.11: 27040974-27040974 on Build GRCh38
rs1021435792 Chr.11: 27041244-27041244 on Build GRCh38
rs1033009510 Chr.11: 27040942-27040942 on Build GRCh38
rs574493906 Chr.11: 27041344-27041344 on Build GRCh38
rs757991269 Chr.11: 27041192-27041192 on Build GRCh38
rs1048535461 Chr.11: 27041116-27041116 on Build GRCh38
rs888621017 Chr.11: 27041123-27041123 on Build GRCh38
rs532217123 Chr.11: 27041003-27041003 on Build GRCh38
rs906100274 Chr.11: 27040924-27040924 on Build GRCh38

View Full Product Details