Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64988271-64988768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BATF2
Entrez Gene ID
116071
Gene Name
basic leucine zipper ATF-like transcription factor 2
Gene Aliases
SARI
UniGene
Hs.124840
Gene Chromosome Location
Chr.11: 64987945-64997045 on Build GRCh38
Transcripts
NM_001300807.1 NM_001300808.1 NM_138456.3

SNP Detail

SNP ID Chromosome Location
rs578122840 Chr.11: 64988435-64988435 on Build GRCh38
rs894939595 Chr.11: 64988473-64988473 on Build GRCh38
rs916366894 Chr.11: 64988726-64988726 on Build GRCh38
rs539955939 Chr.11: 64988498-64988498 on Build GRCh38
rs939068882 Chr.11: 64988414-64988414 on Build GRCh38
rs891143064 Chr.11: 64988566-64988566 on Build GRCh38
rs994166108 Chr.11: 64988510-64988510 on Build GRCh38
rs957533307 Chr.11: 64988593-64988593 on Build GRCh38
rs781366583 Chr.11: 64988502-64988502 on Build GRCh38
rs1018317905 Chr.11: 64988585-64988585 on Build GRCh38
rs897090300 Chr.11: 64988499-64988499 on Build GRCh38
rs1041633189 Chr.11: 64988337-64988337 on Build GRCh38
rs747480767 Chr.11: 64988388-64988388 on Build GRCh38
rs187222961 Chr.11: 64988606-64988606 on Build GRCh38
rs551691301 Chr.11: 64988708-64988708 on Build GRCh38
rs894204246 Chr.11: 64988471-64988471 on Build GRCh38
rs781063487 Chr.11: 64988447-64988447 on Build GRCh38
rs1019855780 Chr.11: 64988547-64988547 on Build GRCh38
rs1028344057 Chr.11: 64988616-64988616 on Build GRCh38
rs927670020 Chr.11: 64988407-64988407 on Build GRCh38
rs977667189 Chr.11: 64988728-64988728 on Build GRCh38
rs968010859 Chr.11: 64988647-64988647 on Build GRCh38
rs562445544 Chr.11: 64988648-64988648 on Build GRCh38
rs914013846 Chr.11: 64988698-64988698 on Build GRCh38
rs148730001 Chr.11: 64988578-64988578 on Build GRCh38
rs531669479 Chr.11: 64988680-64988680 on Build GRCh38
rs990322936 Chr.11: 64988382-64988382 on Build GRCh38
rs1020481202 Chr.11: 64988623-64988623 on Build GRCh38
rs558110256 Chr.11: 64988374-64988374 on Build GRCh38
rs928591107 Chr.11: 64988392-64988392 on Build GRCh38
rs981730138 Chr.11: 64988677-64988677 on Build GRCh38
rs945054168 Chr.11: 64988389-64988389 on Build GRCh38
rs941211722 Chr.11: 64988474-64988474 on Build GRCh38
rs745953402 Chr.11: 64988546-64988546 on Build GRCh38
rs542616741 Chr.11: 64988609-64988609 on Build GRCh38
rs957544561 Chr.11: 64988334-64988334 on Build GRCh38
rs1054888561 Chr.11: 64988466-64988466 on Build GRCh38
rs887262304 Chr.11: 64988501-64988501 on Build GRCh38
rs978185164 Chr.11: 64988394-64988394 on Build GRCh38
rs371784875 Chr.11: 64988618-64988618 on Build GRCh38
rs771591488 Chr.11: 64988440-64988440 on Build GRCh38
rs778242680 Chr.11: 64988387-64988387 on Build GRCh38
rs142322447 Chr.11: 64988710-64988710 on Build GRCh38
rs1018370517 Chr.11: 64988321-64988321 on Build GRCh38
rs960405265 Chr.11: 64988689-64988689 on Build GRCh38

View Full Product Details