Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65018273-65018772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2
Entrez Gene ID
402
Gene Name
ADP ribosylation factor like GTPase 2
Gene Aliases
ARFL2
UniGene
Hs.502836
Gene Chromosome Location
Chr.11: 65014113-65022185 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Gene Symbol
ARL2-SNX15
Entrez Gene ID
100528018
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65014113-65040572 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Gene Symbol
MIR6879
Entrez Gene ID
102466756
Gene Name
microRNA 6879
Gene Aliases
hsa-mir-6879
UniGene
Gene Chromosome Location
Chr.11: 65018505-65018570 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397433 c.209A>G p.Q70R Chr.11: 65018603-65018603 on Build GRCh38
6226809 c.66-8C>T p.? Chr.11: 65018356-65018356 on Build GRCh38
3810004 c.221G>T p.R74L Chr.11: 65018615-65018615 on Build GRCh38
6194326 c.119A>G p.D40G Chr.11: 65018417-65018417 on Build GRCh38
3726594 c.127A>G p.T43A Chr.11: 65018425-65018425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907294168 Chr.11: 65018421-65018421 on Build GRCh38
rs955172753 Chr.11: 65018301-65018301 on Build GRCh38
rs761946079 Chr.11: 65018470-65018470 on Build GRCh38
rs770092004 Chr.11: 65018452-65018452 on Build GRCh38
rs763847712 Chr.11: 65018658-65018658 on Build GRCh38
rs190800247 Chr.11: 65018350-65018350 on Build GRCh38
rs761010941 Chr.11: 65018485-65018485 on Build GRCh38
rs780868896 Chr.11: 65018425-65018425 on Build GRCh38
rs750050216 Chr.11: 65018539-65018539 on Build GRCh38
rs751493254 Chr.11: 65018514-65018514 on Build GRCh38
rs1007133912 Chr.11: 65018673-65018673 on Build GRCh38
rs773435992 Chr.11: 65018483-65018483 on Build GRCh38
rs911123085 Chr.11: 65018331-65018331 on Build GRCh38
rs773171394 Chr.11: 65018627-65018627 on Build GRCh38
rs144161068 Chr.11: 65018460-65018460 on Build GRCh38
rs758414914 Chr.11: 65018419-65018419 on Build GRCh38
rs780425731 Chr.11: 65018453-65018453 on Build GRCh38
rs78241716 Chr.11: 65018337-65018337 on Build GRCh38
rs201379160 Chr.11: 65018555-65018555 on Build GRCh38
rs750676722 Chr.11: 65018356-65018356 on Build GRCh38
rs748651201 Chr.11: 65018615-65018615 on Build GRCh38
rs772153074 Chr.11: 65018621-65018621 on Build GRCh38
rs764092292 Chr.11: 65018513-65018513 on Build GRCh38
rs906537666 Chr.11: 65018561-65018561 on Build GRCh38
rs771266504 Chr.11: 65018646-65018646 on Build GRCh38
rs545247831 Chr.11: 65018321-65018321 on Build GRCh38
rs775951828 Chr.11: 65018322-65018322 on Build GRCh38
rs370884100 Chr.11: 65018324-65018324 on Build GRCh38
rs752554416 Chr.11: 65018487-65018487 on Build GRCh38
rs769161323 Chr.11: 65018462-65018462 on Build GRCh38
rs201625837 Chr.11: 65018655-65018655 on Build GRCh38
rs202163474 Chr.11: 65018523-65018523 on Build GRCh38
rs554217695 Chr.11: 65018718-65018718 on Build GRCh38
rs766982672 Chr.11: 65018674-65018674 on Build GRCh38
rs571758436 Chr.11: 65018687-65018687 on Build GRCh38
rs749724483 Chr.11: 65018457-65018457 on Build GRCh38
rs1056004482 Chr.11: 65018439-65018439 on Build GRCh38
rs777361704 Chr.11: 65018420-65018420 on Build GRCh38
rs376934064 Chr.11: 65018667-65018667 on Build GRCh38
rs373886698 Chr.11: 65018680-65018680 on Build GRCh38
rs937482758 Chr.11: 65018435-65018435 on Build GRCh38
rs955159677 Chr.11: 65018656-65018656 on Build GRCh38
rs543741844 Chr.11: 65018601-65018601 on Build GRCh38
rs778704995 Chr.11: 65018317-65018317 on Build GRCh38
rs746518542 Chr.11: 65018422-65018422 on Build GRCh38
rs570221663 Chr.11: 65018558-65018558 on Build GRCh38
rs936823261 Chr.11: 65018397-65018397 on Build GRCh38
rs778402408 Chr.11: 65018704-65018704 on Build GRCh38
rs201673198 Chr.11: 65018408-65018408 on Build GRCh38
rs768454577 Chr.11: 65018731-65018731 on Build GRCh38
rs773198920 Chr.11: 65018747-65018747 on Build GRCh38
rs755708380 Chr.11: 65018675-65018675 on Build GRCh38
rs774017732 Chr.11: 65018739-65018739 on Build GRCh38
rs753529299 Chr.11: 65018686-65018686 on Build GRCh38
rs749944867 Chr.11: 65018679-65018679 on Build GRCh38
rs774777357 Chr.11: 65018469-65018469 on Build GRCh38
rs745559338 Chr.11: 65018451-65018451 on Build GRCh38
rs779442670 Chr.11: 65018607-65018607 on Build GRCh38
rs761749850 Chr.11: 65018742-65018742 on Build GRCh38
rs748623680 Chr.11: 65018690-65018690 on Build GRCh38
rs779177133 Chr.11: 65018688-65018688 on Build GRCh38
rs1045806113 Chr.11: 65018415-65018415 on Build GRCh38
rs755763132 Chr.11: 65018553-65018553 on Build GRCh38
rs749014112 Chr.11: 65018697-65018697 on Build GRCh38
rs747084254 Chr.11: 65018705-65018705 on Build GRCh38
rs547038303 Chr.11: 65018357-65018357 on Build GRCh38
rs150997435 Chr.11: 65018681-65018681 on Build GRCh38
rs777063183 Chr.11: 65018647-65018647 on Build GRCh38
rs746118890 Chr.11: 65018727-65018727 on Build GRCh38
rs112889675 Chr.11: 65018363-65018363 on Build GRCh38
rs74814065 Chr.11: 65018557-65018557 on Build GRCh38
rs764114808 Chr.11: 65018329-65018329 on Build GRCh38
rs376542433 Chr.11: 65018749-65018749 on Build GRCh38
rs199961468 Chr.11: 65018471-65018471 on Build GRCh38
rs762903453 Chr.11: 65018490-65018490 on Build GRCh38
rs765057682 Chr.11: 65018486-65018486 on Build GRCh38
rs747772006 Chr.11: 65018341-65018343 on Build GRCh38
rs765147433 Chr.11: 65018406-65018406 on Build GRCh38
rs760922486 Chr.11: 65018641-65018641 on Build GRCh38
rs780586325 Chr.11: 65018534-65018534 on Build GRCh38
rs533989493 Chr.11: 65018710-65018710 on Build GRCh38
rs369088336 Chr.11: 65018565-65018565 on Build GRCh38
rs139269006 Chr.11: 65018424-65018424 on Build GRCh38

View Full Product Details