Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65018435-65018929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2
Entrez Gene ID
402
Gene Name
ADP ribosylation factor like GTPase 2
Gene Aliases
ARFL2
UniGene
Hs.502836
Gene Chromosome Location
Chr.11: 65014113-65022185 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Gene Symbol
ARL2-SNX15
Entrez Gene ID
100528018
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65014113-65040572 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Gene Symbol
MIR6879
Entrez Gene ID
102466756
Gene Name
microRNA 6879
Gene Aliases
hsa-mir-6879
UniGene
Gene Chromosome Location
Chr.11: 65018505-65018570 on Build GRCh38
Transcripts
NM_001199745.1 NM_001667.3 NR_037650.1 NR_106939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397433 c.209A>G p.Q70R Chr.11: 65018603-65018603 on Build GRCh38
3810004 c.221G>T p.R74L Chr.11: 65018615-65018615 on Build GRCh38
1283501 c.248G>A p.S83N Chr.11: 65018642-65018642 on Build GRCh38
6552251 c.172C>T p.R58* Chr.11: 65018470-65018470 on Build GRCh38
1135231 c.293G>A p.R98H Chr.11: 65018687-65018687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765580189 Chr.11: 65018779-65018779 on Build GRCh38
rs761946079 Chr.11: 65018470-65018470 on Build GRCh38
rs763847712 Chr.11: 65018658-65018658 on Build GRCh38
rs761010941 Chr.11: 65018485-65018485 on Build GRCh38
rs757367469 Chr.11: 65018806-65018806 on Build GRCh38
rs750050216 Chr.11: 65018539-65018539 on Build GRCh38
rs751493254 Chr.11: 65018514-65018514 on Build GRCh38
rs1007133912 Chr.11: 65018673-65018673 on Build GRCh38
rs763407595 Chr.11: 65018855-65018877 on Build GRCh38
rs371041179 Chr.11: 65018761-65018761 on Build GRCh38
rs928698954 Chr.11: 65018791-65018791 on Build GRCh38
rs771940608 Chr.11: 65018859-65018859 on Build GRCh38
rs773435992 Chr.11: 65018483-65018483 on Build GRCh38
rs760627312 Chr.11: 65018871-65018871 on Build GRCh38
rs144161068 Chr.11: 65018460-65018460 on Build GRCh38
rs773171394 Chr.11: 65018627-65018627 on Build GRCh38
rs753439287 Chr.11: 65018770-65018770 on Build GRCh38
rs775836232 Chr.11: 65018850-65018850 on Build GRCh38
rs201379160 Chr.11: 65018555-65018555 on Build GRCh38
rs772153074 Chr.11: 65018621-65018621 on Build GRCh38
rs748651201 Chr.11: 65018615-65018615 on Build GRCh38
rs764092292 Chr.11: 65018513-65018513 on Build GRCh38
rs1028484501 Chr.11: 65018867-65018867 on Build GRCh38
rs906537666 Chr.11: 65018561-65018561 on Build GRCh38
rs771266504 Chr.11: 65018646-65018646 on Build GRCh38
rs752554416 Chr.11: 65018487-65018487 on Build GRCh38
rs769161323 Chr.11: 65018462-65018462 on Build GRCh38
rs201625837 Chr.11: 65018655-65018655 on Build GRCh38
rs202163474 Chr.11: 65018523-65018523 on Build GRCh38
rs554217695 Chr.11: 65018718-65018718 on Build GRCh38
rs766982672 Chr.11: 65018674-65018674 on Build GRCh38
rs770331127 Chr.11: 65018780-65018781 on Build GRCh38
rs571758436 Chr.11: 65018687-65018687 on Build GRCh38
rs749724483 Chr.11: 65018457-65018457 on Build GRCh38
rs545626345 Chr.11: 65018907-65018907 on Build GRCh38
rs556922598 Chr.11: 65018818-65018818 on Build GRCh38
rs769917578 Chr.11: 65018843-65018843 on Build GRCh38
rs376934064 Chr.11: 65018667-65018667 on Build GRCh38
rs373886698 Chr.11: 65018680-65018680 on Build GRCh38
rs955159677 Chr.11: 65018656-65018656 on Build GRCh38
rs543741844 Chr.11: 65018601-65018601 on Build GRCh38
rs746488072 Chr.11: 65018779-65018779 on Build GRCh38
rs570221663 Chr.11: 65018558-65018558 on Build GRCh38
rs772668773 Chr.11: 65018777-65018778 on Build GRCh38
rs778402408 Chr.11: 65018704-65018704 on Build GRCh38
rs574046543 Chr.11: 65018804-65018804 on Build GRCh38
rs768454577 Chr.11: 65018731-65018731 on Build GRCh38
rs765810555 Chr.11: 65018875-65018875 on Build GRCh38
rs776132002 Chr.11: 65018890-65018890 on Build GRCh38
rs773198920 Chr.11: 65018747-65018747 on Build GRCh38
rs755708380 Chr.11: 65018675-65018675 on Build GRCh38
rs1019267962 Chr.11: 65018778-65018778 on Build GRCh38
rs774017732 Chr.11: 65018739-65018739 on Build GRCh38
rs753529299 Chr.11: 65018686-65018686 on Build GRCh38
rs749944867 Chr.11: 65018679-65018679 on Build GRCh38
rs774777357 Chr.11: 65018469-65018469 on Build GRCh38
rs748623680 Chr.11: 65018690-65018690 on Build GRCh38
rs779442670 Chr.11: 65018607-65018607 on Build GRCh38
rs779177133 Chr.11: 65018688-65018688 on Build GRCh38
rs761749850 Chr.11: 65018742-65018742 on Build GRCh38
rs749014112 Chr.11: 65018697-65018697 on Build GRCh38
rs755763132 Chr.11: 65018553-65018553 on Build GRCh38
rs747084254 Chr.11: 65018705-65018705 on Build GRCh38
rs370568230 Chr.11: 65018855-65018855 on Build GRCh38
rs771746476 Chr.11: 65018776-65018777 on Build GRCh38
rs752992346 Chr.11: 65018781-65018781 on Build GRCh38
rs150997435 Chr.11: 65018681-65018681 on Build GRCh38
rs777063183 Chr.11: 65018647-65018647 on Build GRCh38
rs74814065 Chr.11: 65018557-65018557 on Build GRCh38
rs746118890 Chr.11: 65018727-65018727 on Build GRCh38
rs754591820 Chr.11: 65018773-65018773 on Build GRCh38
rs773108986 Chr.11: 65018866-65018866 on Build GRCh38
rs376542433 Chr.11: 65018749-65018749 on Build GRCh38
rs199961468 Chr.11: 65018471-65018471 on Build GRCh38
rs762903453 Chr.11: 65018490-65018490 on Build GRCh38
rs765057682 Chr.11: 65018486-65018486 on Build GRCh38
rs778213020 Chr.11: 65018818-65018818 on Build GRCh38
rs144619946 Chr.11: 65018807-65018807 on Build GRCh38
rs758866064 Chr.11: 65018783-65018783 on Build GRCh38
rs760922486 Chr.11: 65018641-65018641 on Build GRCh38
rs780586325 Chr.11: 65018534-65018534 on Build GRCh38
rs533989493 Chr.11: 65018710-65018710 on Build GRCh38
rs369088336 Chr.11: 65018565-65018565 on Build GRCh38

View Full Product Details