Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117292104-117292545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACE1
Entrez Gene ID
23621
Gene Name
beta-secretase 1
Gene Aliases
ASP2, BACE, HSPC104
UniGene
Hs.504003
Gene Chromosome Location
Chr.11: 117285686-117316256 on Build GRCh38
Transcripts
NM_001207048.1 NM_001207049.1 NM_012104.4 NM_138971.3 NM_138972.3 NM_138973.3 NR_037803.2

Gene Symbol
BACE1-AS
Entrez Gene ID
100379571
Gene Name
BACE1 antisense RNA
Gene Aliases
BACE1-AS1, BACE1AS, NCRNA00177
UniGene
Hs.736305
Gene Chromosome Location
Chr.11: 117291346-117292170 on Build GRCh38
Transcripts
NM_001207048.1 NM_001207049.1 NM_012104.4 NM_138971.3 NM_138972.3 NM_138973.3 NR_037803.2

SNP Detail

SNP ID Chromosome Location
rs903254463 Chr.11: 117292188-117292188 on Build GRCh38
rs75236964 Chr.11: 117292158-117292158 on Build GRCh38
rs938004692 Chr.11: 117292290-117292290 on Build GRCh38
rs879007792 Chr.11: 117292208-117292234 on Build GRCh38
rs569230884 Chr.11: 117292410-117292410 on Build GRCh38
rs904589697 Chr.11: 117292444-117292444 on Build GRCh38
rs1032235216 Chr.11: 117292447-117292447 on Build GRCh38
rs888963367 Chr.11: 117292402-117292402 on Build GRCh38
rs1009386182 Chr.11: 117292470-117292470 on Build GRCh38
rs971244302 Chr.11: 117292260-117292260 on Build GRCh38
rs1021025654 Chr.11: 117292259-117292259 on Build GRCh38
rs926644500 Chr.11: 117292268-117292268 on Build GRCh38
rs190663181 Chr.11: 117292265-117292265 on Build GRCh38
rs982161413 Chr.11: 117292486-117292486 on Build GRCh38
rs1023090615 Chr.11: 117292175-117292175 on Build GRCh38
rs76039038 Chr.11: 117292206-117292206 on Build GRCh38
rs767365119 Chr.11: 117292283-117292283 on Build GRCh38
rs557957196 Chr.11: 117292443-117292443 on Build GRCh38
rs918896433 Chr.11: 117292334-117292334 on Build GRCh38
rs573393773 Chr.11: 117292390-117292390 on Build GRCh38
rs971710468 Chr.11: 117292327-117292327 on Build GRCh38
rs1021246164 Chr.11: 117292473-117292473 on Build GRCh38
rs998964798 Chr.11: 117292190-117292190 on Build GRCh38
rs547978217 Chr.11: 117292340-117292347 on Build GRCh38
rs1032227885 Chr.11: 117292163-117292163 on Build GRCh38
rs980579033 Chr.11: 117292191-117292191 on Build GRCh38
rs554720503 Chr.11: 117292363-117292363 on Build GRCh38
rs538077560 Chr.11: 117292431-117292431 on Build GRCh38
rs540510924 Chr.11: 117292474-117292474 on Build GRCh38
rs375461112 Chr.11: 117292181-117292181 on Build GRCh38
rs1037633690 Chr.11: 117292383-117292383 on Build GRCh38
rs933108210 Chr.11: 117292421-117292421 on Build GRCh38
rs866636177 Chr.11: 117292186-117292186 on Build GRCh38
rs893727499 Chr.11: 117292465-117292465 on Build GRCh38
rs34135426 Chr.11: 117292466-117292467 on Build GRCh38
rs908951987 Chr.11: 117292233-117292233 on Build GRCh38
rs761952154 Chr.11: 117292252-117292252 on Build GRCh38
rs940176873 Chr.11: 117292378-117292378 on Build GRCh38
rs1051368446 Chr.11: 117292426-117292426 on Build GRCh38
rs957855513 Chr.11: 117292228-117292228 on Build GRCh38
rs138557212 Chr.11: 117292317-117292317 on Build GRCh38

View Full Product Details