Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99690901-99691460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99659506-99711245 on Build GRCh38
Transcripts
NM_020354.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4506229 c.713C>T p.S238L Chr.10: 99691388-99691388 on Build GRCh38
6518185 c.784G>A p.A262T Chr.10: 99691459-99691459 on Build GRCh38
4596711 c.730C>T p.Q244* Chr.10: 99691405-99691405 on Build GRCh38
5049819 c.748C>T p.R250W Chr.10: 99691423-99691423 on Build GRCh38
426907 c.772G>T p.D258Y Chr.10: 99691447-99691447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050813097 Chr.10: 99690978-99690978 on Build GRCh38
rs910924404 Chr.10: 99691164-99691164 on Build GRCh38
rs764386935 Chr.10: 99691370-99691370 on Build GRCh38
rs755967451 Chr.10: 99691049-99691049 on Build GRCh38
rs1001704810 Chr.10: 99691116-99691116 on Build GRCh38
rs779124352 Chr.10: 99691341-99691341 on Build GRCh38
rs755830723 Chr.10: 99691433-99691433 on Build GRCh38
rs985391118 Chr.10: 99691392-99691392 on Build GRCh38
rs772356200 Chr.10: 99691354-99691354 on Build GRCh38
rs6584309 Chr.10: 99691072-99691072 on Build GRCh38
rs6584308 Chr.10: 99690937-99690937 on Build GRCh38
rs979319898 Chr.10: 99691175-99691175 on Build GRCh38
rs903649231 Chr.10: 99691025-99691025 on Build GRCh38
rs771181785 Chr.10: 99691378-99691378 on Build GRCh38
rs538209413 Chr.10: 99691164-99691164 on Build GRCh38
rs767106170 Chr.10: 99691424-99691424 on Build GRCh38
rs747354667 Chr.10: 99691375-99691375 on Build GRCh38
rs935169530 Chr.10: 99691031-99691031 on Build GRCh38
rs901076162 Chr.10: 99691149-99691149 on Build GRCh38
rs575922825 Chr.10: 99690995-99690996 on Build GRCh38
rs891870713 Chr.10: 99691327-99691327 on Build GRCh38
rs771397300 Chr.10: 99691107-99691107 on Build GRCh38
rs1039365760 Chr.10: 99691141-99691141 on Build GRCh38
rs11190244 Chr.10: 99691163-99691163 on Build GRCh38
rs555177484 Chr.10: 99691393-99691393 on Build GRCh38
rs72832512 Chr.10: 99691437-99691437 on Build GRCh38
rs530777063 Chr.10: 99691106-99691106 on Build GRCh38
rs200496455 Chr.10: 99691162-99691162 on Build GRCh38
rs750297462 Chr.10: 99690991-99690994 on Build GRCh38
rs759943729 Chr.10: 99691398-99691398 on Build GRCh38
rs532366423 Chr.10: 99691038-99691038 on Build GRCh38
rs764259944 Chr.10: 99691266-99691266 on Build GRCh38
rs551673847 Chr.10: 99691071-99691071 on Build GRCh38
rs1052289755 Chr.10: 99691253-99691253 on Build GRCh38
rs918356694 Chr.10: 99690941-99690941 on Build GRCh38
rs557643955 Chr.10: 99691198-99691198 on Build GRCh38
rs368813452 Chr.10: 99691221-99691221 on Build GRCh38
rs942365761 Chr.10: 99691127-99691127 on Build GRCh38
rs763022800 Chr.10: 99691220-99691220 on Build GRCh38
rs919432364 Chr.10: 99691092-99691092 on Build GRCh38
rs891906119 Chr.10: 99691050-99691050 on Build GRCh38
rs991933107 Chr.10: 99690985-99690985 on Build GRCh38
rs12249456 Chr.10: 99691252-99691252 on Build GRCh38
rs761342650 Chr.10: 99691423-99691423 on Build GRCh38
rs534399682 Chr.10: 99691161-99691161 on Build GRCh38
rs1033388063 Chr.10: 99690947-99690947 on Build GRCh38
rs565262367 Chr.10: 99691036-99691036 on Build GRCh38
rs774034291 Chr.10: 99691372-99691375 on Build GRCh38
rs199637784 Chr.10: 99691156-99691156 on Build GRCh38
rs750022624 Chr.10: 99691428-99691428 on Build GRCh38
rs983882011 Chr.10: 99691120-99691120 on Build GRCh38
rs765745100 Chr.10: 99691411-99691411 on Build GRCh38
rs933625403 Chr.10: 99690955-99690955 on Build GRCh38
rs1043571460 Chr.10: 99691017-99691017 on Build GRCh38
rs34046167 Chr.10: 99691175-99691175 on Build GRCh38
rs548766249 Chr.10: 99691123-99691123 on Build GRCh38
rs557092189 Chr.10: 99691151-99691151 on Build GRCh38
rs958732206 Chr.10: 99690952-99690952 on Build GRCh38
rs748253142 Chr.10: 99691343-99691343 on Build GRCh38
rs139033055 Chr.10: 99691032-99691032 on Build GRCh38
rs971605659 Chr.10: 99691292-99691292 on Build GRCh38
rs773602293 Chr.10: 99691370-99691370 on Build GRCh38
rs192655451 Chr.10: 99691419-99691419 on Build GRCh38
rs972310592 Chr.10: 99691317-99691317 on Build GRCh38

View Full Product Details