Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123787712-123788210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH10
Entrez Gene ID
196385
Gene Name
dynein axonemal heavy chain 10
Gene Aliases
-
UniGene
Hs.622654
Gene Chromosome Location
Chr.12: 123761995-123935720 on Build GRCh38
Transcripts
NM_207437.3 XM_005253555.3 XM_011538014.2 XM_011538015.2 XM_011538016.2 XM_011538017.2 XM_011538020.2 XM_017018960.1 XM_017018961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4427791 c.824G>A p.R275Q Chr.12: 123787935-123787935 on Build GRCh38
6051563 c.746G>A p.R249K Chr.12: 123787857-123787857 on Build GRCh38
3457576 c.760G>A p.A254T Chr.12: 123787871-123787871 on Build GRCh38
6359023 c.1391C>T p.T464M Chr.12: 123787956-123787956 on Build GRCh38
4833344 c.822C>T p.D274D Chr.12: 123787933-123787933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771535684 Chr.12: 123787790-123787791 on Build GRCh38
rs764942262 Chr.12: 123788024-123788024 on Build GRCh38
rs558231333 Chr.12: 123787736-123787736 on Build GRCh38
rs923031229 Chr.12: 123787773-123787773 on Build GRCh38
rs760229533 Chr.12: 123787815-123787815 on Build GRCh38
rs752170573 Chr.12: 123787944-123787944 on Build GRCh38
rs994141300 Chr.12: 123787735-123787735 on Build GRCh38
rs781151744 Chr.12: 123788008-123788008 on Build GRCh38
rs751954619 Chr.12: 123787817-123787817 on Build GRCh38
rs994172250 Chr.12: 123787872-123787872 on Build GRCh38
rs771011276 Chr.12: 123787799-123787799 on Build GRCh38
rs1031814218 Chr.12: 123788066-123788066 on Build GRCh38
rs778668046 Chr.12: 123787783-123787783 on Build GRCh38
rs932400614 Chr.12: 123787801-123787801 on Build GRCh38
rs200629703 Chr.12: 123787895-123787895 on Build GRCh38
rs186760005 Chr.12: 123787991-123787991 on Build GRCh38
rs867025519 Chr.12: 123788079-123788079 on Build GRCh38
rs947065260 Chr.12: 123788129-123788129 on Build GRCh38
rs965644697 Chr.12: 123788073-123788073 on Build GRCh38
rs914263127 Chr.12: 123787811-123787811 on Build GRCh38
rs760248622 Chr.12: 123787884-123787884 on Build GRCh38
rs543786627 Chr.12: 123787753-123787753 on Build GRCh38
rs765224002 Chr.12: 123787812-123787812 on Build GRCh38
rs986154223 Chr.12: 123787745-123787745 on Build GRCh38
rs966016303 Chr.12: 123787772-123787772 on Build GRCh38
rs376640267 Chr.12: 123788012-123788012 on Build GRCh38
rs758016401 Chr.12: 123787867-123787867 on Build GRCh38
rs766869862 Chr.12: 123787941-123787941 on Build GRCh38
rs145065895 Chr.12: 123787905-123787905 on Build GRCh38
rs776774116 Chr.12: 123787886-123787886 on Build GRCh38
rs775148949 Chr.12: 123787871-123787872 on Build GRCh38
rs767865064 Chr.12: 123787927-123787927 on Build GRCh38
rs144145237 Chr.12: 123787873-123787873 on Build GRCh38
rs781637400 Chr.12: 123787759-123787759 on Build GRCh38
rs371001710 Chr.12: 123787760-123787760 on Build GRCh38
rs777604662 Chr.12: 123787957-123787957 on Build GRCh38
rs764312185 Chr.12: 123787816-123787816 on Build GRCh38
rs778299462 Chr.12: 123787865-123787865 on Build GRCh38
rs751868475 Chr.12: 123787758-123787758 on Build GRCh38
rs547637372 Chr.12: 123788036-123788036 on Build GRCh38
rs772885473 Chr.12: 123787793-123787793 on Build GRCh38
rs939803008 Chr.12: 123787849-123787849 on Build GRCh38
rs776684495 Chr.12: 123787800-123787800 on Build GRCh38
rs747741367 Chr.12: 123787786-123787786 on Build GRCh38
rs201134504 Chr.12: 123787803-123787803 on Build GRCh38
rs897252466 Chr.12: 123787871-123787871 on Build GRCh38
rs559230848 Chr.12: 123787738-123787739 on Build GRCh38
rs914315044 Chr.12: 123788096-123788096 on Build GRCh38
rs560467277 Chr.12: 123787819-123787819 on Build GRCh38
rs752318669 Chr.12: 123787761-123787761 on Build GRCh38
rs138266985 Chr.12: 123787779-123787779 on Build GRCh38
rs762207670 Chr.12: 123787919-123787919 on Build GRCh38
rs567684796 Chr.12: 123787934-123787934 on Build GRCh38
rs542109616 Chr.12: 123787790-123787790 on Build GRCh38
rs760505364 Chr.12: 123787796-123787796 on Build GRCh38
rs927225904 Chr.12: 123788184-123788184 on Build GRCh38
rs965593256 Chr.12: 123787942-123787942 on Build GRCh38
rs141195166 Chr.12: 123787902-123787902 on Build GRCh38
rs757900300 Chr.12: 123787949-123787949 on Build GRCh38
rs777516903 Chr.12: 123787868-123787868 on Build GRCh38
rs139225647 Chr.12: 123787835-123787835 on Build GRCh38
rs1021697769 Chr.12: 123787982-123787982 on Build GRCh38
rs764477500 Chr.12: 123788008-123788018 on Build GRCh38
rs756788063 Chr.12: 123787855-123787855 on Build GRCh38
rs958788319 Chr.12: 123787732-123787732 on Build GRCh38
rs989002381 Chr.12: 123788094-123788094 on Build GRCh38
rs749377912 Chr.12: 123788025-123788025 on Build GRCh38
rs201388729 Chr.12: 123787959-123787959 on Build GRCh38
rs746375819 Chr.12: 123788158-123788167 on Build GRCh38
rs746846748 Chr.12: 123787870-123787870 on Build GRCh38
rs769655318 Chr.12: 123788015-123788015 on Build GRCh38
rs775439814 Chr.12: 123787896-123787896 on Build GRCh38
rs774535004 Chr.12: 123787906-123787906 on Build GRCh38
rs768691327 Chr.12: 123788037-123788037 on Build GRCh38
rs576504798 Chr.12: 123787749-123787749 on Build GRCh38
rs775045548 Chr.12: 123787748-123787748 on Build GRCh38
rs976928734 Chr.12: 123787948-123787948 on Build GRCh38
rs952993665 Chr.12: 123787739-123787739 on Build GRCh38
rs146557304 Chr.12: 123787889-123787889 on Build GRCh38
rs200152174 Chr.12: 123787935-123787935 on Build GRCh38
rs759048360 Chr.12: 123788009-123788009 on Build GRCh38
rs956857459 Chr.12: 123788093-123788093 on Build GRCh38
rs566079965 Chr.12: 123788157-123788157 on Build GRCh38
rs1031137969 Chr.12: 123788001-123788001 on Build GRCh38
rs768063276 Chr.12: 123787821-123787821 on Build GRCh38
rs750898775 Chr.12: 123787825-123787825 on Build GRCh38
rs775453594 Chr.12: 123788019-123788019 on Build GRCh38
rs552134248 Chr.12: 123787866-123787866 on Build GRCh38

View Full Product Details