Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124953939-124954378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX37
Entrez Gene ID
57647
Gene Name
DEAH-box helicase 37
Gene Aliases
DDX37
UniGene
Hs.107382
Gene Chromosome Location
Chr.12: 124946824-124989125 on Build GRCh38
Transcripts
NM_032656.3 XM_005253590.3 XM_011538598.2 XM_011538600.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
203005 c.2605A>G p.S869G Chr.12: 124953970-124953970 on Build GRCh38
4432695 c.2523G>C p.R841S Chr.12: 124954142-124954142 on Build GRCh38
936794 c.2565C>A p.L855L Chr.12: 124954100-124954100 on Build GRCh38
936793 c.2566A>G p.M856V Chr.12: 124954099-124954099 on Build GRCh38
4853763 c.2514G>T p.Q838H Chr.12: 124954151-124954151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748058922 Chr.12: 124953991-124953991 on Build GRCh38
rs942596185 Chr.12: 124954271-124954271 on Build GRCh38
rs750847950 Chr.12: 124954056-124954056 on Build GRCh38
rs186513005 Chr.12: 124954205-124954205 on Build GRCh38
rs776767613 Chr.12: 124954005-124954005 on Build GRCh38
rs781259803 Chr.12: 124954255-124954255 on Build GRCh38
rs752439923 Chr.12: 124954223-124954223 on Build GRCh38
rs780796755 Chr.12: 124954168-124954168 on Build GRCh38
rs764591153 Chr.12: 124954039-124954039 on Build GRCh38
rs4258464 Chr.12: 124953977-124953977 on Build GRCh38
rs759206908 Chr.12: 124954154-124954154 on Build GRCh38
rs557192637 Chr.12: 124954066-124954066 on Build GRCh38
rs375161391 Chr.12: 124954071-124954071 on Build GRCh38
rs773001817 Chr.12: 124953995-124953995 on Build GRCh38
rs763395834 Chr.12: 124954022-124954022 on Build GRCh38
rs920164504 Chr.12: 124954294-124954294 on Build GRCh38
rs781073177 Chr.12: 124954098-124954098 on Build GRCh38
rs553181503 Chr.12: 124953971-124953971 on Build GRCh38
rs11831506 Chr.12: 124954070-124954070 on Build GRCh38
rs199885185 Chr.12: 124954059-124954059 on Build GRCh38
rs750208899 Chr.12: 124954258-124954258 on Build GRCh38
rs4606565 Chr.12: 124954003-124954003 on Build GRCh38
rs762202883 Chr.12: 124954130-124954130 on Build GRCh38
rs777630341 Chr.12: 124954091-124954091 on Build GRCh38
rs961536319 Chr.12: 124954290-124954290 on Build GRCh38
rs757245596 Chr.12: 124954167-124954167 on Build GRCh38
rs768653623 Chr.12: 124954187-124954187 on Build GRCh38
rs368057884 Chr.12: 124954079-124954079 on Build GRCh38
rs545379306 Chr.12: 124954249-124954249 on Build GRCh38
rs149339717 Chr.12: 124954172-124954172 on Build GRCh38
rs774517204 Chr.12: 124954191-124954191 on Build GRCh38
rs11829165 Chr.12: 124954166-124954166 on Build GRCh38
rs973016488 Chr.12: 124954293-124954293 on Build GRCh38
rs372411913 Chr.12: 124954278-124954278 on Build GRCh38
rs749815996 Chr.12: 124954326-124954326 on Build GRCh38
rs370825217 Chr.12: 124954056-124954056 on Build GRCh38
rs910848384 Chr.12: 124954313-124954313 on Build GRCh38
rs548994653 Chr.12: 124954292-124954292 on Build GRCh38
rs771174000 Chr.12: 124954004-124954004 on Build GRCh38
rs543372890 Chr.12: 124954095-124954095 on Build GRCh38
rs943808392 Chr.12: 124954355-124954355 on Build GRCh38
rs372541666 Chr.12: 124954277-124954277 on Build GRCh38
rs774767034 Chr.12: 124954127-124954127 on Build GRCh38
rs754580190 Chr.12: 124953973-124953973 on Build GRCh38
rs764963329 Chr.12: 124954208-124954208 on Build GRCh38
rs371881147 Chr.12: 124954165-124954165 on Build GRCh38
rs764983929 Chr.12: 124954155-124954155 on Build GRCh38
rs747672861 Chr.12: 124954072-124954072 on Build GRCh38
rs1010225446 Chr.12: 124953992-124953992 on Build GRCh38
rs755949436 Chr.12: 124954262-124954262 on Build GRCh38
rs374743786 Chr.12: 124954006-124954006 on Build GRCh38
rs573793703 Chr.12: 124954160-124954160 on Build GRCh38
rs138006043 Chr.12: 124954139-124954139 on Build GRCh38
rs751886485 Chr.12: 124953967-124953967 on Build GRCh38
rs755499369 Chr.12: 124954116-124954116 on Build GRCh38
rs772157759 Chr.12: 124953994-124953994 on Build GRCh38
rs769190348 Chr.12: 124954126-124954126 on Build GRCh38
rs770021642 Chr.12: 124954106-124954106 on Build GRCh38
rs199924927 Chr.12: 124954195-124954195 on Build GRCh38
rs376497208 Chr.12: 124954206-124954206 on Build GRCh38
rs199731932 Chr.12: 124954174-124954174 on Build GRCh38
rs142687812 Chr.12: 124954161-124954161 on Build GRCh38
rs762727129 Chr.12: 124954233-124954233 on Build GRCh38
rs930988286 Chr.12: 124954296-124954296 on Build GRCh38
rs772647267 Chr.12: 124954196-124954196 on Build GRCh38
rs770672674 Chr.12: 124954096-124954096 on Build GRCh38
rs1051096213 Chr.12: 124954269-124954269 on Build GRCh38
rs886858409 Chr.12: 124954303-124954303 on Build GRCh38
rs370360134 Chr.12: 124954271-124954271 on Build GRCh38
rs11829181 Chr.12: 124954270-124954270 on Build GRCh38
rs748815708 Chr.12: 124953972-124953972 on Build GRCh38
rs764396850 Chr.12: 124953960-124953960 on Build GRCh38
rs367662120 Chr.12: 124954058-124954058 on Build GRCh38
rs749346050 Chr.12: 124954180-124954180 on Build GRCh38
rs375953533 Chr.12: 124954246-124954246 on Build GRCh38
rs139700161 Chr.12: 124954171-124954171 on Build GRCh38
rs560993526 Chr.12: 124954309-124954309 on Build GRCh38
rs758851392 Chr.12: 124954250-124954250 on Build GRCh38
rs767364946 Chr.12: 124954019-124954019 on Build GRCh38
rs4516060 Chr.12: 124953970-124953970 on Build GRCh38
rs946079535 Chr.12: 124954261-124954261 on Build GRCh38
rs763090216 Chr.12: 124954123-124954123 on Build GRCh38
rs765729465 Chr.12: 124954019-124954020 on Build GRCh38
rs767730811 Chr.12: 124954149-124954149 on Build GRCh38
rs746044350 Chr.12: 124954105-124954105 on Build GRCh38
rs750821411 Chr.12: 124954152-124954152 on Build GRCh38
rs200490767 Chr.12: 124954109-124954109 on Build GRCh38

View Full Product Details