Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24600855-24601365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCMAP
Entrez Gene ID
400746
Gene Name
non-compact myelin associated protein
Gene Aliases
C1orf130, MP11
UniGene
Hs.200253
Gene Chromosome Location
Chr.1: 24556076-24609328 on Build GRCh38
Transcripts
NM_001010980.4 XM_005245872.3 XM_011541463.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2153579 c.106G>A p.V36I Chr.1: 24600963-24600963 on Build GRCh38
1198394 c.86C>A p.S29Y Chr.1: 24600943-24600943 on Build GRCh38
3689529 c.148C>A p.L50M Chr.1: 24601005-24601005 on Build GRCh38
7379130 c.98T>C p.V33A Chr.1: 24600955-24600955 on Build GRCh38
6032383 c.113T>C p.V38A Chr.1: 24600970-24600970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777038587 Chr.1: 24601020-24601020 on Build GRCh38
rs1038471791 Chr.1: 24601069-24601069 on Build GRCh38
rs892398866 Chr.1: 24601281-24601281 on Build GRCh38
rs751497680 Chr.1: 24600909-24600909 on Build GRCh38
rs1048549736 Chr.1: 24601149-24601149 on Build GRCh38
rs567192754 Chr.1: 24600939-24600939 on Build GRCh38
rs1030952737 Chr.1: 24600991-24600991 on Build GRCh38
rs548583547 Chr.1: 24600913-24600913 on Build GRCh38
rs528883787 Chr.1: 24600908-24600908 on Build GRCh38
rs569598945 Chr.1: 24601139-24601143 on Build GRCh38
rs752164623 Chr.1: 24600914-24600914 on Build GRCh38
rs371911941 Chr.1: 24601058-24601058 on Build GRCh38
rs901046850 Chr.1: 24601226-24601226 on Build GRCh38
rs969499304 Chr.1: 24601331-24601331 on Build GRCh38
rs756907584 Chr.1: 24601059-24601059 on Build GRCh38
rs146873574 Chr.1: 24600879-24600879 on Build GRCh38
rs898754856 Chr.1: 24600891-24600891 on Build GRCh38
rs556724113 Chr.1: 24601095-24601095 on Build GRCh38
rs142867139 Chr.1: 24600963-24600963 on Build GRCh38
rs559061502 Chr.1: 24601035-24601035 on Build GRCh38
rs779190565 Chr.1: 24601066-24601066 on Build GRCh38
rs542282779 Chr.1: 24601116-24601116 on Build GRCh38
rs760761838 Chr.1: 24601330-24601330 on Build GRCh38
rs767414418 Chr.1: 24601034-24601034 on Build GRCh38
rs775435956 Chr.1: 24600889-24600889 on Build GRCh38
rs928163698 Chr.1: 24601110-24601110 on Build GRCh38
rs759463152 Chr.1: 24601031-24601031 on Build GRCh38
rs768966470 Chr.1: 24601004-24601004 on Build GRCh38
rs753266077 Chr.1: 24601275-24601275 on Build GRCh38
rs1039971307 Chr.1: 24600961-24600961 on Build GRCh38
rs775625305 Chr.1: 24600978-24600978 on Build GRCh38
rs117081131 Chr.1: 24600907-24600907 on Build GRCh38
rs527942993 Chr.1: 24601138-24601138 on Build GRCh38
rs919512993 Chr.1: 24601122-24601122 on Build GRCh38
rs56316061 Chr.1: 24601240-24601240 on Build GRCh38
rs147452276 Chr.1: 24600955-24600955 on Build GRCh38
rs774025806 Chr.1: 24601029-24601029 on Build GRCh38
rs372348645 Chr.1: 24601028-24601028 on Build GRCh38
rs76868886 Chr.1: 24600973-24600973 on Build GRCh38
rs4378146 Chr.1: 24601134-24601134 on Build GRCh38
rs764466662 Chr.1: 24600904-24600904 on Build GRCh38
rs755662758 Chr.1: 24601052-24601052 on Build GRCh38
rs755779908 Chr.1: 24600915-24600915 on Build GRCh38
rs113221702 Chr.1: 24601014-24601014 on Build GRCh38
rs942466993 Chr.1: 24601206-24601206 on Build GRCh38
rs750235923 Chr.1: 24600905-24600905 on Build GRCh38
rs762145101 Chr.1: 24601024-24601024 on Build GRCh38
rs760892207 Chr.1: 24600894-24600894 on Build GRCh38
rs772469044 Chr.1: 24600974-24600974 on Build GRCh38
rs1008300002 Chr.1: 24600921-24600921 on Build GRCh38
rs55691364 Chr.1: 24601236-24601236 on Build GRCh38
rs4553122 Chr.1: 24601070-24601070 on Build GRCh38
rs774829761 Chr.1: 24601158-24601158 on Build GRCh38
rs1049958539 Chr.1: 24601102-24601102 on Build GRCh38
rs754952110 Chr.1: 24601308-24601308 on Build GRCh38
rs909619930 Chr.1: 24601154-24601154 on Build GRCh38
rs754906607 Chr.1: 24600912-24600912 on Build GRCh38
rs969888072 Chr.1: 24601068-24601068 on Build GRCh38
rs531426734 Chr.1: 24601290-24601290 on Build GRCh38
rs574871483 Chr.1: 24601108-24601108 on Build GRCh38
rs908295580 Chr.1: 24601055-24601055 on Build GRCh38
rs538276712 Chr.1: 24601286-24601289 on Build GRCh38
rs747122535 Chr.1: 24600992-24600992 on Build GRCh38
rs757667971 Chr.1: 24600962-24600962 on Build GRCh38
rs758937172 Chr.1: 24601075-24601075 on Build GRCh38
rs777470341 Chr.1: 24600922-24600922 on Build GRCh38
rs891343938 Chr.1: 24601107-24601107 on Build GRCh38
rs1039535269 Chr.1: 24601212-24601213 on Build GRCh38

View Full Product Details