Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26469206-26469707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHDDS
Entrez Gene ID
79947
Gene Name
dehydrodolichyl diphosphate synthase subunit
Gene Aliases
CIT, CPT, DS, HDS, RP59
UniGene
Hs.369385
Gene Chromosome Location
Chr.1: 26432282-26471306 on Build GRCh38
Transcripts
NM_001243564.1 NM_001243565.1 NM_001319959.1 NM_024887.3 NM_205861.2 XM_006710912.2 XM_006710913.2 XM_006710914.2 XM_006710915.2 XM_006710916.2 XM_006710917.2 XM_006710918.2 XM_011542183.2 XM_011542184.2 XM_011542185.2 XM_011542186.2 XM_017002379.1 XM_017002380.1 XM_017002381.1 XM_017002382.1 XM_017002383.1 XM_017002384.1

SNP Detail

SNP ID Chromosome Location
rs552046225 Chr.1: 26469611-26469611 on Build GRCh38
rs1057515469 Chr.1: 26469443-26469443 on Build GRCh38
rs933126536 Chr.1: 26469494-26469494 on Build GRCh38
rs756248551 Chr.1: 26469275-26469275 on Build GRCh38
rs761300466 Chr.1: 26469512-26469512 on Build GRCh38
rs551261460 Chr.1: 26469441-26469441 on Build GRCh38
rs187250338 Chr.1: 26469254-26469254 on Build GRCh38
rs553705354 Chr.1: 26469609-26469609 on Build GRCh38
rs983520261 Chr.1: 26469342-26469342 on Build GRCh38
rs571731712 Chr.1: 26469501-26469501 on Build GRCh38
rs74979801 Chr.1: 26469504-26469504 on Build GRCh38
rs1057515436 Chr.1: 26469395-26469395 on Build GRCh38
rs1042787798 Chr.1: 26469558-26469558 on Build GRCh38
rs1032593244 Chr.1: 26469240-26469240 on Build GRCh38
rs572009346 Chr.1: 26469535-26469535 on Build GRCh38
rs1000926401 Chr.1: 26469605-26469606 on Build GRCh38
rs536605809 Chr.1: 26469598-26469598 on Build GRCh38
rs536975761 Chr.1: 26469264-26469264 on Build GRCh38
rs981126429 Chr.1: 26469374-26469374 on Build GRCh38
rs970147808 Chr.1: 26469349-26469349 on Build GRCh38
rs567665048 Chr.1: 26469595-26469595 on Build GRCh38
rs1002494844 Chr.1: 26469652-26469652 on Build GRCh38
rs931273148 Chr.1: 26469499-26469499 on Build GRCh38
rs963136115 Chr.1: 26469405-26469405 on Build GRCh38
rs1035397746 Chr.1: 26469678-26469678 on Build GRCh38
rs961152986 Chr.1: 26469434-26469434 on Build GRCh38
rs754078948 Chr.1: 26469454-26469454 on Build GRCh38
rs919835526 Chr.1: 26469488-26469488 on Build GRCh38
rs944044952 Chr.1: 26469537-26469537 on Build GRCh38
rs140277372 Chr.1: 26469550-26469550 on Build GRCh38
rs950449228 Chr.1: 26469316-26469318 on Build GRCh38
rs747837144 Chr.1: 26469646-26469646 on Build GRCh38
rs180690900 Chr.1: 26469564-26469564 on Build GRCh38
rs921784315 Chr.1: 26469465-26469465 on Build GRCh38
rs1024319488 Chr.1: 26469621-26469621 on Build GRCh38
rs895348953 Chr.1: 26469623-26469623 on Build GRCh38
rs1057515546 Chr.1: 26469305-26469305 on Build GRCh38
rs189263364 Chr.1: 26469430-26469430 on Build GRCh38
rs780471395 Chr.1: 26469447-26469447 on Build GRCh38
rs1025265736 Chr.1: 26469663-26469663 on Build GRCh38
rs1022655060 Chr.1: 26469305-26469305 on Build GRCh38
rs146706631 Chr.1: 26469259-26469259 on Build GRCh38
rs767843924 Chr.1: 26469242-26469242 on Build GRCh38
rs902639485 Chr.1: 26469554-26469554 on Build GRCh38

View Full Product Details