Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9032209-9032720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9B
Entrez Gene ID
171483
Gene Name
family with sequence similarity 9 member B
Gene Aliases
TEX39B
UniGene
Hs.733351
Gene Chromosome Location
Chr.X: 9024232-9197176 on Build GRCh38
Transcripts
NM_205849.3 XM_011545464.2 XM_011545465.2

SNP Detail

SNP ID Chromosome Location
rs774998270 Chr.X: 9032310-9032310 on Build GRCh38
rs773009993 Chr.X: 9032351-9032351 on Build GRCh38
rs762329143 Chr.X: 9032354-9032354 on Build GRCh38
rs200710909 Chr.X: 9032422-9032422 on Build GRCh38
rs371784822 Chr.X: 9032473-9032473 on Build GRCh38
rs200449273 Chr.X: 9032554-9032554 on Build GRCh38
rs747575371 Chr.X: 9032486-9032486 on Build GRCh38
rs761494466 Chr.X: 9032324-9032324 on Build GRCh38
rs766362562 Chr.X: 9032370-9032370 on Build GRCh38
rs780593316 Chr.X: 9032461-9032461 on Build GRCh38
rs764409804 Chr.X: 9032344-9032344 on Build GRCh38
rs796909241 Chr.X: 9032564-9032565 on Build GRCh38
rs1019589796 Chr.X: 9032559-9032559 on Build GRCh38
rs768136875 Chr.X: 9032309-9032309 on Build GRCh38
rs925772419 Chr.X: 9032244-9032244 on Build GRCh38
rs777302447 Chr.X: 9032557-9032557 on Build GRCh38
rs768195808 Chr.X: 9032462-9032462 on Build GRCh38
rs766734802 Chr.X: 9032554-9032556 on Build GRCh38
rs905986127 Chr.X: 9032441-9032441 on Build GRCh38
rs780730044 Chr.X: 9032463-9032463 on Build GRCh38
rs5934524 Chr.X: 9032690-9032690 on Build GRCh38
rs770845783 Chr.X: 9032496-9032496 on Build GRCh38
rs370616228 Chr.X: 9032562-9032564 on Build GRCh38
rs760825913 Chr.X: 9032607-9032607 on Build GRCh38
rs756901678 Chr.X: 9032560-9032560 on Build GRCh38
rs762901395 Chr.X: 9032489-9032489 on Build GRCh38
rs757803656 Chr.X: 9032555-9032555 on Build GRCh38
rs5979009 Chr.X: 9032280-9032280 on Build GRCh38
rs746163014 Chr.X: 9032445-9032445 on Build GRCh38
rs375440422 Chr.X: 9032308-9032308 on Build GRCh38
rs934627743 Chr.X: 9032255-9032255 on Build GRCh38
rs900849801 Chr.X: 9032564-9032564 on Build GRCh38
rs796307197 Chr.X: 9032554-9032555 on Build GRCh38
rs917262179 Chr.X: 9032330-9032330 on Build GRCh38
rs745317637 Chr.X: 9032246-9032246 on Build GRCh38
rs772558643 Chr.X: 9032298-9032298 on Build GRCh38
rs142867505 Chr.X: 9032449-9032449 on Build GRCh38
rs769445974 Chr.X: 9032480-9032480 on Build GRCh38
rs779179061 Chr.X: 9032439-9032439 on Build GRCh38
rs756328331 Chr.X: 9032397-9032397 on Build GRCh38
rs184865805 Chr.X: 9032502-9032502 on Build GRCh38
rs759528063 Chr.X: 9032511-9032511 on Build GRCh38
rs772897944 Chr.X: 9032486-9032486 on Build GRCh38
rs998288363 Chr.X: 9032632-9032632 on Build GRCh38
rs879003606 Chr.X: 9032556-9032556 on Build GRCh38
rs757454422 Chr.X: 9032438-9032438 on Build GRCh38
rs768051414 Chr.X: 9032553-9032554 on Build GRCh38
rs369906919 Chr.X: 9032356-9032356 on Build GRCh38
rs751530233 Chr.X: 9032383-9032383 on Build GRCh38
rs111614282 Chr.X: 9032554-9032554 on Build GRCh38
rs113943818 Chr.X: 9032348-9032348 on Build GRCh38
rs746725564 Chr.X: 9032558-9032558 on Build GRCh38
rs996091308 Chr.X: 9032437-9032437 on Build GRCh38
rs878943286 Chr.X: 9032364-9032364 on Build GRCh38
rs5979010 Chr.X: 9032660-9032660 on Build GRCh38
rs745607663 Chr.X: 9032685-9032685 on Build GRCh38
rs772525351 Chr.X: 9032298-9032298 on Build GRCh38

View Full Product Details