Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9032805-9033347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9B
Entrez Gene ID
171483
Gene Name
family with sequence similarity 9 member B
Gene Aliases
TEX39B
UniGene
Hs.733351
Gene Chromosome Location
Chr.X: 9024232-9197176 on Build GRCh38
Transcripts
NM_205849.3 XM_011545464.2 XM_011545465.2

SNP Detail

SNP ID Chromosome Location
rs180695457 Chr.X: 9033290-9033290 on Build GRCh38
rs750679970 Chr.X: 9033024-9033024 on Build GRCh38
rs755462433 Chr.X: 9032940-9032940 on Build GRCh38
rs755443091 Chr.X: 9033064-9033064 on Build GRCh38
rs5933708 Chr.X: 9032851-9032851 on Build GRCh38
rs371572820 Chr.X: 9032909-9032909 on Build GRCh38
rs188156078 Chr.X: 9032900-9032900 on Build GRCh38
rs773227423 Chr.X: 9033136-9033136 on Build GRCh38
rs766519675 Chr.X: 9033158-9033158 on Build GRCh38
rs763139671 Chr.X: 9033025-9033025 on Build GRCh38
rs373460079 Chr.X: 9032960-9032960 on Build GRCh38
rs971760544 Chr.X: 9033322-9033322 on Build GRCh38
rs769174837 Chr.X: 9032959-9032959 on Build GRCh38
rs767743161 Chr.X: 9033169-9033169 on Build GRCh38
rs902114625 Chr.X: 9033197-9033197 on Build GRCh38
rs776585457 Chr.X: 9032986-9032986 on Build GRCh38
rs774242312 Chr.X: 9032968-9032968 on Build GRCh38
rs779876689 Chr.X: 9033282-9033282 on Build GRCh38
rs755902915 Chr.X: 9033226-9033226 on Build GRCh38
rs761942596 Chr.X: 9032914-9032914 on Build GRCh38
rs765215414 Chr.X: 9033164-9033164 on Build GRCh38
rs775560253 Chr.X: 9033118-9033118 on Build GRCh38
rs765438760 Chr.X: 9032922-9032922 on Build GRCh38
rs780775539 Chr.X: 9033172-9033172 on Build GRCh38
rs760014502 Chr.X: 9033165-9033165 on Build GRCh38
rs753141625 Chr.X: 9033068-9033068 on Build GRCh38
rs771748153 Chr.X: 9033109-9033109 on Build GRCh38
rs777400675 Chr.X: 9033071-9033071 on Build GRCh38
rs780596472 Chr.X: 9032942-9032942 on Build GRCh38
rs750265003 Chr.X: 9033186-9033186 on Build GRCh38
rs770902868 Chr.X: 9032966-9032966 on Build GRCh38
rs760079615 Chr.X: 9032912-9032912 on Build GRCh38
rs752606889 Chr.X: 9033254-9033254 on Build GRCh38
rs1010516017 Chr.X: 9033319-9033319 on Build GRCh38
rs1024124537 Chr.X: 9033321-9033321 on Build GRCh38
rs760897197 Chr.X: 9032884-9032912 on Build GRCh38
rs72611469 Chr.X: 9033123-9033123 on Build GRCh38
rs748974724 Chr.X: 9033084-9033084 on Build GRCh38
rs369036793 Chr.X: 9032932-9032932 on Build GRCh38
rs996425870 Chr.X: 9033240-9033240 on Build GRCh38
rs751006711 Chr.X: 9033085-9033085 on Build GRCh38
rs759581024 Chr.X: 9032981-9032981 on Build GRCh38
rs772351987 Chr.X: 9032847-9032847 on Build GRCh38
rs375029458 Chr.X: 9032910-9032910 on Build GRCh38
rs762007446 Chr.X: 9033000-9033000 on Build GRCh38
rs988764513 Chr.X: 9032828-9032828 on Build GRCh38
rs267606528 Chr.X: 9033065-9033065 on Build GRCh38
rs748917729 Chr.X: 9032965-9032965 on Build GRCh38
rs745621193 Chr.X: 9033106-9033106 on Build GRCh38
rs1032740447 Chr.X: 9033257-9033257 on Build GRCh38
rs768634326 Chr.X: 9033301-9033301 on Build GRCh38
rs755689969 Chr.X: 9033069-9033069 on Build GRCh38
rs750627252 Chr.X: 9032928-9032928 on Build GRCh38
rs765224028 Chr.X: 9033022-9033022 on Build GRCh38
rs999950662 Chr.X: 9033255-9033255 on Build GRCh38
rs1049454503 Chr.X: 9033245-9033245 on Build GRCh38
rs763664402 Chr.X: 9033044-9033044 on Build GRCh38
rs375803513 Chr.X: 9032870-9032870 on Build GRCh38
rs979981690 Chr.X: 9033323-9033323 on Build GRCh38
rs746887111 Chr.X: 9033122-9033122 on Build GRCh38
rs780364669 Chr.X: 9032934-9032934 on Build GRCh38
rs747638327 Chr.X: 9032948-9032948 on Build GRCh38
rs752108331 Chr.X: 9032938-9032938 on Build GRCh38
rs377436464 Chr.X: 9033056-9033056 on Build GRCh38
rs763192649 Chr.X: 9033157-9033157 on Build GRCh38
rs917127755 Chr.X: 9032858-9032858 on Build GRCh38

View Full Product Details