Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130911713-130912151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIBCD1
Entrez Gene ID
84929
Gene Name
fibrinogen C domain containing 1
Gene Aliases
-
UniGene
Hs.133205
Gene Chromosome Location
Chr.9: 130902438-130939068 on Build GRCh38
Transcripts
NM_001145106.1 NM_032843.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368276 c.907C>T p.R303* Chr.9: 130911831-130911831 on Build GRCh38
3718725 c.768C>G p.Y256* Chr.9: 130911831-130911831 on Build GRCh38
6323295 c.902C>T p.A301V Chr.9: 130911836-130911836 on Build GRCh38
239877 c.860G>C p.R287P Chr.9: 130911878-130911878 on Build GRCh38
6020777 c.723C>T p.R241R Chr.9: 130911876-130911876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040732143 Chr.9: 130911997-130911997 on Build GRCh38
rs765223888 Chr.9: 130911876-130911876 on Build GRCh38
rs769347517 Chr.9: 130911817-130911817 on Build GRCh38
rs1012245597 Chr.9: 130911760-130911760 on Build GRCh38
rs7035155 Chr.9: 130911961-130911961 on Build GRCh38
rs770468815 Chr.9: 130911973-130911973 on Build GRCh38
rs528592841 Chr.9: 130912095-130912095 on Build GRCh38
rs773144212 Chr.9: 130911868-130911868 on Build GRCh38
rs1008345190 Chr.9: 130911992-130911992 on Build GRCh38
rs763876802 Chr.9: 130911787-130911787 on Build GRCh38
rs1020793575 Chr.9: 130911743-130911743 on Build GRCh38
rs142317287 Chr.9: 130911870-130911870 on Build GRCh38
rs555125900 Chr.9: 130912013-130912013 on Build GRCh38
rs769443515 Chr.9: 130911849-130911849 on Build GRCh38
rs924779318 Chr.9: 130912085-130912085 on Build GRCh38
rs779807050 Chr.9: 130911886-130911886 on Build GRCh38
rs768960152 Chr.9: 130911757-130911757 on Build GRCh38
rs1050067423 Chr.9: 130911972-130911972 on Build GRCh38
rs377304322 Chr.9: 130911770-130911770 on Build GRCh38
rs925141690 Chr.9: 130912043-130912043 on Build GRCh38
rs767769869 Chr.9: 130911940-130911940 on Build GRCh38
rs376857307 Chr.9: 130911923-130911923 on Build GRCh38
rs925087114 Chr.9: 130911965-130911965 on Build GRCh38
rs912127805 Chr.9: 130912093-130912093 on Build GRCh38
rs550474087 Chr.9: 130911837-130911837 on Build GRCh38
rs936465815 Chr.9: 130912055-130912055 on Build GRCh38
rs1008062686 Chr.9: 130911890-130911890 on Build GRCh38
rs762300224 Chr.9: 130911820-130911820 on Build GRCh38
rs750650927 Chr.9: 130911797-130911797 on Build GRCh38
rs190575249 Chr.9: 130911988-130911988 on Build GRCh38
rs199643737 Chr.9: 130911848-130911848 on Build GRCh38
rs756213471 Chr.9: 130911748-130911748 on Build GRCh38
rs896296414 Chr.9: 130911735-130911735 on Build GRCh38
rs780867280 Chr.9: 130911808-130911808 on Build GRCh38
rs548275524 Chr.9: 130912127-130912127 on Build GRCh38
rs950517943 Chr.9: 130912025-130912025 on Build GRCh38
rs758922636 Chr.9: 130911878-130911878 on Build GRCh38
rs368873188 Chr.9: 130911774-130911774 on Build GRCh38
rs754584372 Chr.9: 130911895-130911895 on Build GRCh38
rs999078279 Chr.9: 130911871-130911871 on Build GRCh38
rs753167205 Chr.9: 130911843-130911843 on Build GRCh38
rs201574586 Chr.9: 130911924-130911924 on Build GRCh38
rs559677479 Chr.9: 130912039-130912039 on Build GRCh38
rs149922280 Chr.9: 130911826-130911826 on Build GRCh38
rs866063692 Chr.9: 130911814-130911814 on Build GRCh38
rs1052054752 Chr.9: 130912089-130912089 on Build GRCh38
rs776008115 Chr.9: 130911989-130911989 on Build GRCh38
rs780755370 Chr.9: 130911750-130911750 on Build GRCh38
rs772166026 Chr.9: 130911903-130911903 on Build GRCh38
rs760073718 Chr.9: 130911750-130911752 on Build GRCh38
rs889465108 Chr.9: 130911985-130911985 on Build GRCh38
rs552218149 Chr.9: 130911958-130911958 on Build GRCh38
rs958997550 Chr.9: 130911927-130911927 on Build GRCh38
rs745511635 Chr.9: 130911815-130911815 on Build GRCh38
rs371263618 Chr.9: 130911807-130911807 on Build GRCh38
rs144181457 Chr.9: 130911831-130911831 on Build GRCh38
rs750656149 Chr.9: 130911805-130911805 on Build GRCh38
rs990483042 Chr.9: 130912037-130912037 on Build GRCh38
rs918039525 Chr.9: 130911949-130911949 on Build GRCh38
rs6597650 Chr.9: 130911838-130911838 on Build GRCh38
rs978206458 Chr.9: 130912067-130912067 on Build GRCh38
rs763277435 Chr.9: 130912036-130912036 on Build GRCh38
rs748861995 Chr.9: 130911891-130911891 on Build GRCh38
rs1017855522 Chr.9: 130911823-130911823 on Build GRCh38
rs147669758 Chr.9: 130911861-130911861 on Build GRCh38
rs759172126 Chr.9: 130912026-130912026 on Build GRCh38
rs559408176 Chr.9: 130911738-130911738 on Build GRCh38
rs775076933 Chr.9: 130911819-130911819 on Build GRCh38
rs369193753 Chr.9: 130911867-130911867 on Build GRCh38
rs779520322 Chr.9: 130911842-130911842 on Build GRCh38
rs762983731 Chr.9: 130911918-130911918 on Build GRCh38
rs180860396 Chr.9: 130912033-130912033 on Build GRCh38
rs943953897 Chr.9: 130912120-130912120 on Build GRCh38
rs56126622 Chr.9: 130911769-130911769 on Build GRCh38
rs372918822 Chr.9: 130911902-130911902 on Build GRCh38
rs541870250 Chr.9: 130911780-130911780 on Build GRCh38
rs568093970 Chr.9: 130911873-130911873 on Build GRCh38
rs552451805 Chr.9: 130911847-130911847 on Build GRCh38
rs752104263 Chr.9: 130911745-130911745 on Build GRCh38
rs1001176261 Chr.9: 130911859-130911859 on Build GRCh38
rs770597300 Chr.9: 130911910-130911910 on Build GRCh38
rs757517191 Chr.9: 130911883-130911883 on Build GRCh38
rs914862943 Chr.9: 130912044-130912044 on Build GRCh38
rs750732451 Chr.9: 130911835-130911835 on Build GRCh38
rs375155907 Chr.9: 130911906-130911906 on Build GRCh38
rs770604739 Chr.9: 130911761-130911761 on Build GRCh38
rs564014410 Chr.9: 130912014-130912014 on Build GRCh38
rs372500939 Chr.9: 130911939-130911939 on Build GRCh38
rs1049575845 Chr.9: 130912068-130912068 on Build GRCh38
rs952289131 Chr.9: 130912010-130912010 on Build GRCh38
rs368351261 Chr.9: 130911752-130911752 on Build GRCh38
rs375911377 Chr.9: 130911862-130911862 on Build GRCh38
rs556831334 Chr.9: 130911914-130911914 on Build GRCh38
rs768667343 Chr.9: 130911825-130911825 on Build GRCh38
rs943394735 Chr.9: 130912109-130912109 on Build GRCh38
rs746977133 Chr.9: 130911966-130911966 on Build GRCh38
rs142360752 Chr.9: 130911877-130911877 on Build GRCh38
rs775352223 Chr.9: 130911777-130911777 on Build GRCh38
rs199554608 Chr.9: 130911915-130911915 on Build GRCh38
rs778299636 Chr.9: 130911899-130911899 on Build GRCh38
rs775555444 Chr.9: 130911917-130911917 on Build GRCh38
rs751954546 Chr.9: 130911879-130911879 on Build GRCh38
rs755514768 Chr.9: 130911804-130911804 on Build GRCh38
rs796767271 Chr.9: 130912002-130912002 on Build GRCh38
rs574982274 Chr.9: 130912034-130912034 on Build GRCh38
rs753982129 Chr.9: 130911872-130911872 on Build GRCh38
rs748396074 Chr.9: 130911759-130911759 on Build GRCh38
rs373150753 Chr.9: 130911911-130911911 on Build GRCh38
rs780268824 Chr.9: 130911800-130911800 on Build GRCh38
rs757075901 Chr.9: 130911796-130911796 on Build GRCh38
rs890420744 Chr.9: 130911883-130911884 on Build GRCh38
rs565623490 Chr.9: 130912003-130912003 on Build GRCh38
rs986924724 Chr.9: 130911784-130911784 on Build GRCh38
rs765585062 Chr.9: 130911768-130911769 on Build GRCh38
rs532727695 Chr.9: 130911846-130911846 on Build GRCh38
rs1039049419 Chr.9: 130912122-130912122 on Build GRCh38
rs760505314 Chr.9: 130911869-130911869 on Build GRCh38
rs145385892 Chr.9: 130911850-130911850 on Build GRCh38
rs11794418 Chr.9: 130912011-130912011 on Build GRCh38
rs934901744 Chr.9: 130911987-130911987 on Build GRCh38
rs762931223 Chr.9: 130911781-130911781 on Build GRCh38
rs945444955 Chr.9: 130911955-130911955 on Build GRCh38
rs911170547 Chr.9: 130912100-130912100 on Build GRCh38
rs761838548 Chr.9: 130911866-130911866 on Build GRCh38
rs766443740 Chr.9: 130911836-130911836 on Build GRCh38
rs947570804 Chr.9: 130911962-130911962 on Build GRCh38
rs1051980121 Chr.9: 130911995-130911995 on Build GRCh38
rs934805199 Chr.9: 130912088-130912088 on Build GRCh38
rs751371909 Chr.9: 130911792-130911792 on Build GRCh38
rs749270628 Chr.9: 130911801-130911801 on Build GRCh38
rs896934089 Chr.9: 130912001-130912001 on Build GRCh38

View Full Product Details