Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132542854-132543353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP77
Entrez Gene ID
389799
Gene Name
cilia and flagella associated protein 77
Gene Aliases
C9orf171
UniGene
Hs.201709
Gene Chromosome Location
Chr.9: 132410043-132573319 on Build GRCh38
Transcripts
NM_001282957.1 NM_207417.2 XM_011518670.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3212563 c.743T>C p.V248A Chr.9: 132542950-132542950 on Build GRCh38
69969 c.796C>A p.P266T Chr.9: 132543003-132543003 on Build GRCh38
6550562 c.742G>T p.V248F Chr.9: 132542949-132542949 on Build GRCh38
6213401 c.748G>C p.G250R Chr.9: 132542955-132542955 on Build GRCh38
6616858 c.800T>C p.V267A Chr.9: 132543007-132543007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376031788 Chr.9: 132543095-132543095 on Build GRCh38
rs966529766 Chr.9: 132543296-132543296 on Build GRCh38
rs367948498 Chr.9: 132543149-132543149 on Build GRCh38
rs753565718 Chr.9: 132543072-132543072 on Build GRCh38
rs375517993 Chr.9: 132543163-132543164 on Build GRCh38
rs367674163 Chr.9: 132542988-132542988 on Build GRCh38
rs763954954 Chr.9: 132542951-132542951 on Build GRCh38
rs766142719 Chr.9: 132543001-132543001 on Build GRCh38
rs1040447847 Chr.9: 132543054-132543054 on Build GRCh38
rs375418327 Chr.9: 132543215-132543215 on Build GRCh38
rs1050507589 Chr.9: 132543326-132543326 on Build GRCh38
rs55720639 Chr.9: 132543066-132543066 on Build GRCh38
rs201630263 Chr.9: 132542973-132542973 on Build GRCh38
rs773286184 Chr.9: 132542931-132542931 on Build GRCh38
rs1024386815 Chr.9: 132543181-132543181 on Build GRCh38
rs749555389 Chr.9: 132543310-132543310 on Build GRCh38
rs563639643 Chr.9: 132542974-132542974 on Build GRCh38
rs376871470 Chr.9: 132543027-132543027 on Build GRCh38
rs34939624 Chr.9: 132543040-132543041 on Build GRCh38
rs765676695 Chr.9: 132543076-132543076 on Build GRCh38
rs778110990 Chr.9: 132542988-132542988 on Build GRCh38
rs532808059 Chr.9: 132543180-132543180 on Build GRCh38
rs372180431 Chr.9: 132543015-132543015 on Build GRCh38
rs55777723 Chr.9: 132543164-132543164 on Build GRCh38
rs1000463178 Chr.9: 132543153-132543153 on Build GRCh38
rs887759062 Chr.9: 132543328-132543328 on Build GRCh38
rs984038354 Chr.9: 132543218-132543218 on Build GRCh38
rs61308567 Chr.9: 132543002-132543002 on Build GRCh38
rs374197369 Chr.9: 132542916-132542916 on Build GRCh38
rs772719147 Chr.9: 132543058-132543058 on Build GRCh38
rs920750392 Chr.9: 132543033-132543033 on Build GRCh38
rs748136073 Chr.9: 132542919-132542919 on Build GRCh38
rs755374000 Chr.9: 132543073-132543073 on Build GRCh38
rs142726335 Chr.9: 132543006-132543006 on Build GRCh38
rs768486195 Chr.9: 132543022-132543022 on Build GRCh38
rs904354662 Chr.9: 132543152-132543152 on Build GRCh38
rs946336627 Chr.9: 132543298-132543298 on Build GRCh38
rs932567845 Chr.9: 132543112-132543112 on Build GRCh38
rs78783314 Chr.9: 132542920-132542920 on Build GRCh38
rs868400778 Chr.9: 132543008-132543008 on Build GRCh38
rs776847264 Chr.9: 132543017-132543017 on Build GRCh38
rs969742829 Chr.9: 132543183-132543183 on Build GRCh38
rs112763937 Chr.9: 132543255-132543255 on Build GRCh38
rs935692676 Chr.9: 132543264-132543264 on Build GRCh38
rs369040131 Chr.9: 132543044-132543044 on Build GRCh38
rs944694340 Chr.9: 132543070-132543070 on Build GRCh38
rs1040903116 Chr.9: 132543304-132543304 on Build GRCh38
rs749045226 Chr.9: 132543019-132543019 on Build GRCh38
rs761872564 Chr.9: 132543038-132543038 on Build GRCh38
rs753141286 Chr.9: 132543082-132543082 on Build GRCh38
rs759002753 Chr.9: 132543094-132543094 on Build GRCh38
rs961991848 Chr.9: 132542889-132542889 on Build GRCh38
rs148870688 Chr.9: 132543005-132543005 on Build GRCh38
rs147829744 Chr.9: 132543004-132543004 on Build GRCh38
rs901900149 Chr.9: 132543065-132543065 on Build GRCh38
rs971517143 Chr.9: 132543178-132543178 on Build GRCh38
rs779238098 Chr.9: 132543003-132543003 on Build GRCh38
rs766087714 Chr.9: 132543071-132543071 on Build GRCh38
rs56229164 Chr.9: 132542942-132542942 on Build GRCh38
rs1028915671 Chr.9: 132543101-132543101 on Build GRCh38
rs891554168 Chr.9: 132543169-132543169 on Build GRCh38
rs369216787 Chr.9: 132542896-132542896 on Build GRCh38
rs551417708 Chr.9: 132543217-132543217 on Build GRCh38
rs751549774 Chr.9: 132542966-132542966 on Build GRCh38
rs35315271 Chr.9: 132542999-132543000 on Build GRCh38
rs779564995 Chr.9: 132542918-132542918 on Build GRCh38
rs760997701 Chr.9: 132542934-132542934 on Build GRCh38
rs932014966 Chr.9: 132543324-132543324 on Build GRCh38
rs1001771265 Chr.9: 132543193-132543193 on Build GRCh38
rs749798662 Chr.9: 132542908-132542908 on Build GRCh38
rs577981040 Chr.9: 132542939-132542939 on Build GRCh38
rs976232430 Chr.9: 132543299-132543299 on Build GRCh38
rs199758334 Chr.9: 132542907-132542907 on Build GRCh38
rs1054696869 Chr.9: 132543292-132543292 on Build GRCh38
rs370753369 Chr.9: 132542905-132542905 on Build GRCh38
rs368494178 Chr.9: 132543096-132543096 on Build GRCh38

View Full Product Details