Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154010994-154011487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK1
Entrez Gene ID
3654
Gene Name
interleukin 1 receptor associated kinase 1
Gene Aliases
IRAK, pelle
UniGene
Hs.522819
Gene Chromosome Location
Chr.X: 154010506-154019906 on Build GRCh38
Transcripts
NM_001025242.1 NM_001025243.1 NM_001569.3 XM_005274668.3 XM_011531158.2

SNP Detail

SNP ID Chromosome Location
rs782308632 Chr.X: 154011170-154011170 on Build GRCh38
rs1041558395 Chr.X: 154011386-154011386 on Build GRCh38
rs900966517 Chr.X: 154011379-154011379 on Build GRCh38
rs782275173 Chr.X: 154011100-154011100 on Build GRCh38
rs959187249 Chr.X: 154011229-154011229 on Build GRCh38
rs904605468 Chr.X: 154011391-154011391 on Build GRCh38
rs983774311 Chr.X: 154011258-154011258 on Build GRCh38
rs185172267 Chr.X: 154011450-154011450 on Build GRCh38
rs1014887397 Chr.X: 154011436-154011436 on Build GRCh38
rs1020330734 Chr.X: 154011179-154011179 on Build GRCh38
rs1803779 Chr.X: 154011069-154011069 on Build GRCh38
rs1000137506 Chr.X: 154011199-154011199 on Build GRCh38
rs1000252339 Chr.X: 154011398-154011398 on Build GRCh38
rs182367373 Chr.X: 154011242-154011242 on Build GRCh38
rs901021144 Chr.X: 154011128-154011128 on Build GRCh38
rs904657738 Chr.X: 154011193-154011193 on Build GRCh38
rs910901502 Chr.X: 154011313-154011313 on Build GRCh38
rs782722587 Chr.X: 154011380-154011380 on Build GRCh38
rs782105100 Chr.X: 154011044-154011044 on Build GRCh38
rs918337813 Chr.X: 154011250-154011250 on Build GRCh38
rs931073457 Chr.X: 154011254-154011254 on Build GRCh38
rs11556421 Chr.X: 154011115-154011115 on Build GRCh38
rs781893812 Chr.X: 154011432-154011432 on Build GRCh38
rs993106750 Chr.X: 154011230-154011230 on Build GRCh38
rs782229055 Chr.X: 154011088-154011088 on Build GRCh38
rs782442824 Chr.X: 154011263-154011263 on Build GRCh38
rs942658174 Chr.X: 154011027-154011027 on Build GRCh38
rs1055699592 Chr.X: 154011403-154011403 on Build GRCh38
rs782323844 Chr.X: 154011162-154011162 on Build GRCh38
rs782287578 Chr.X: 154011372-154011372 on Build GRCh38
rs1034334232 Chr.X: 154011210-154011210 on Build GRCh38
rs942588241 Chr.X: 154011370-154011370 on Build GRCh38
rs894480859 Chr.X: 154011409-154011409 on Build GRCh38
rs1024859110 Chr.X: 154011233-154011233 on Build GRCh38
rs144858531 Chr.X: 154011234-154011234 on Build GRCh38

View Full Product Details