Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154011386-154011891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK1
Entrez Gene ID
3654
Gene Name
interleukin 1 receptor associated kinase 1
Gene Aliases
IRAK, pelle
UniGene
Hs.522819
Gene Chromosome Location
Chr.X: 154010506-154019906 on Build GRCh38
Transcripts
NM_001025242.1 NM_001025243.1 NM_001569.3 XM_005274668.3 XM_011531158.2

SNP Detail

SNP ID Chromosome Location
rs3027901 Chr.X: 154011829-154011829 on Build GRCh38
rs782406832 Chr.X: 154011711-154011711 on Build GRCh38
rs782019810 Chr.X: 154011754-154011754 on Build GRCh38
rs781970018 Chr.X: 154011799-154011799 on Build GRCh38
rs782256946 Chr.X: 154011764-154011764 on Build GRCh38
rs781935869 Chr.X: 154011714-154011714 on Build GRCh38
rs1027800961 Chr.X: 154011854-154011854 on Build GRCh38
rs782222241 Chr.X: 154011685-154011685 on Build GRCh38
rs781912525 Chr.X: 154011833-154011833 on Build GRCh38
rs781998565 Chr.X: 154011690-154011690 on Build GRCh38
rs782199923 Chr.X: 154011788-154011788 on Build GRCh38
rs782327366 Chr.X: 154011741-154011741 on Build GRCh38
rs372331737 Chr.X: 154011721-154011721 on Build GRCh38
rs781826721 Chr.X: 154011841-154011841 on Build GRCh38
rs782579760 Chr.X: 154011743-154011743 on Build GRCh38
rs894586240 Chr.X: 154011767-154011767 on Build GRCh38
rs781993534 Chr.X: 154011553-154011553 on Build GRCh38
rs782428563 Chr.X: 154011488-154011488 on Build GRCh38
rs782090043 Chr.X: 154011802-154011802 on Build GRCh38
rs782574273 Chr.X: 154011675-154011675 on Build GRCh38
rs952410596 Chr.X: 154011582-154011582 on Build GRCh38
rs373959036 Chr.X: 154011615-154011615 on Build GRCh38
rs782674809 Chr.X: 154011787-154011787 on Build GRCh38
rs781786701 Chr.X: 154011748-154011748 on Build GRCh38
rs1027853408 Chr.X: 154011578-154011578 on Build GRCh38
rs367879651 Chr.X: 154011742-154011742 on Build GRCh38
rs922539349 Chr.X: 154011723-154011723 on Build GRCh38
rs782514504 Chr.X: 154011842-154011842 on Build GRCh38
rs781995118 Chr.X: 154011839-154011839 on Build GRCh38
rs781828069 Chr.X: 154011720-154011720 on Build GRCh38
rs782399705 Chr.X: 154011814-154011814 on Build GRCh38
rs191455046 Chr.X: 154011688-154011688 on Build GRCh38
rs962647785 Chr.X: 154011502-154011502 on Build GRCh38
rs1014887397 Chr.X: 154011436-154011436 on Build GRCh38
rs185172267 Chr.X: 154011450-154011450 on Build GRCh38
rs782283731 Chr.X: 154011795-154011795 on Build GRCh38
rs782367305 Chr.X: 154011769-154011769 on Build GRCh38
rs781893812 Chr.X: 154011432-154011432 on Build GRCh38
rs782486025 Chr.X: 154011750-154011750 on Build GRCh38
rs781990807 Chr.X: 154011815-154011815 on Build GRCh38
rs781966531 Chr.X: 154011648-154011648 on Build GRCh38
rs782116854 Chr.X: 154011699-154011699 on Build GRCh38
rs11556422 Chr.X: 154011516-154011516 on Build GRCh38
rs3027900 Chr.X: 154011717-154011717 on Build GRCh38
rs983888763 Chr.X: 154011606-154011606 on Build GRCh38
rs782778618 Chr.X: 154011828-154011828 on Build GRCh38
rs782628025 Chr.X: 154011562-154011562 on Build GRCh38
rs894480859 Chr.X: 154011409-154011409 on Build GRCh38
rs782169186 Chr.X: 154011820-154011820 on Build GRCh38

View Full Product Details