Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154013320-154013829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRAK1
Entrez Gene ID
3654
Gene Name
interleukin 1 receptor associated kinase 1
Gene Aliases
IRAK, pelle
UniGene
Hs.522819
Gene Chromosome Location
Chr.X: 154010506-154019906 on Build GRCh38
Transcripts
NM_001025242.1 NM_001025243.1 NM_001569.3 XM_005274668.3 XM_011531158.2

SNP Detail

SNP ID Chromosome Location
rs150089165 Chr.X: 154013347-154013347 on Build GRCh38
rs782152214 Chr.X: 154013427-154013427 on Build GRCh38
rs782540043 Chr.X: 154013482-154013482 on Build GRCh38
rs933617119 Chr.X: 154013475-154013475 on Build GRCh38
rs1016760551 Chr.X: 154013750-154013750 on Build GRCh38
rs782767944 Chr.X: 154013416-154013416 on Build GRCh38
rs144148891 Chr.X: 154013736-154013736 on Build GRCh38
rs782498713 Chr.X: 154013453-154013453 on Build GRCh38
rs781882635 Chr.X: 154013746-154013746 on Build GRCh38
rs1002802175 Chr.X: 154013700-154013700 on Build GRCh38
rs974811749 Chr.X: 154013386-154013386 on Build GRCh38
rs782718514 Chr.X: 154013431-154013431 on Build GRCh38
rs964848327 Chr.X: 154013801-154013801 on Build GRCh38
rs1012993209 Chr.X: 154013603-154013603 on Build GRCh38
rs924130150 Chr.X: 154013500-154013500 on Build GRCh38
rs187973801 Chr.X: 154013563-154013563 on Build GRCh38
rs1059703 Chr.X: 154013378-154013378 on Build GRCh38
rs781791980 Chr.X: 154013441-154013441 on Build GRCh38
rs781960874 Chr.X: 154013401-154013401 on Build GRCh38
rs11556420 Chr.X: 154013424-154013424 on Build GRCh38
rs782476487 Chr.X: 154013770-154013770 on Build GRCh38
rs923501483 Chr.X: 154013474-154013474 on Build GRCh38
rs781889502 Chr.X: 154013638-154013638 on Build GRCh38
rs782043087 Chr.X: 154013395-154013395 on Build GRCh38
rs111637323 Chr.X: 154013578-154013578 on Build GRCh38
rs781931439 Chr.X: 154013390-154013390 on Build GRCh38
rs907098777 Chr.X: 154013697-154013697 on Build GRCh38
rs782545852 Chr.X: 154013731-154013731 on Build GRCh38
rs3027906 Chr.X: 154013520-154013520 on Build GRCh38
rs368063747 Chr.X: 154013385-154013385 on Build GRCh38
rs782346380 Chr.X: 154013397-154013397 on Build GRCh38
rs371569686 Chr.X: 154013407-154013407 on Build GRCh38
rs1054139830 Chr.X: 154013523-154013523 on Build GRCh38
rs781952209 Chr.X: 154013519-154013519 on Build GRCh38
rs782777000 Chr.X: 154013483-154013483 on Build GRCh38
rs781911608 Chr.X: 154013418-154013418 on Build GRCh38
rs781854558 Chr.X: 154013481-154013481 on Build GRCh38
rs975136768 Chr.X: 154013803-154013803 on Build GRCh38
rs1006093510 Chr.X: 154013748-154013748 on Build GRCh38
rs782290230 Chr.X: 154013526-154013526 on Build GRCh38
rs782639963 Chr.X: 154013476-154013476 on Build GRCh38

View Full Product Details