Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154431699-154432198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP1
Entrez Gene ID
537
Gene Name
ATPase H+ transporting accessory protein 1
Gene Aliases
16A, ATP6IP1, ATP6S1, Ac45, CF2, VATPS1, XAP-3, XAP3
UniGene
Hs.633817
Gene Chromosome Location
Chr.X: 154428632-154436517 on Build GRCh38
Transcripts
NM_001183.5 XM_011531179.1

SNP Detail

SNP ID Chromosome Location
rs782254911 Chr.X: 154431807-154431807 on Build GRCh38
rs1005586818 Chr.X: 154431819-154431819 on Build GRCh38
rs782574022 Chr.X: 154431722-154431722 on Build GRCh38
rs1046310918 Chr.X: 154432037-154432037 on Build GRCh38
rs782687619 Chr.X: 154431791-154431791 on Build GRCh38
rs188599778 Chr.X: 154432017-154432017 on Build GRCh38
rs782743255 Chr.X: 154432079-154432079 on Build GRCh38
rs782310185 Chr.X: 154431946-154431946 on Build GRCh38
rs782629683 Chr.X: 154431954-154431954 on Build GRCh38
rs781978458 Chr.X: 154432085-154432085 on Build GRCh38
rs781786319 Chr.X: 154431732-154431732 on Build GRCh38
rs948008046 Chr.X: 154432034-154432034 on Build GRCh38
rs3737557 Chr.X: 154431724-154431724 on Build GRCh38
rs782743847 Chr.X: 154431886-154431886 on Build GRCh38
rs888563405 Chr.X: 154432117-154432117 on Build GRCh38
rs782081475 Chr.X: 154432033-154432033 on Build GRCh38
rs782239813 Chr.X: 154431844-154431844 on Build GRCh38
rs782269208 Chr.X: 154431956-154431956 on Build GRCh38
rs782780282 Chr.X: 154432134-154432134 on Build GRCh38
rs906335347 Chr.X: 154431779-154431779 on Build GRCh38
rs782469174 Chr.X: 154431789-154431789 on Build GRCh38
rs1053854906 Chr.X: 154431733-154431733 on Build GRCh38
rs782256057 Chr.X: 154431754-154431754 on Build GRCh38
rs782538885 Chr.X: 154431788-154431788 on Build GRCh38
rs782606567 Chr.X: 154431860-154431860 on Build GRCh38
rs781908417 Chr.X: 154431922-154431922 on Build GRCh38
rs782435833 Chr.X: 154431929-154431929 on Build GRCh38
rs148803059 Chr.X: 154431902-154431902 on Build GRCh38
rs782291765 Chr.X: 154431887-154431888 on Build GRCh38
rs782275475 Chr.X: 154431977-154431977 on Build GRCh38
rs369607006 Chr.X: 154431834-154431834 on Build GRCh38
rs782275748 Chr.X: 154431926-154431926 on Build GRCh38
rs782664781 Chr.X: 154431920-154431920 on Build GRCh38
rs782201822 Chr.X: 154432018-154432018 on Build GRCh38
rs782061281 Chr.X: 154432098-154432098 on Build GRCh38
rs781941934 Chr.X: 154431949-154431949 on Build GRCh38
rs782133223 Chr.X: 154432111-154432111 on Build GRCh38
rs782547343 Chr.X: 154431952-154431952 on Build GRCh38
rs782161019 Chr.X: 154431852-154431852 on Build GRCh38
rs1046244673 Chr.X: 154431777-154431777 on Build GRCh38
rs375234358 Chr.X: 154431814-154431814 on Build GRCh38
rs782336286 Chr.X: 154431811-154431811 on Build GRCh38
rs782174698 Chr.X: 154431816-154431816 on Build GRCh38
rs782448013 Chr.X: 154431936-154431936 on Build GRCh38
rs199872479 Chr.X: 154431885-154431885 on Build GRCh38
rs371825213 Chr.X: 154431914-154431914 on Build GRCh38
rs782321781 Chr.X: 154431822-154431822 on Build GRCh38
rs781997607 Chr.X: 154432027-154432028 on Build GRCh38
rs782504984 Chr.X: 154431782-154431782 on Build GRCh38
rs190513781 Chr.X: 154431751-154431751 on Build GRCh38
rs781917775 Chr.X: 154432094-154432094 on Build GRCh38
rs782449304 Chr.X: 154431742-154431742 on Build GRCh38
rs782002491 Chr.X: 154431882-154431882 on Build GRCh38
rs782559988 Chr.X: 154431886-154431887 on Build GRCh38
rs781924823 Chr.X: 154431887-154431887 on Build GRCh38
rs1006031978 Chr.X: 154432126-154432126 on Build GRCh38
rs183479139 Chr.X: 154431752-154431752 on Build GRCh38

View Full Product Details