Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154433547-154434043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP1
Entrez Gene ID
537
Gene Name
ATPase H+ transporting accessory protein 1
Gene Aliases
16A, ATP6IP1, ATP6S1, Ac45, CF2, VATPS1, XAP-3, XAP3
UniGene
Hs.633817
Gene Chromosome Location
Chr.X: 154428632-154436517 on Build GRCh38
Transcripts
NM_001183.5 XM_011531179.1

SNP Detail

SNP ID Chromosome Location
rs782667538 Chr.X: 154433966-154433966 on Build GRCh38
rs782295007 Chr.X: 154434002-154434002 on Build GRCh38
rs782686160 Chr.X: 154433875-154433875 on Build GRCh38
rs35734764 Chr.X: 154433751-154433752 on Build GRCh38
rs782271128 Chr.X: 154433709-154433709 on Build GRCh38
rs781976520 Chr.X: 154433572-154433572 on Build GRCh38
rs141473098 Chr.X: 154433992-154433992 on Build GRCh38
rs370990970 Chr.X: 154433752-154433752 on Build GRCh38
rs782436864 Chr.X: 154433673-154433673 on Build GRCh38
rs782696227 Chr.X: 154433646-154433646 on Build GRCh38
rs782138982 Chr.X: 154433630-154433630 on Build GRCh38
rs782597304 Chr.X: 154433692-154433692 on Build GRCh38
rs781836064 Chr.X: 154433672-154433672 on Build GRCh38
rs1056188193 Chr.X: 154433953-154433953 on Build GRCh38
rs191969885 Chr.X: 154433832-154433832 on Build GRCh38
rs782518476 Chr.X: 154433652-154433652 on Build GRCh38
rs782057748 Chr.X: 154433577-154433577 on Build GRCh38
rs782097620 Chr.X: 154433863-154433863 on Build GRCh38
rs782642386 Chr.X: 154433708-154433708 on Build GRCh38
rs150353253 Chr.X: 154433705-154433705 on Build GRCh38
rs782396742 Chr.X: 154433733-154433733 on Build GRCh38
rs782275544 Chr.X: 154433704-154433704 on Build GRCh38
rs782085826 Chr.X: 154433587-154433587 on Build GRCh38
rs782451012 Chr.X: 154433659-154433659 on Build GRCh38
rs782349780 Chr.X: 154433710-154433710 on Build GRCh38
rs146695597 Chr.X: 154433645-154433645 on Build GRCh38
rs782187940 Chr.X: 154433719-154433719 on Build GRCh38
rs781977533 Chr.X: 154433712-154433712 on Build GRCh38
rs782762748 Chr.X: 154433785-154433785 on Build GRCh38
rs782244743 Chr.X: 154433805-154433805 on Build GRCh38
rs782702726 Chr.X: 154433617-154433617 on Build GRCh38
rs782611558 Chr.X: 154433674-154433674 on Build GRCh38
rs377605162 Chr.X: 154433963-154433963 on Build GRCh38
rs782728896 Chr.X: 154433656-154433656 on Build GRCh38
rs1003302630 Chr.X: 154433907-154433907 on Build GRCh38

View Full Product Details