Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154435167-154435671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP1
Entrez Gene ID
537
Gene Name
ATPase H+ transporting accessory protein 1
Gene Aliases
16A, ATP6IP1, ATP6S1, Ac45, CF2, VATPS1, XAP-3, XAP3
UniGene
Hs.633817
Gene Chromosome Location
Chr.X: 154428632-154436517 on Build GRCh38
Transcripts
NM_001183.5 XM_011531179.1

SNP Detail

SNP ID Chromosome Location
rs376661461 Chr.X: 154435518-154435518 on Build GRCh38
rs781834817 Chr.X: 154435199-154435199 on Build GRCh38
rs372228206 Chr.X: 154435436-154435436 on Build GRCh38
rs782455566 Chr.X: 154435538-154435538 on Build GRCh38
rs782243961 Chr.X: 154435293-154435293 on Build GRCh38
rs782286220 Chr.X: 154435471-154435471 on Build GRCh38
rs782819704 Chr.X: 154435352-154435352 on Build GRCh38
rs782330877 Chr.X: 154435291-154435291 on Build GRCh38
rs782362083 Chr.X: 154435474-154435474 on Build GRCh38
rs200315745 Chr.X: 154435459-154435459 on Build GRCh38
rs1049404900 Chr.X: 154435612-154435612 on Build GRCh38
rs782183151 Chr.X: 154435424-154435424 on Build GRCh38
rs782690014 Chr.X: 154435332-154435332 on Build GRCh38
rs782447719 Chr.X: 154435207-154435207 on Build GRCh38
rs781993994 Chr.X: 154435508-154435508 on Build GRCh38
rs782186939 Chr.X: 154435227-154435228 on Build GRCh38
rs781850155 Chr.X: 154435403-154435403 on Build GRCh38
rs878853277 Chr.X: 154435338-154435338 on Build GRCh38
rs781909953 Chr.X: 154435620-154435620 on Build GRCh38
rs373029897 Chr.X: 154435536-154435536 on Build GRCh38
rs782533572 Chr.X: 154435515-154435515 on Build GRCh38
rs890764219 Chr.X: 154435644-154435644 on Build GRCh38
rs782384749 Chr.X: 154435312-154435312 on Build GRCh38
rs782248279 Chr.X: 154435267-154435267 on Build GRCh38
rs200598349 Chr.X: 154435465-154435465 on Build GRCh38
rs973971109 Chr.X: 154435489-154435489 on Build GRCh38
rs782048010 Chr.X: 154435611-154435611 on Build GRCh38
rs1012080406 Chr.X: 154435235-154435235 on Build GRCh38
rs782744522 Chr.X: 154435197-154435197 on Build GRCh38
rs919714787 Chr.X: 154435514-154435514 on Build GRCh38
rs371934768 Chr.X: 154435260-154435260 on Build GRCh38
rs781865181 Chr.X: 154435519-154435519 on Build GRCh38
rs782644354 Chr.X: 154435455-154435455 on Build GRCh38
rs143004832 Chr.X: 154435313-154435313 on Build GRCh38
rs782662082 Chr.X: 154435540-154435540 on Build GRCh38
rs782284513 Chr.X: 154435441-154435441 on Build GRCh38
rs11539773 Chr.X: 154435430-154435430 on Build GRCh38
rs782140448 Chr.X: 154435337-154435337 on Build GRCh38
rs376840212 Chr.X: 154435647-154435647 on Build GRCh38
rs782188842 Chr.X: 154435463-154435463 on Build GRCh38
rs782708418 Chr.X: 154435502-154435502 on Build GRCh38
rs368929031 Chr.X: 154435327-154435327 on Build GRCh38
rs369039111 Chr.X: 154435529-154435529 on Build GRCh38
rs781921302 Chr.X: 154435461-154435461 on Build GRCh38
rs782108324 Chr.X: 154435478-154435478 on Build GRCh38
rs782492576 Chr.X: 154435454-154435454 on Build GRCh38
rs782264629 Chr.X: 154435256-154435256 on Build GRCh38
rs782299035 Chr.X: 154435536-154435536 on Build GRCh38
rs782117222 Chr.X: 154435466-154435466 on Build GRCh38
rs782737307 Chr.X: 154435362-154435362 on Build GRCh38
rs782615040 Chr.X: 154435361-154435361 on Build GRCh38
rs781862242 Chr.X: 154435359-154435359 on Build GRCh38
rs782180502 Chr.X: 154435277-154435277 on Build GRCh38
rs781936333 Chr.X: 154435333-154435333 on Build GRCh38
rs782554726 Chr.X: 154435237-154435237 on Build GRCh38
rs375162515 Chr.X: 154435290-154435290 on Build GRCh38

View Full Product Details