Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154677307-154677854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB3
Entrez Gene ID
139716
Gene Name
GRB2 associated binding protein 3
Gene Aliases
-
UniGene
Hs.496982
Gene Chromosome Location
Chr.X: 154675249-154751583 on Build GRCh38
Transcripts
NM_001081573.2 NM_001282283.1 NM_080612.3 NR_104114.1 XM_005274648.1 XM_006724804.1 XM_011531103.1 XM_011531105.1 XM_011531106.1 XM_017029276.1

SNP Detail

SNP ID Chromosome Location
rs141555380 Chr.X: 154677735-154677735 on Build GRCh38
rs1034253836 Chr.X: 154677550-154677550 on Build GRCh38
rs782742034 Chr.X: 154677627-154677627 on Build GRCh38
rs781819647 Chr.X: 154677635-154677635 on Build GRCh38
rs191782113 Chr.X: 154677639-154677639 on Build GRCh38
rs782535702 Chr.X: 154677742-154677742 on Build GRCh38
rs782554128 Chr.X: 154677721-154677721 on Build GRCh38
rs972190622 Chr.X: 154677693-154677693 on Build GRCh38
rs28637142 Chr.X: 154677359-154677359 on Build GRCh38
rs904202957 Chr.X: 154677544-154677544 on Build GRCh38
rs910842431 Chr.X: 154677736-154677736 on Build GRCh38
rs182341863 Chr.X: 154677348-154677348 on Build GRCh38
rs992654743 Chr.X: 154677592-154677592 on Build GRCh38
rs930657386 Chr.X: 154677729-154677729 on Build GRCh38
rs958611953 Chr.X: 154677565-154677565 on Build GRCh38
rs944950136 Chr.X: 154677761-154677761 on Build GRCh38
rs782114360 Chr.X: 154677529-154677529 on Build GRCh38
rs782222243 Chr.X: 154677708-154677708 on Build GRCh38
rs186673379 Chr.X: 154677528-154677528 on Build GRCh38
rs892595686 Chr.X: 154677499-154677499 on Build GRCh38
rs1040558951 Chr.X: 154677410-154677410 on Build GRCh38
rs999805683 Chr.X: 154677549-154677549 on Build GRCh38

View Full Product Details