Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154695835-154696345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB3
Entrez Gene ID
139716
Gene Name
GRB2 associated binding protein 3
Gene Aliases
-
UniGene
Hs.496982
Gene Chromosome Location
Chr.X: 154675249-154751583 on Build GRCh38
Transcripts
NM_001081573.2 NM_001282283.1 NM_080612.3 NR_104114.1 XM_005274648.1 XM_006724804.1 XM_011531103.1 XM_011531105.1 XM_011531106.1 XM_017029276.1

SNP Detail

SNP ID Chromosome Location
rs370569226 Chr.X: 154696040-154696040 on Build GRCh38
rs782210679 Chr.X: 154696035-154696035 on Build GRCh38
rs893613331 Chr.X: 154696110-154696110 on Build GRCh38
rs910053089 Chr.X: 154696263-154696263 on Build GRCh38
rs782788126 Chr.X: 154695924-154695924 on Build GRCh38
rs183011802 Chr.X: 154695907-154695907 on Build GRCh38
rs782602979 Chr.X: 154695948-154695948 on Build GRCh38
rs969419430 Chr.X: 154696178-154696178 on Build GRCh38
rs782473076 Chr.X: 154695938-154695938 on Build GRCh38
rs35105330 Chr.X: 154696298-154696299 on Build GRCh38
rs138941436 Chr.X: 154696029-154696029 on Build GRCh38
rs782633613 Chr.X: 154696033-154696033 on Build GRCh38
rs1031271492 Chr.X: 154696074-154696074 on Build GRCh38
rs983029728 Chr.X: 154696259-154696259 on Build GRCh38
rs782005897 Chr.X: 154696101-154696101 on Build GRCh38
rs782665358 Chr.X: 154696240-154696240 on Build GRCh38
rs1023703724 Chr.X: 154696140-154696140 on Build GRCh38
rs982102880 Chr.X: 154696187-154696187 on Build GRCh38
rs782548838 Chr.X: 154696141-154696141 on Build GRCh38
rs781800638 Chr.X: 154695953-154695953 on Build GRCh38
rs782096395 Chr.X: 154696273-154696273 on Build GRCh38
rs782678443 Chr.X: 154695967-154695967 on Build GRCh38
rs782724992 Chr.X: 154696299-154696299 on Build GRCh38
rs782414307 Chr.X: 154696129-154696129 on Build GRCh38
rs112121559 Chr.X: 154696055-154696055 on Build GRCh38
rs1011438851 Chr.X: 154696139-154696139 on Build GRCh38
rs781950875 Chr.X: 154695873-154695873 on Build GRCh38
rs941524133 Chr.X: 154696279-154696279 on Build GRCh38
rs781785441 Chr.X: 154696126-154696126 on Build GRCh38
rs782069241 Chr.X: 154695885-154695885 on Build GRCh38
rs951610066 Chr.X: 154696246-154696246 on Build GRCh38
rs781866223 Chr.X: 154695937-154695937 on Build GRCh38
rs1037426080 Chr.X: 154695896-154695896 on Build GRCh38
rs782445662 Chr.X: 154695963-154695963 on Build GRCh38
rs900194387 Chr.X: 154695908-154695908 on Build GRCh38

View Full Product Details