Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154886002-154886495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F8
Entrez Gene ID
2157
Gene Name
coagulation factor VIII
Gene Aliases
AHF, DXS1253E, F8B, F8C, FVIII, HEMA
UniGene
Hs.632836
Gene Chromosome Location
Chr.X: 154835788-155022723 on Build GRCh38
Transcripts
NM_000132.3 NM_019863.2 NM_012151.3

Gene Symbol
F8A1
Entrez Gene ID
8263
Gene Name
coagulation factor VIII-associated 1
Gene Aliases
DXS522E, F8A, HAP40
UniGene
Hs.533543
Gene Chromosome Location
Chr.X: 154886360-154888061 on Build GRCh38
Transcripts
NM_000132.3 NM_019863.2 NM_012151.3

SNP Detail

SNP ID Chromosome Location
rs782391117 Chr.X: 154886110-154886110 on Build GRCh38
rs782308962 Chr.X: 154886173-154886173 on Build GRCh38
rs113683431 Chr.X: 154886026-154886026 on Build GRCh38
rs781938341 Chr.X: 154886185-154886185 on Build GRCh38
rs782295624 Chr.X: 154886044-154886044 on Build GRCh38
rs782101726 Chr.X: 154886158-154886158 on Build GRCh38
rs370287354 Chr.X: 154886234-154886234 on Build GRCh38
rs781949659 Chr.X: 154886129-154886129 on Build GRCh38

View Full Product Details