Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58196281-58196781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPMK
Entrez Gene ID
253430
Gene Name
inositol polyphosphate multikinase
Gene Aliases
-
UniGene
Hs.30280
Gene Chromosome Location
Chr.10: 58191517-58269837 on Build GRCh38
Transcripts
NM_152230.4 XM_011539565.2 XM_017016013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
386277 c.824A>T p.E275V Chr.10: 58196503-58196503 on Build GRCh38
7137530 c.809A>G p.D270G Chr.10: 58196518-58196518 on Build GRCh38
5969850 c.820G>A p.A274T Chr.10: 58196507-58196507 on Build GRCh38
3807382 c.906T>G p.A302A Chr.10: 58196421-58196421 on Build GRCh38
391624 c.986delA p.H329fs*7 Chr.10: 58196341-58196341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914004944 Chr.10: 58196689-58196689 on Build GRCh38
rs762465521 Chr.10: 58196697-58196697 on Build GRCh38
rs541806725 Chr.10: 58196401-58196401 on Build GRCh38
rs200416444 Chr.10: 58196547-58196547 on Build GRCh38
rs375383787 Chr.10: 58196371-58196371 on Build GRCh38
rs543365928 Chr.10: 58196673-58196673 on Build GRCh38
rs762937469 Chr.10: 58196713-58196714 on Build GRCh38
rs780067236 Chr.10: 58196444-58196444 on Build GRCh38
rs774063548 Chr.10: 58196481-58196481 on Build GRCh38
rs766769958 Chr.10: 58196715-58196715 on Build GRCh38
rs776881750 Chr.10: 58196462-58196462 on Build GRCh38
rs1044344662 Chr.10: 58196411-58196411 on Build GRCh38
rs770946028 Chr.10: 58196325-58196325 on Build GRCh38
rs759410890 Chr.10: 58196369-58196369 on Build GRCh38
rs775525235 Chr.10: 58196544-58196544 on Build GRCh38
rs767989494 Chr.10: 58196721-58196721 on Build GRCh38
rs749426932 Chr.10: 58196633-58196633 on Build GRCh38
rs768041734 Chr.10: 58196553-58196553 on Build GRCh38
rs148537566 Chr.10: 58196488-58196488 on Build GRCh38
rs61751225 Chr.10: 58196641-58196641 on Build GRCh38
rs772305007 Chr.10: 58196338-58196338 on Build GRCh38
rs777938568 Chr.10: 58196629-58196629 on Build GRCh38
rs768810591 Chr.10: 58196457-58196457 on Build GRCh38
rs772940182 Chr.10: 58196353-58196353 on Build GRCh38
rs988205742 Chr.10: 58196687-58196687 on Build GRCh38
rs201015068 Chr.10: 58196504-58196504 on Build GRCh38
rs766613884 Chr.10: 58196577-58196577 on Build GRCh38
rs142574341 Chr.10: 58196406-58196406 on Build GRCh38
rs370658364 Chr.10: 58196678-58196678 on Build GRCh38
rs752758405 Chr.10: 58196601-58196601 on Build GRCh38
rs535168447 Chr.10: 58196368-58196368 on Build GRCh38
rs762715336 Chr.10: 58196363-58196363 on Build GRCh38
rs752811210 Chr.10: 58196725-58196725 on Build GRCh38
rs900806853 Chr.10: 58196717-58196717 on Build GRCh38
rs764972658 Chr.10: 58196565-58196565 on Build GRCh38
rs748319721 Chr.10: 58196465-58196465 on Build GRCh38
rs766938208 Chr.10: 58196370-58196370 on Build GRCh38
rs202113550 Chr.10: 58196720-58196720 on Build GRCh38
rs776342351 Chr.10: 58196668-58196668 on Build GRCh38
rs185185082 Chr.10: 58196339-58196339 on Build GRCh38
rs913891219 Chr.10: 58196400-58196400 on Build GRCh38
rs1005862697 Chr.10: 58196546-58196546 on Build GRCh38
rs768475708 Chr.10: 58196647-58196647 on Build GRCh38
rs951698763 Chr.10: 58196527-58196527 on Build GRCh38
rs755776415 Chr.10: 58196398-58196398 on Build GRCh38
rs368295594 Chr.10: 58196425-58196425 on Build GRCh38
rs755373508 Chr.10: 58196586-58196586 on Build GRCh38
rs61757082 Chr.10: 58196509-58196509 on Build GRCh38
rs761503472 Chr.10: 58196501-58196501 on Build GRCh38
rs376895646 Chr.10: 58196531-58196531 on Build GRCh38
rs142776806 Chr.10: 58196525-58196525 on Build GRCh38
rs1046822225 Chr.10: 58196749-58196749 on Build GRCh38
rs763347320 Chr.10: 58196570-58196570 on Build GRCh38
rs779660196 Chr.10: 58196642-58196642 on Build GRCh38
rs754291508 Chr.10: 58196413-58196413 on Build GRCh38
rs372263875 Chr.10: 58196520-58196520 on Build GRCh38
rs146319184 Chr.10: 58196431-58196431 on Build GRCh38
rs756213118 Chr.10: 58196740-58196740 on Build GRCh38
rs769612886 Chr.10: 58196674-58196674 on Build GRCh38
rs761628068 Chr.10: 58196562-58196562 on Build GRCh38
rs367920159 Chr.10: 58196536-58196536 on Build GRCh38
rs765751052 Chr.10: 58196343-58196343 on Build GRCh38
rs760196498 Chr.10: 58196494-58196494 on Build GRCh38
rs746395205 Chr.10: 58196428-58196428 on Build GRCh38
rs576272977 Chr.10: 58196580-58196580 on Build GRCh38
rs747099599 Chr.10: 58196452-58196452 on Build GRCh38
rs774909915 Chr.10: 58196493-58196493 on Build GRCh38
rs76733713 Chr.10: 58196455-58196455 on Build GRCh38
rs964659053 Chr.10: 58196548-58196548 on Build GRCh38
rs746974757 Chr.10: 58196645-58196645 on Build GRCh38
rs781190690 Chr.10: 58196530-58196530 on Build GRCh38
rs757723741 Chr.10: 58196415-58196415 on Build GRCh38
rs746340310 Chr.10: 58196324-58196324 on Build GRCh38
rs769997362 Chr.10: 58196533-58196533 on Build GRCh38
rs770725627 Chr.10: 58196472-58196472 on Build GRCh38
rs750816294 Chr.10: 58196505-58196505 on Build GRCh38
rs756374477 Chr.10: 58196616-58196616 on Build GRCh38
rs766345094 Chr.10: 58196365-58196365 on Build GRCh38
rs182040233 Chr.10: 58196543-58196543 on Build GRCh38
rs781314381 Chr.10: 58196597-58196597 on Build GRCh38
rs779527924 Chr.10: 58196309-58196309 on Build GRCh38
rs750867741 Chr.10: 58196567-58196567 on Build GRCh38
rs753571526 Chr.10: 58196499-58196499 on Build GRCh38

View Full Product Details