Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58513230-58513728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICC1
Entrez Gene ID
80114
Gene Name
BicC family RNA binding protein 1
Gene Aliases
BICC, CYSRD
UniGene
Hs.100261
Gene Chromosome Location
Chr.10: 58512970-58831437 on Build GRCh38
Transcripts
NM_001080512.2 XM_011540185.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611475 c.121C>A p.L41M Chr.10: 58513264-58513264 on Build GRCh38
4573680 c.182T>C p.M61T Chr.10: 58513325-58513325 on Build GRCh38
5360191 c.87C>A p.P29P Chr.10: 58513230-58513230 on Build GRCh38
1246100 c.143A>C p.E48A Chr.10: 58513286-58513286 on Build GRCh38
1246101 c.149G>A p.R50H Chr.10: 58513292-58513292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960126438 Chr.10: 58513525-58513525 on Build GRCh38
rs1025276939 Chr.10: 58513452-58513452 on Build GRCh38
rs900901566 Chr.10: 58513647-58513647 on Build GRCh38
rs1046692947 Chr.10: 58513666-58513666 on Build GRCh38
rs141140671 Chr.10: 58513305-58513305 on Build GRCh38
rs921766642 Chr.10: 58513251-58513251 on Build GRCh38
rs565122891 Chr.10: 58513567-58513567 on Build GRCh38
rs199873943 Chr.10: 58513355-58513355 on Build GRCh38
rs1023517177 Chr.10: 58513682-58513682 on Build GRCh38
rs778387594 Chr.10: 58513625-58513625 on Build GRCh38
rs765473594 Chr.10: 58513352-58513352 on Build GRCh38
rs543353661 Chr.10: 58513498-58513498 on Build GRCh38
rs756936659 Chr.10: 58513558-58513558 on Build GRCh38
rs7100474 Chr.10: 58513361-58513361 on Build GRCh38
rs182394260 Chr.10: 58513578-58513578 on Build GRCh38
rs888347685 Chr.10: 58513459-58513459 on Build GRCh38
rs369591398 Chr.10: 58513323-58513323 on Build GRCh38
rs537799012 Chr.10: 58513324-58513324 on Build GRCh38
rs767927498 Chr.10: 58513476-58513476 on Build GRCh38
rs778725648 Chr.10: 58513372-58513372 on Build GRCh38
rs754126869 Chr.10: 58513249-58513249 on Build GRCh38
rs369199088 Chr.10: 58513343-58513343 on Build GRCh38
rs1004640288 Chr.10: 58513672-58513672 on Build GRCh38
rs372711421 Chr.10: 58513347-58513347 on Build GRCh38
rs903823349 Chr.10: 58513357-58513357 on Build GRCh38
rs555230848 Chr.10: 58513255-58513255 on Build GRCh38
rs748986819 Chr.10: 58513292-58513292 on Build GRCh38
rs867134439 Chr.10: 58513461-58513461 on Build GRCh38
rs751379394 Chr.10: 58513273-58513273 on Build GRCh38
rs764122252 Chr.10: 58513342-58513342 on Build GRCh38
rs762179200 Chr.10: 58513252-58513257 on Build GRCh38
rs10571555 Chr.10: 58513430-58513433 on Build GRCh38
rs996240579 Chr.10: 58513688-58513688 on Build GRCh38
rs200248207 Chr.10: 58513274-58513274 on Build GRCh38
rs561519989 Chr.10: 58513513-58513513 on Build GRCh38
rs1023884899 Chr.10: 58513482-58513482 on Build GRCh38
rs774094478 Chr.10: 58513298-58513298 on Build GRCh38
rs912679976 Chr.10: 58513514-58513514 on Build GRCh38
rs890000259 Chr.10: 58513427-58513427 on Build GRCh38
rs7078900 Chr.10: 58513670-58513670 on Build GRCh38
rs776017402 Chr.10: 58513705-58513705 on Build GRCh38
rs1040736695 Chr.10: 58513439-58513439 on Build GRCh38
rs969281471 Chr.10: 58513507-58513507 on Build GRCh38
rs1043600018 Chr.10: 58513416-58513416 on Build GRCh38
rs559953643 Chr.10: 58513650-58513650 on Build GRCh38
rs1005873256 Chr.10: 58513677-58513677 on Build GRCh38
rs7079648 Chr.10: 58513395-58513395 on Build GRCh38
rs761833071 Chr.10: 58513348-58513348 on Build GRCh38
rs981947396 Chr.10: 58513510-58513510 on Build GRCh38
rs543766542 Chr.10: 58513564-58513564 on Build GRCh38
rs753892784 Chr.10: 58513344-58513344 on Build GRCh38
rs921836643 Chr.10: 58513497-58513497 on Build GRCh38
rs372002626 Chr.10: 58513605-58513605 on Build GRCh38
rs905128626 Chr.10: 58513701-58513701 on Build GRCh38
rs769244104 Chr.10: 58513281-58513281 on Build GRCh38
rs548866269 Chr.10: 58513707-58513707 on Build GRCh38
rs777329269 Chr.10: 58513287-58513287 on Build GRCh38
rs189341200 Chr.10: 58513571-58513571 on Build GRCh38
rs143335902 Chr.10: 58513543-58513543 on Build GRCh38
rs71033620 Chr.10: 58513437-58513440 on Build GRCh38
rs775922178 Chr.10: 58513322-58513322 on Build GRCh38
rs554454704 Chr.10: 58513409-58513409 on Build GRCh38
rs912646689 Chr.10: 58513321-58513321 on Build GRCh38
rs779873399 Chr.10: 58513266-58513266 on Build GRCh38
rs1055344443 Chr.10: 58513606-58513606 on Build GRCh38
rs1051212806 Chr.10: 58513314-58513314 on Build GRCh38
rs370077280 Chr.10: 58513362-58513362 on Build GRCh38
rs572667497 Chr.10: 58513437-58513437 on Build GRCh38
rs947693321 Chr.10: 58513405-58513405 on Build GRCh38
rs754740911 Chr.10: 58513354-58513354 on Build GRCh38
rs993769810 Chr.10: 58513450-58513450 on Build GRCh38
rs954564744 Chr.10: 58513504-58513504 on Build GRCh38
rs770680277 Chr.10: 58513293-58513293 on Build GRCh38
rs759986530 Chr.10: 58513299-58513299 on Build GRCh38
rs780995274 Chr.10: 58513276-58513276 on Build GRCh38
rs745504989 Chr.10: 58513376-58513376 on Build GRCh38
rs980248083 Chr.10: 58513254-58513254 on Build GRCh38
rs547568860 Chr.10: 58513626-58513626 on Build GRCh38
rs900932663 Chr.10: 58513441-58513441 on Build GRCh38
rs748825695 Chr.10: 58513363-58513363 on Build GRCh38

View Full Product Details