Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58784898-58785418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICC1
Entrez Gene ID
80114
Gene Name
BicC family RNA binding protein 1
Gene Aliases
BICC, CYSRD
UniGene
Hs.100261
Gene Chromosome Location
Chr.10: 58512970-58831437 on Build GRCh38
Transcripts
NM_001080512.2 XM_005270169.4 XM_011540185.2 XM_011540188.2 XM_011540189.2 XM_011540190.2 XM_011540191.2 XM_017016677.1 XM_017016678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1240892 c.313C>A p.H105N Chr.10: 58785006-58785006 on Build GRCh38
1184696 c.320A>C p.K107T Chr.10: 58785013-58785013 on Build GRCh38
2148185 c.315T>C p.H105H Chr.10: 58785008-58785008 on Build GRCh38
919156 c.326C>A p.S109Y Chr.10: 58785019-58785019 on Build GRCh38
5007041 c.361A>T p.M121L Chr.10: 58785054-58785054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11006261 Chr.10: 58785301-58785301 on Build GRCh38
rs549855727 Chr.10: 58785373-58785373 on Build GRCh38
rs1031526797 Chr.10: 58785136-58785136 on Build GRCh38
rs778832785 Chr.10: 58785081-58785081 on Build GRCh38
rs867771558 Chr.10: 58785174-58785174 on Build GRCh38
rs929476502 Chr.10: 58784975-58784975 on Build GRCh38
rs776548074 Chr.10: 58784985-58784985 on Build GRCh38
rs12356664 Chr.10: 58785285-58785285 on Build GRCh38
rs895354836 Chr.10: 58785282-58785282 on Build GRCh38
rs573800211 Chr.10: 58785165-58785165 on Build GRCh38
rs931903605 Chr.10: 58785169-58785169 on Build GRCh38
rs781582242 Chr.10: 58785032-58785032 on Build GRCh38
rs778345096 Chr.10: 58785055-58785055 on Build GRCh38
rs1034397630 Chr.10: 58785279-58785279 on Build GRCh38
rs761169936 Chr.10: 58785364-58785364 on Build GRCh38
rs117782728 Chr.10: 58785326-58785326 on Build GRCh38
rs114251685 Chr.10: 58785367-58785367 on Build GRCh38
rs950911995 Chr.10: 58785289-58785289 on Build GRCh38
rs772491320 Chr.10: 58785304-58785304 on Build GRCh38
rs960031689 Chr.10: 58784942-58784942 on Build GRCh38
rs983563559 Chr.10: 58785293-58785293 on Build GRCh38
rs895407187 Chr.10: 58785044-58785044 on Build GRCh38
rs913690975 Chr.10: 58785120-58785132 on Build GRCh38
rs749748141 Chr.10: 58785060-58785060 on Build GRCh38
rs573010214 Chr.10: 58785076-58785076 on Build GRCh38
rs560925833 Chr.10: 58785361-58785361 on Build GRCh38
rs891867898 Chr.10: 58785338-58785338 on Build GRCh38
rs536345560 Chr.10: 58785129-58785129 on Build GRCh38
rs868026674 Chr.10: 58785359-58785359 on Build GRCh38
rs74423803 Chr.10: 58785339-58785339 on Build GRCh38
rs1047999068 Chr.10: 58784990-58784990 on Build GRCh38
rs756427798 Chr.10: 58785047-58785047 on Build GRCh38
rs931442607 Chr.10: 58785268-58785268 on Build GRCh38
rs987834013 Chr.10: 58785175-58785175 on Build GRCh38
rs533560347 Chr.10: 58785287-58785287 on Build GRCh38
rs113800716 Chr.10: 58785362-58785362 on Build GRCh38
rs898936036 Chr.10: 58785263-58785263 on Build GRCh38
rs878885102 Chr.10: 58785234-58785234 on Build GRCh38
rs746145409 Chr.10: 58785088-58785088 on Build GRCh38
rs768616894 Chr.10: 58784981-58784981 on Build GRCh38
rs909485248 Chr.10: 58785295-58785295 on Build GRCh38
rs918330487 Chr.10: 58785287-58785288 on Build GRCh38
rs771051560 Chr.10: 58785187-58785187 on Build GRCh38
rs540236147 Chr.10: 58785126-58785126 on Build GRCh38
rs568696613 Chr.10: 58784974-58784974 on Build GRCh38
rs911824464 Chr.10: 58785390-58785390 on Build GRCh38
rs554300271 Chr.10: 58785148-58785148 on Build GRCh38
rs372045739 Chr.10: 58785029-58785029 on Build GRCh38
rs1025675101 Chr.10: 58785291-58785291 on Build GRCh38
rs1010656328 Chr.10: 58785349-58785349 on Build GRCh38
rs1021576792 Chr.10: 58785206-58785206 on Build GRCh38
rs559764770 Chr.10: 58784933-58784933 on Build GRCh38
rs539392881 Chr.10: 58784991-58784991 on Build GRCh38
rs189890114 Chr.10: 58785264-58785264 on Build GRCh38
rs1031054461 Chr.10: 58785329-58785329 on Build GRCh38
rs995701758 Chr.10: 58785022-58785022 on Build GRCh38
rs1052314520 Chr.10: 58785330-58785330 on Build GRCh38
rs957795762 Chr.10: 58785116-58785116 on Build GRCh38
rs75436133 Chr.10: 58785352-58785352 on Build GRCh38
rs1048595464 Chr.10: 58785274-58785274 on Build GRCh38
rs971167370 Chr.10: 58785114-58785114 on Build GRCh38
rs911554938 Chr.10: 58785210-58785210 on Build GRCh38
rs375158306 Chr.10: 58785104-58785104 on Build GRCh38
rs921819039 Chr.10: 58785163-58785163 on Build GRCh38
rs1034270557 Chr.10: 58785380-58785380 on Build GRCh38
rs79744401 Chr.10: 58785391-58785391 on Build GRCh38
rs774695094 Chr.10: 58784960-58784960 on Build GRCh38
rs761652604 Chr.10: 58784988-58784988 on Build GRCh38
rs927083859 Chr.10: 58784941-58784941 on Build GRCh38
rs750263349 Chr.10: 58784989-58784989 on Build GRCh38
rs957843089 Chr.10: 58785144-58785144 on Build GRCh38
rs558031043 Chr.10: 58785020-58785020 on Build GRCh38
rs992348854 Chr.10: 58784948-58784948 on Build GRCh38
rs897310594 Chr.10: 58785009-58785009 on Build GRCh38
rs555365290 Chr.10: 58785143-58785143 on Build GRCh38
rs753079034 Chr.10: 58785034-58785034 on Build GRCh38
rs749291082 Chr.10: 58785110-58785110 on Build GRCh38
rs765791783 Chr.10: 58784997-58784997 on Build GRCh38
rs1036097084 Chr.10: 58785244-58785244 on Build GRCh38
rs775814783 Chr.10: 58785107-58785107 on Build GRCh38
rs940381417 Chr.10: 58785231-58785231 on Build GRCh38

View Full Product Details